Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61757208-61757728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZP434K028
Entrez Gene ID
26070
Gene Name
uncharacterized LOC26070
Gene Aliases
-
UniGene
Hs.564254
Gene Chromosome Location
Chr.11: 61754029-61757664 on Build GRCh38
Transcripts
NR_026882.1 NM_001127392.2 NM_013279.3 XM_005274222.1 XM_005274223.1 XM_005274224.1 XM_005274225.1 XM_005274226.1 XM_005274227.1 XM_005274228.1 XM_011545234.2

Gene Symbol
MYRF
Entrez Gene ID
745
Gene Name
myelin regulatory factor
Gene Aliases
C11orf9, MRF, Ndt80, pqn-47
UniGene
Hs.473109
Gene Chromosome Location
Chr.11: 61752617-61788518 on Build GRCh38
Transcripts
NR_026882.1 NM_001127392.2 NM_013279.3 XM_005274222.1 XM_005274223.1 XM_005274224.1 XM_005274225.1 XM_005274226.1 XM_005274227.1 XM_005274228.1 XM_011545234.2

SNP Detail

SNP ID Chromosome Location
rs761387890 Chr.11: 61757265-61757265 on Build GRCh38
rs1047051872 Chr.11: 61757234-61757234 on Build GRCh38
rs1043187814 Chr.11: 61757624-61757624 on Build GRCh38
rs557380012 Chr.11: 61757680-61757680 on Build GRCh38
rs867291974 Chr.11: 61757531-61757531 on Build GRCh38
rs1003286533 Chr.11: 61757287-61757287 on Build GRCh38
rs953281625 Chr.11: 61757336-61757337 on Build GRCh38
rs541361421 Chr.11: 61757424-61757424 on Build GRCh38
rs117536695 Chr.11: 61757574-61757574 on Build GRCh38
rs1013522585 Chr.11: 61757253-61757253 on Build GRCh38
rs903344554 Chr.11: 61757688-61757688 on Build GRCh38
rs567571087 Chr.11: 61757666-61757666 on Build GRCh38
rs774057156 Chr.11: 61757405-61757405 on Build GRCh38
rs1010990380 Chr.11: 61757627-61757627 on Build GRCh38
rs748831110 Chr.11: 61757329-61757329 on Build GRCh38
rs1035939511 Chr.11: 61757372-61757372 on Build GRCh38
rs907367751 Chr.11: 61757522-61757522 on Build GRCh38
rs377576348 Chr.11: 61757683-61757683 on Build GRCh38
rs198459 Chr.11: 61757548-61757548 on Build GRCh38
rs747603606 Chr.11: 61757229-61757229 on Build GRCh38
rs527260618 Chr.11: 61757601-61757601 on Build GRCh38
rs999000498 Chr.11: 61757698-61757698 on Build GRCh38
rs562605356 Chr.11: 61757602-61757602 on Build GRCh38
rs767347456 Chr.11: 61757409-61757409 on Build GRCh38
rs570927327 Chr.11: 61757614-61757614 on Build GRCh38
rs558576656 Chr.11: 61757354-61757354 on Build GRCh38
rs969550889 Chr.11: 61757402-61757402 on Build GRCh38
rs772989481 Chr.11: 61757454-61757454 on Build GRCh38
rs545055266 Chr.11: 61757332-61757332 on Build GRCh38
rs198460 Chr.11: 61757502-61757502 on Build GRCh38
rs1039740789 Chr.11: 61757592-61757592 on Build GRCh38
rs754871461 Chr.11: 61757618-61757618 on Build GRCh38
rs879636785 Chr.11: 61757652-61757652 on Build GRCh38
rs991803986 Chr.11: 61757382-61757382 on Build GRCh38
rs906291354 Chr.11: 61757272-61757272 on Build GRCh38
rs1039792855 Chr.11: 61757294-61757294 on Build GRCh38
rs1008261219 Chr.11: 61757279-61757279 on Build GRCh38
rs201312715 Chr.11: 61757692-61757692 on Build GRCh38
rs999513698 Chr.11: 61757368-61757368 on Build GRCh38
rs35607948 Chr.11: 61757686-61757686 on Build GRCh38
rs930650214 Chr.11: 61757432-61757432 on Build GRCh38
rs753965339 Chr.11: 61757486-61757486 on Build GRCh38
rs766130038 Chr.11: 61757558-61757558 on Build GRCh38
rs769143992 Chr.11: 61757249-61757249 on Build GRCh38
rs571303456 Chr.11: 61757594-61757594 on Build GRCh38
rs1030712358 Chr.11: 61757289-61757289 on Build GRCh38
rs758816566 Chr.11: 61757562-61757562 on Build GRCh38
rs956556940 Chr.11: 61757311-61757311 on Build GRCh38
rs757792091 Chr.11: 61757599-61757599 on Build GRCh38
rs561225633 Chr.11: 61757481-61757481 on Build GRCh38
rs919751791 Chr.11: 61757473-61757473 on Build GRCh38
rs544011389 Chr.11: 61757487-61757487 on Build GRCh38
rs894700189 Chr.11: 61757252-61757252 on Build GRCh38
rs572100327 Chr.11: 61757362-61757362 on Build GRCh38
rs759834141 Chr.11: 61757512-61757512 on Build GRCh38
rs533691050 Chr.11: 61757651-61757651 on Build GRCh38
rs973893253 Chr.11: 61757426-61757426 on Build GRCh38
rs775085264 Chr.11: 61757266-61757266 on Build GRCh38
rs760699774 Chr.11: 61757477-61757477 on Build GRCh38
rs775302734 Chr.11: 61757400-61757400 on Build GRCh38
rs1022220324 Chr.11: 61757348-61757348 on Build GRCh38
rs530302731 Chr.11: 61757482-61757482 on Build GRCh38
rs929884394 Chr.11: 61757233-61757233 on Build GRCh38
rs753880565 Chr.11: 61757509-61757510 on Build GRCh38
rs868034792 Chr.11: 61757465-61757465 on Build GRCh38
rs761581736 Chr.11: 61757408-61757408 on Build GRCh38
rs767019578 Chr.11: 61757342-61757342 on Build GRCh38
rs916301844 Chr.11: 61757385-61757385 on Build GRCh38
rs960341931 Chr.11: 61757374-61757374 on Build GRCh38

View Full Product Details