Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65424522-65425021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEAT1
Entrez Gene ID
283131
Gene Name
nuclear paraspeckle assembly transcript 1 (non-protein coding)
Gene Aliases
LINC00084, NCRNA00084, TncRNA, VINC
UniGene
Hs.523789
Gene Chromosome Location
Chr.11: 65422798-65445540 on Build GRCh38
Transcripts
NR_028272.1 NR_131012.1

SNP Detail

SNP ID Chromosome Location
rs965609550 Chr.11: 65424936-65424937 on Build GRCh38
rs868576943 Chr.11: 65424894-65424894 on Build GRCh38
rs559633501 Chr.11: 65424798-65424798 on Build GRCh38
rs894840258 Chr.11: 65424773-65424773 on Build GRCh38
rs1038322508 Chr.11: 65424787-65424787 on Build GRCh38
rs191394840 Chr.11: 65424907-65424907 on Build GRCh38
rs955450900 Chr.11: 65424862-65424862 on Build GRCh38
rs1002280235 Chr.11: 65424858-65424858 on Build GRCh38
rs965168995 Chr.11: 65424765-65424765 on Build GRCh38
rs1009774347 Chr.11: 65424652-65424652 on Build GRCh38
rs967608069 Chr.11: 65424969-65424969 on Build GRCh38
rs1033699987 Chr.11: 65424685-65424686 on Build GRCh38
rs975185954 Chr.11: 65424987-65424987 on Build GRCh38
rs921486353 Chr.11: 65424581-65424581 on Build GRCh38
rs914166141 Chr.11: 65424809-65424809 on Build GRCh38
rs561355953 Chr.11: 65424864-65424864 on Build GRCh38
rs746681072 Chr.11: 65424565-65424565 on Build GRCh38
rs1019429540 Chr.11: 65424933-65424933 on Build GRCh38
rs1029912244 Chr.11: 65424853-65424853 on Build GRCh38
rs541143543 Chr.11: 65424741-65424741 on Build GRCh38
rs906583233 Chr.11: 65424827-65424827 on Build GRCh38
rs1020471344 Chr.11: 65424949-65424949 on Build GRCh38
rs576326652 Chr.11: 65424642-65424642 on Build GRCh38
rs767750452 Chr.11: 65424908-65424908 on Build GRCh38
rs1026937204 Chr.11: 65424702-65424702 on Build GRCh38
rs1002592156 Chr.11: 65424667-65424667 on Build GRCh38
rs1005452137 Chr.11: 65424783-65424783 on Build GRCh38
rs775061847 Chr.11: 65424822-65424822 on Build GRCh38
rs951159176 Chr.11: 65424707-65424708 on Build GRCh38
rs906622305 Chr.11: 65424659-65424659 on Build GRCh38
rs1055239978 Chr.11: 65424863-65424864 on Build GRCh38
rs1028065032 Chr.11: 65424992-65424992 on Build GRCh38
rs151283333 Chr.11: 65424814-65424814 on Build GRCh38
rs770651035 Chr.11: 65424651-65424651 on Build GRCh38
rs77364365 Chr.11: 65424698-65424698 on Build GRCh38
rs1053986967 Chr.11: 65424636-65424636 on Build GRCh38
rs142359507 Chr.11: 65424654-65424654 on Build GRCh38
rs911221246 Chr.11: 65424586-65424586 on Build GRCh38
rs556180460 Chr.11: 65424608-65424608 on Build GRCh38
rs927698045 Chr.11: 65424733-65424733 on Build GRCh38
rs975595294 Chr.11: 65424768-65424768 on Build GRCh38
rs994846486 Chr.11: 65424870-65424870 on Build GRCh38
rs140061339 Chr.11: 65424588-65424588 on Build GRCh38
rs376648561 Chr.11: 65424600-65424600 on Build GRCh38
rs1033541250 Chr.11: 65424705-65424705 on Build GRCh38
rs939108151 Chr.11: 65424736-65424736 on Build GRCh38
rs963401577 Chr.11: 65424695-65424695 on Build GRCh38
rs957636027 Chr.11: 65424602-65424602 on Build GRCh38
rs572143293 Chr.11: 65424715-65424715 on Build GRCh38
rs563032755 Chr.11: 65424986-65424986 on Build GRCh38
rs936378494 Chr.11: 65424775-65424775 on Build GRCh38
rs1039387039 Chr.11: 65424546-65424546 on Build GRCh38
rs769222125 Chr.11: 65424734-65424734 on Build GRCh38
rs1018257244 Chr.11: 65424900-65424900 on Build GRCh38
rs1004422744 Chr.11: 65424914-65424914 on Build GRCh38
rs1054793477 Chr.11: 65424757-65424757 on Build GRCh38

View Full Product Details