Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67321070-67321586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100130987
Entrez Gene ID
100130987
Gene Name
uncharacterized LOC100130987
Gene Aliases
-
UniGene
Hs.664585
Gene Chromosome Location
Chr.11: 67317839-67391687 on Build GRCh38
Transcripts
NR_024469.1

SNP Detail

SNP ID Chromosome Location
rs1018183206 Chr.11: 67321534-67321534 on Build GRCh38
rs883189 Chr.11: 67321147-67321147 on Build GRCh38
rs895424836 Chr.11: 67321324-67321324 on Build GRCh38
rs1021495132 Chr.11: 67321140-67321140 on Build GRCh38
rs143479260 Chr.11: 67321212-67321212 on Build GRCh38
rs1038803702 Chr.11: 67321401-67321401 on Build GRCh38
rs941361340 Chr.11: 67321346-67321346 on Build GRCh38
rs750861877 Chr.11: 67321408-67321408 on Build GRCh38
rs376425643 Chr.11: 67321451-67321451 on Build GRCh38
rs939552307 Chr.11: 67321285-67321285 on Build GRCh38
rs557907130 Chr.11: 67321103-67321103 on Build GRCh38
rs994229324 Chr.11: 67321412-67321412 on Build GRCh38
rs928131668 Chr.11: 67321264-67321264 on Build GRCh38
rs763539848 Chr.11: 67321325-67321325 on Build GRCh38
rs774838339 Chr.11: 67321224-67321224 on Build GRCh38
rs909421768 Chr.11: 67321364-67321364 on Build GRCh38
rs890965794 Chr.11: 67321422-67321422 on Build GRCh38
rs537660907 Chr.11: 67321149-67321149 on Build GRCh38
rs957547046 Chr.11: 67321548-67321548 on Build GRCh38
rs756626199 Chr.11: 67321413-67321413 on Build GRCh38
rs148013431 Chr.11: 67321276-67321276 on Build GRCh38
rs1055385696 Chr.11: 67321323-67321323 on Build GRCh38
rs962509586 Chr.11: 67321387-67321387 on Build GRCh38
rs897226238 Chr.11: 67321402-67321402 on Build GRCh38
rs887315 Chr.11: 67321249-67321249 on Build GRCh38
rs184904291 Chr.11: 67321470-67321470 on Build GRCh38
rs574120077 Chr.11: 67321362-67321362 on Build GRCh38
rs1012604846 Chr.11: 67321281-67321281 on Build GRCh38
rs766715449 Chr.11: 67321541-67321541 on Build GRCh38
rs370099789 Chr.11: 67321231-67321232 on Build GRCh38
rs764977410 Chr.11: 67321216-67321216 on Build GRCh38
rs978233298 Chr.11: 67321253-67321253 on Build GRCh38
rs957057680 Chr.11: 67321198-67321198 on Build GRCh38
rs188134964 Chr.11: 67321561-67321561 on Build GRCh38
rs912968356 Chr.11: 67321232-67321232 on Build GRCh38
rs866093946 Chr.11: 67321426-67321426 on Build GRCh38
rs1022597468 Chr.11: 67321300-67321300 on Build GRCh38
rs577984491 Chr.11: 67321286-67321286 on Build GRCh38

View Full Product Details