Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 81890403-81890920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR4300
Entrez Gene ID
100422823
Gene Name
microRNA 4300
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 81890741-81890836 on Build GRCh38
Transcripts
NR_036186.1 NR_120571.1

Gene Symbol
MIR4300HG
Entrez Gene ID
101928989
Gene Name
MIR4300 host gene
Gene Aliases
-
UniGene
Hs.145117
Gene Chromosome Location
Chr.11: 81879851-82403913 on Build GRCh38
Transcripts
NR_036186.1 NR_120571.1

SNP Detail

SNP ID Chromosome Location
rs77127366 Chr.11: 81890430-81890430 on Build GRCh38
rs11607302 Chr.11: 81890533-81890533 on Build GRCh38
rs1044486546 Chr.11: 81890659-81890659 on Build GRCh38
rs777297232 Chr.11: 81890439-81890439 on Build GRCh38
rs779487079 Chr.11: 81890726-81890726 on Build GRCh38
rs947918138 Chr.11: 81890853-81890853 on Build GRCh38
rs901072483 Chr.11: 81890647-81890647 on Build GRCh38
rs1028755978 Chr.11: 81890824-81890824 on Build GRCh38
rs755713936 Chr.11: 81890850-81890850 on Build GRCh38
rs935622084 Chr.11: 81890459-81890459 on Build GRCh38
rs543747004 Chr.11: 81890707-81890707 on Build GRCh38
rs776197531 Chr.11: 81890463-81890464 on Build GRCh38
rs780443161 Chr.11: 81890852-81890852 on Build GRCh38
rs570516494 Chr.11: 81890525-81890525 on Build GRCh38
rs930299271 Chr.11: 81890522-81890522 on Build GRCh38
rs915428735 Chr.11: 81890494-81890494 on Build GRCh38
rs75322260 Chr.11: 81890566-81890566 on Build GRCh38
rs11601151 Chr.11: 81890451-81890451 on Build GRCh38
rs984475749 Chr.11: 81890847-81890847 on Build GRCh38
rs754924320 Chr.11: 81890746-81890746 on Build GRCh38
rs957150579 Chr.11: 81890851-81890851 on Build GRCh38
rs1054361771 Chr.11: 81890683-81890683 on Build GRCh38
rs907521492 Chr.11: 81890847-81890848 on Build GRCh38
rs1023668708 Chr.11: 81890784-81890784 on Build GRCh38
rs905868344 Chr.11: 81890443-81890443 on Build GRCh38
rs184272314 Chr.11: 81890744-81890744 on Build GRCh38
rs150809979 Chr.11: 81890579-81890579 on Build GRCh38
rs776855136 Chr.11: 81890848-81890848 on Build GRCh38
rs988683984 Chr.11: 81890461-81890461 on Build GRCh38
rs1035058782 Chr.11: 81890846-81890846 on Build GRCh38
rs940566435 Chr.11: 81890562-81890562 on Build GRCh38
rs147006890 Chr.11: 81890821-81890821 on Build GRCh38
rs1000111274 Chr.11: 81890450-81890450 on Build GRCh38
rs950215582 Chr.11: 81890622-81890622 on Build GRCh38
rs149503927 Chr.11: 81890851-81890852 on Build GRCh38
rs566062453 Chr.11: 81890524-81890524 on Build GRCh38
rs142477884 Chr.11: 81890428-81890428 on Build GRCh38
rs1019093199 Chr.11: 81890489-81890489 on Build GRCh38
rs192418226 Chr.11: 81890519-81890519 on Build GRCh38
rs576120127 Chr.11: 81890661-81890661 on Build GRCh38
rs892069174 Chr.11: 81890688-81890688 on Build GRCh38
rs369351267 Chr.11: 81890444-81890445 on Build GRCh38
rs1945895 Chr.11: 81890845-81890845 on Build GRCh38
rs1045271452 Chr.11: 81890854-81890854 on Build GRCh38
rs974511535 Chr.11: 81890517-81890517 on Build GRCh38
rs986336032 Chr.11: 81890868-81890868 on Build GRCh38
rs946858131 Chr.11: 81890863-81890863 on Build GRCh38
rs966215249 Chr.11: 81890506-81890506 on Build GRCh38
rs555110079 Chr.11: 81890684-81890684 on Build GRCh38
rs144577738 Chr.11: 81890441-81890441 on Build GRCh38
rs780110695 Chr.11: 81890875-81890875 on Build GRCh38
rs770698427 Chr.11: 81890475-81890475 on Build GRCh38
rs76174975 Chr.11: 81890556-81890556 on Build GRCh38
rs11603185 Chr.11: 81890842-81890842 on Build GRCh38
rs1000163767 Chr.11: 81890674-81890674 on Build GRCh38
rs915360249 Chr.11: 81890862-81890862 on Build GRCh38
rs554773458 Chr.11: 81890603-81890603 on Build GRCh38
rs1019564967 Chr.11: 81890738-81890738 on Build GRCh38
rs988410914 Chr.11: 81890852-81890852 on Build GRCh38
rs969485859 Chr.11: 81890790-81890790 on Build GRCh38
rs141825015 Chr.11: 81890572-81890572 on Build GRCh38

View Full Product Details