Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49910707-49911211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC283332
Entrez Gene ID
283332
Gene Name
uncharacterized LOC283332
Gene Aliases
-
UniGene
Hs.127688
Gene Chromosome Location
Chr.12: 49908882-49911863 on Build GRCh38
Transcripts
NR_026948.1

SNP Detail

SNP ID Chromosome Location
rs74089088 Chr.12: 49910790-49910790 on Build GRCh38
rs992747908 Chr.12: 49910949-49910949 on Build GRCh38
rs549714354 Chr.12: 49911155-49911155 on Build GRCh38
rs879237155 Chr.12: 49911084-49911084 on Build GRCh38
rs2298640 Chr.12: 49910791-49910791 on Build GRCh38
rs563165092 Chr.12: 49910898-49910898 on Build GRCh38
rs960650248 Chr.12: 49910942-49910942 on Build GRCh38
rs994789380 Chr.12: 49910786-49910786 on Build GRCh38
rs921602098 Chr.12: 49910843-49910843 on Build GRCh38
rs912169288 Chr.12: 49910887-49910887 on Build GRCh38
rs960131417 Chr.12: 49910812-49910812 on Build GRCh38
rs1045020459 Chr.12: 49910959-49910959 on Build GRCh38
rs116203235 Chr.12: 49911181-49911181 on Build GRCh38
rs539378725 Chr.12: 49910768-49910768 on Build GRCh38
rs758397821 Chr.12: 49911131-49911131 on Build GRCh38
rs761971561 Chr.12: 49910801-49910801 on Build GRCh38
rs577491532 Chr.12: 49910860-49910860 on Build GRCh38
rs1025550571 Chr.12: 49910929-49910929 on Build GRCh38
rs1006655106 Chr.12: 49910873-49910873 on Build GRCh38
rs554377185 Chr.12: 49910830-49910830 on Build GRCh38
rs487193 Chr.12: 49910815-49910815 on Build GRCh38
rs2062837 Chr.12: 49911102-49911102 on Build GRCh38
rs765101749 Chr.12: 49911016-49911016 on Build GRCh38
rs989425705 Chr.12: 49911063-49911063 on Build GRCh38
rs549651102 Chr.12: 49910782-49910782 on Build GRCh38
rs563975924 Chr.12: 49911105-49911105 on Build GRCh38
rs546552491 Chr.12: 49910877-49910877 on Build GRCh38
rs181307750 Chr.12: 49910908-49910908 on Build GRCh38
rs74089090 Chr.12: 49911017-49911017 on Build GRCh38
rs141845799 Chr.12: 49910731-49910731 on Build GRCh38
rs918521159 Chr.12: 49910755-49910755 on Build GRCh38
rs34869745 Chr.12: 49910769-49910770 on Build GRCh38
rs953005209 Chr.12: 49911137-49911137 on Build GRCh38
rs747793966 Chr.12: 49911109-49911118 on Build GRCh38
rs487165 Chr.12: 49910805-49910805 on Build GRCh38
rs450798 Chr.12: 49911166-49911166 on Build GRCh38
rs775672863 Chr.12: 49910769-49910769 on Build GRCh38
rs559923944 Chr.12: 49910917-49910917 on Build GRCh38
rs970092445 Chr.12: 49910995-49910995 on Build GRCh38
rs58262364 Chr.12: 49911010-49911010 on Build GRCh38
rs527354270 Chr.12: 49911040-49911040 on Build GRCh38
rs768029724 Chr.12: 49911189-49911189 on Build GRCh38
rs577212564 Chr.12: 49910834-49910834 on Build GRCh38
rs370938860 Chr.12: 49911188-49911188 on Build GRCh38
rs892089030 Chr.12: 49910814-49910814 on Build GRCh38
rs558682745 Chr.12: 49910818-49910818 on Build GRCh38
rs987191456 Chr.12: 49911139-49911139 on Build GRCh38
rs889471194 Chr.12: 49911165-49911165 on Build GRCh38
rs1020127327 Chr.12: 49911044-49911044 on Build GRCh38
rs962555609 Chr.12: 49910923-49910923 on Build GRCh38
rs2298641 Chr.12: 49911143-49911143 on Build GRCh38
rs115722759 Chr.12: 49911172-49911172 on Build GRCh38
rs921706951 Chr.12: 49911125-49911125 on Build GRCh38
rs968419861 Chr.12: 49911047-49911047 on Build GRCh38
rs567799236 Chr.12: 49910767-49910767 on Build GRCh38
rs1024274129 Chr.12: 49910970-49910970 on Build GRCh38
rs936177285 Chr.12: 49911060-49911060 on Build GRCh38
rs940849484 Chr.12: 49910888-49910888 on Build GRCh38
rs926160643 Chr.12: 49910800-49910800 on Build GRCh38
rs898272931 Chr.12: 49911173-49911173 on Build GRCh38
rs541769745 Chr.12: 49910925-49910925 on Build GRCh38
rs756770448 Chr.12: 49911186-49911186 on Build GRCh38
rs541054602 Chr.12: 49910901-49910901 on Build GRCh38
rs749858203 Chr.12: 49910783-49910783 on Build GRCh38
rs972692123 Chr.12: 49910751-49910751 on Build GRCh38
rs759824707 Chr.12: 49911187-49911187 on Build GRCh38

View Full Product Details