Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49911808-49912313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101927292
Entrez Gene ID
101927292
Gene Name
uncharacterized LOC101927292
Gene Aliases
-
UniGene
Hs.129345
Gene Chromosome Location
Chr.12: 49911953-49926339 on Build GRCh38
Transcripts
NR_110048.1 NR_026948.1

Gene Symbol
LOC283332
Entrez Gene ID
283332
Gene Name
uncharacterized LOC283332
Gene Aliases
-
UniGene
Hs.127688
Gene Chromosome Location
Chr.12: 49908882-49911863 on Build GRCh38
Transcripts
NR_110048.1 NR_026948.1

SNP Detail

SNP ID Chromosome Location
rs542303403 Chr.12: 49912276-49912276 on Build GRCh38
rs892414513 Chr.12: 49912174-49912174 on Build GRCh38
rs936539934 Chr.12: 49912168-49912168 on Build GRCh38
rs1009175095 Chr.12: 49911842-49911842 on Build GRCh38
rs17261501 Chr.12: 49911977-49911977 on Build GRCh38
rs967203223 Chr.12: 49912126-49912126 on Build GRCh38
rs891181517 Chr.12: 49912266-49912266 on Build GRCh38
rs897540368 Chr.12: 49912173-49912173 on Build GRCh38
rs926470131 Chr.12: 49912156-49912156 on Build GRCh38
rs960894416 Chr.12: 49912027-49912027 on Build GRCh38
rs755400369 Chr.12: 49912113-49912113 on Build GRCh38
rs964298302 Chr.12: 49911988-49911988 on Build GRCh38
rs1010236737 Chr.12: 49912175-49912175 on Build GRCh38
rs887458949 Chr.12: 49912236-49912236 on Build GRCh38
rs1040106187 Chr.12: 49911896-49911896 on Build GRCh38
rs897609548 Chr.12: 49911930-49911930 on Build GRCh38
rs80185865 Chr.12: 49912169-49912169 on Build GRCh38
rs1018419655 Chr.12: 49912278-49912278 on Build GRCh38
rs372299285 Chr.12: 49912228-49912228 on Build GRCh38
rs1033919407 Chr.12: 49912264-49912264 on Build GRCh38
rs935873416 Chr.12: 49912146-49912146 on Build GRCh38
rs755866420 Chr.12: 49911965-49911965 on Build GRCh38
rs35754018 Chr.12: 49912115-49912116 on Build GRCh38
rs563528616 Chr.12: 49912022-49912022 on Build GRCh38
rs147518676 Chr.12: 49911949-49911949 on Build GRCh38
rs150310497 Chr.12: 49911843-49911843 on Build GRCh38
rs796336853 Chr.12: 49912151-49912151 on Build GRCh38
rs899986179 Chr.12: 49912286-49912286 on Build GRCh38
rs1010269198 Chr.12: 49911864-49911864 on Build GRCh38
rs140072881 Chr.12: 49912052-49912052 on Build GRCh38
rs1050320935 Chr.12: 49912147-49912147 on Build GRCh38
rs542950513 Chr.12: 49912102-49912102 on Build GRCh38
rs402468 Chr.12: 49911944-49911944 on Build GRCh38
rs1033782518 Chr.12: 49912024-49912024 on Build GRCh38
rs910483420 Chr.12: 49912153-49912153 on Build GRCh38
rs1020371957 Chr.12: 49911935-49911935 on Build GRCh38
rs982453247 Chr.12: 49912083-49912083 on Build GRCh38
rs540575070 Chr.12: 49911830-49911830 on Build GRCh38
rs530023196 Chr.12: 49912062-49912062 on Build GRCh38
rs951112075 Chr.12: 49912076-49912076 on Build GRCh38
rs145513614 Chr.12: 49912137-49912137 on Build GRCh38
rs573275103 Chr.12: 49912039-49912043 on Build GRCh38
rs1001856967 Chr.12: 49912255-49912255 on Build GRCh38
rs149705227 Chr.12: 49912120-49912120 on Build GRCh38
rs925291419 Chr.12: 49912138-49912138 on Build GRCh38
rs144902253 Chr.12: 49911872-49911872 on Build GRCh38

View Full Product Details