Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53963952-53964451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOTAIR
Entrez Gene ID
100124700
Gene Name
HOX transcript antisense RNA
Gene Aliases
HOXAS, HOXC-AS4, HOXC11-AS1, NCRNA00072
UniGene
Hs.197076
Gene Chromosome Location
Chr.12: 53962308-53974956 on Build GRCh38
Transcripts
NR_003716.3 NR_047517.1 NR_047518.1

SNP Detail

SNP ID Chromosome Location
rs893489335 Chr.12: 53964402-53964402 on Build GRCh38
rs931583861 Chr.12: 53963998-53963998 on Build GRCh38
rs1017398932 Chr.12: 53964086-53964086 on Build GRCh38
rs1009132786 Chr.12: 53964127-53964127 on Build GRCh38
rs767374082 Chr.12: 53964296-53964296 on Build GRCh38
rs35951424 Chr.12: 53964213-53964213 on Build GRCh38
rs202007535 Chr.12: 53964394-53964395 on Build GRCh38
rs376145776 Chr.12: 53964328-53964328 on Build GRCh38
rs200062893 Chr.12: 53963977-53963977 on Build GRCh38
rs976268564 Chr.12: 53964255-53964255 on Build GRCh38
rs529620501 Chr.12: 53964220-53964220 on Build GRCh38
rs901530450 Chr.12: 53964002-53964002 on Build GRCh38
rs763855108 Chr.12: 53963972-53963973 on Build GRCh38
rs957502187 Chr.12: 53964182-53964182 on Build GRCh38
rs1024288330 Chr.12: 53964126-53964126 on Build GRCh38
rs533991618 Chr.12: 53964388-53964389 on Build GRCh38
rs200484491 Chr.12: 53964223-53964223 on Build GRCh38
rs71227278 Chr.12: 53964263-53964264 on Build GRCh38
rs1046162992 Chr.12: 53964048-53964048 on Build GRCh38
rs904946081 Chr.12: 53964052-53964052 on Build GRCh38
rs67188582 Chr.12: 53964264-53964265 on Build GRCh38
rs201719283 Chr.12: 53964230-53964230 on Build GRCh38
rs571190095 Chr.12: 53964130-53964131 on Build GRCh38
rs908729786 Chr.12: 53964035-53964035 on Build GRCh38
rs954591197 Chr.12: 53964122-53964122 on Build GRCh38
rs1021265875 Chr.12: 53964119-53964119 on Build GRCh38
rs34408646 Chr.12: 53964389-53964389 on Build GRCh38
rs1047641591 Chr.12: 53964323-53964323 on Build GRCh38
rs949567501 Chr.12: 53964036-53964036 on Build GRCh38
rs867710570 Chr.12: 53964020-53964020 on Build GRCh38
rs756898416 Chr.12: 53964214-53964214 on Build GRCh38
rs569819779 Chr.12: 53964242-53964242 on Build GRCh38
rs201203822 Chr.12: 53964268-53964268 on Build GRCh38
rs977614250 Chr.12: 53964148-53964148 on Build GRCh38
rs778772853 Chr.12: 53964014-53964014 on Build GRCh38
rs17840857 Chr.12: 53963973-53963973 on Build GRCh38
rs964120087 Chr.12: 53964199-53964199 on Build GRCh38
rs949614562 Chr.12: 53964282-53964282 on Build GRCh38
rs76948242 Chr.12: 53964221-53964222 on Build GRCh38
rs796610683 Chr.12: 53964222-53964222 on Build GRCh38
rs756772639 Chr.12: 53964213-53964213 on Build GRCh38
rs1031901027 Chr.12: 53964178-53964178 on Build GRCh38
rs112955399 Chr.12: 53964224-53964224 on Build GRCh38
rs537377655 Chr.12: 53964281-53964281 on Build GRCh38
rs893440759 Chr.12: 53964090-53964090 on Build GRCh38
rs926753699 Chr.12: 53964352-53964352 on Build GRCh38
rs60732492 Chr.12: 53964265-53964266 on Build GRCh38
rs894551180 Chr.12: 53964118-53964118 on Build GRCh38
rs536505412 Chr.12: 53964056-53964056 on Build GRCh38
rs929197676 Chr.12: 53964026-53964026 on Build GRCh38
rs200571747 Chr.12: 53964263-53964263 on Build GRCh38
rs750425423 Chr.12: 53964401-53964401 on Build GRCh38
rs975452975 Chr.12: 53964214-53964214 on Build GRCh38
rs111434707 Chr.12: 53963973-53963973 on Build GRCh38
rs564341635 Chr.12: 53964271-53964271 on Build GRCh38
rs1053388018 Chr.12: 53964064-53964064 on Build GRCh38
rs756827347 Chr.12: 53964012-53964012 on Build GRCh38
rs753712445 Chr.12: 53964010-53964010 on Build GRCh38
rs937770456 Chr.12: 53964361-53964361 on Build GRCh38
rs552383007 Chr.12: 53964290-53964290 on Build GRCh38
rs749561321 Chr.12: 53964213-53964214 on Build GRCh38
rs199680547 Chr.12: 53964222-53964222 on Build GRCh38
rs183607878 Chr.12: 53964096-53964096 on Build GRCh38
rs922905327 Chr.12: 53964276-53964276 on Build GRCh38
rs1009500773 Chr.12: 53964112-53964112 on Build GRCh38
rs1019744894 Chr.12: 53964146-53964146 on Build GRCh38
rs1046549502 Chr.12: 53964289-53964289 on Build GRCh38
rs143496128 Chr.12: 53964262-53964263 on Build GRCh38

View Full Product Details