Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 97464918-97465419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMST
Entrez Gene ID
196475
Gene Name
rhabdomyosarcoma 2 associated transcript (non-protein coding)
Gene Aliases
LINC00054, NCRMS, NCRNA00054
UniGene
Hs.652568
Gene Chromosome Location
Chr.12: 97465021-97533766 on Build GRCh38
Transcripts
NR_024037.1

SNP Detail

SNP ID Chromosome Location
rs948262896 Chr.12: 97465261-97465261 on Build GRCh38
rs573633417 Chr.12: 97465263-97465263 on Build GRCh38
rs1028929727 Chr.12: 97465125-97465125 on Build GRCh38
rs767502508 Chr.12: 97465039-97465039 on Build GRCh38
rs1048251521 Chr.12: 97465225-97465225 on Build GRCh38
rs1004530735 Chr.12: 97464966-97464966 on Build GRCh38
rs1014012875 Chr.12: 97465044-97465044 on Build GRCh38
rs540098795 Chr.12: 97465155-97465155 on Build GRCh38
rs1037473700 Chr.12: 97465336-97465336 on Build GRCh38
rs1033170302 Chr.12: 97465114-97465114 on Build GRCh38
rs1016477908 Chr.12: 97465043-97465043 on Build GRCh38
rs766512416 Chr.12: 97464993-97464993 on Build GRCh38
rs753554211 Chr.12: 97465378-97465378 on Build GRCh38
rs1015300725 Chr.12: 97464987-97464987 on Build GRCh38
rs759639043 Chr.12: 97465035-97465035 on Build GRCh38
rs534428417 Chr.12: 97465276-97465276 on Build GRCh38
rs930713628 Chr.12: 97465187-97465187 on Build GRCh38
rs1004712382 Chr.12: 97465029-97465029 on Build GRCh38
rs954683181 Chr.12: 97465147-97465147 on Build GRCh38
rs970233768 Chr.12: 97465074-97465074 on Build GRCh38
rs774336808 Chr.12: 97465006-97465006 on Build GRCh38
rs978949986 Chr.12: 97465109-97465109 on Build GRCh38
rs996071931 Chr.12: 97465071-97465071 on Build GRCh38
rs956354769 Chr.12: 97465142-97465142 on Build GRCh38
rs748688724 Chr.12: 97465052-97465052 on Build GRCh38
rs141071985 Chr.12: 97465169-97465169 on Build GRCh38
rs888278308 Chr.12: 97465233-97465233 on Build GRCh38
rs190161352 Chr.12: 97465016-97465016 on Build GRCh38
rs367597181 Chr.12: 97465321-97465321 on Build GRCh38
rs763821828 Chr.12: 97465269-97465269 on Build GRCh38
rs915353934 Chr.12: 97465246-97465246 on Build GRCh38
rs928248091 Chr.12: 97465271-97465271 on Build GRCh38
rs553095978 Chr.12: 97465368-97465368 on Build GRCh38
rs536557144 Chr.12: 97465254-97465254 on Build GRCh38
rs957039596 Chr.12: 97465207-97465207 on Build GRCh38
rs558469581 Chr.12: 97465158-97465158 on Build GRCh38
rs938330396 Chr.12: 97465273-97465273 on Build GRCh38
rs566157815 Chr.12: 97465055-97465055 on Build GRCh38
rs755912551 Chr.12: 97465237-97465237 on Build GRCh38
rs989585560 Chr.12: 97465226-97465226 on Build GRCh38
rs963246853 Chr.12: 97465064-97465064 on Build GRCh38
rs940396610 Chr.12: 97465319-97465319 on Build GRCh38
rs974870871 Chr.12: 97465181-97465181 on Build GRCh38
rs922042241 Chr.12: 97465185-97465185 on Build GRCh38
rs898965962 Chr.12: 97465338-97465338 on Build GRCh38
rs1056763106 Chr.12: 97465275-97465275 on Build GRCh38
rs181303613 Chr.12: 97465163-97465163 on Build GRCh38
rs781736463 Chr.12: 97465032-97465032 on Build GRCh38

View Full Product Details