Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98502522-98503068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC643770
Entrez Gene ID
643770
Gene Name
uncharacterized LOC643770
Gene Aliases
-
UniGene
Hs.129589
Gene Chromosome Location
Chr.12: 98485544-98503855 on Build GRCh38
Transcripts
NR_038383.1

SNP Detail

SNP ID Chromosome Location
rs911260230 Chr.12: 98503000-98503000 on Build GRCh38
rs969800043 Chr.12: 98503032-98503032 on Build GRCh38
rs541331305 Chr.12: 98502768-98502768 on Build GRCh38
rs546925646 Chr.12: 98502970-98502970 on Build GRCh38
rs186025229 Chr.12: 98502640-98502640 on Build GRCh38
rs972945619 Chr.12: 98502909-98502909 on Build GRCh38
rs558296855 Chr.12: 98502695-98502695 on Build GRCh38
rs915578275 Chr.12: 98502709-98502710 on Build GRCh38
rs548745093 Chr.12: 98502890-98502890 on Build GRCh38
rs55846846 Chr.12: 98502837-98502837 on Build GRCh38
rs145996959 Chr.12: 98502660-98502660 on Build GRCh38
rs796072356 Chr.12: 98502839-98502843 on Build GRCh38
rs757713451 Chr.12: 98502609-98502609 on Build GRCh38
rs1027227023 Chr.12: 98502671-98502671 on Build GRCh38
rs566447510 Chr.12: 98503006-98503006 on Build GRCh38
rs959139819 Chr.12: 98502680-98502680 on Build GRCh38
rs552949796 Chr.12: 98502597-98502597 on Build GRCh38
rs3825263 Chr.12: 98502724-98502724 on Build GRCh38
rs894810364 Chr.12: 98502870-98502870 on Build GRCh38
rs991901293 Chr.12: 98502692-98502692 on Build GRCh38
rs867328857 Chr.12: 98502845-98502845 on Build GRCh38
rs982931627 Chr.12: 98502684-98502684 on Build GRCh38
rs866246625 Chr.12: 98503009-98503009 on Build GRCh38
rs1027402374 Chr.12: 98502880-98502880 on Build GRCh38
rs199663641 Chr.12: 98502840-98502840 on Build GRCh38
rs777255419 Chr.12: 98502636-98502636 on Build GRCh38
rs559482298 Chr.12: 98502771-98502771 on Build GRCh38
rs899836813 Chr.12: 98502867-98502867 on Build GRCh38
rs973280183 Chr.12: 98502699-98502699 on Build GRCh38
rs944105676 Chr.12: 98502831-98502831 on Build GRCh38
rs944043428 Chr.12: 98503025-98503025 on Build GRCh38
rs754334622 Chr.12: 98502598-98502598 on Build GRCh38
rs249816 Chr.12: 98502841-98502841 on Build GRCh38
rs945219129 Chr.12: 98502790-98502790 on Build GRCh38
rs921745706 Chr.12: 98502855-98502855 on Build GRCh38
rs11109503 Chr.12: 98502833-98502833 on Build GRCh38
rs763632131 Chr.12: 98502678-98502678 on Build GRCh38
rs190651470 Chr.12: 98502641-98502641 on Build GRCh38
rs908718053 Chr.12: 98502686-98502686 on Build GRCh38
rs199793476 Chr.12: 98502843-98502843 on Build GRCh38
rs1053367551 Chr.12: 98502628-98502628 on Build GRCh38
rs11836259 Chr.12: 98502630-98502630 on Build GRCh38
rs867922081 Chr.12: 98502838-98502838 on Build GRCh38
rs911198612 Chr.12: 98502740-98502740 on Build GRCh38
rs1025187827 Chr.12: 98502696-98502696 on Build GRCh38
rs56351219 Chr.12: 98502843-98502843 on Build GRCh38
rs976686679 Chr.12: 98503036-98503036 on Build GRCh38
rs866966592 Chr.12: 98502830-98502830 on Build GRCh38
rs574300317 Chr.12: 98502752-98502752 on Build GRCh38
rs986106902 Chr.12: 98502986-98502986 on Build GRCh38
rs544675959 Chr.12: 98502818-98502818 on Build GRCh38
rs549239494 Chr.12: 98502558-98502558 on Build GRCh38
rs925854754 Chr.12: 98502769-98502769 on Build GRCh38
rs944159576 Chr.12: 98502757-98502757 on Build GRCh38
rs561231366 Chr.12: 98502905-98502905 on Build GRCh38
rs370722970 Chr.12: 98502553-98502553 on Build GRCh38
rs1033386058 Chr.12: 98502806-98502806 on Build GRCh38
rs1013219517 Chr.12: 98502887-98502887 on Build GRCh38
rs961089824 Chr.12: 98502907-98502907 on Build GRCh38
rs1015745688 Chr.12: 98502914-98502914 on Build GRCh38
rs386765642 Chr.12: 98502640-98502641 on Build GRCh38
rs903786806 Chr.12: 98502797-98502797 on Build GRCh38
rs934593478 Chr.12: 98502775-98502775 on Build GRCh38
rs891839467 Chr.12: 98502782-98502782 on Build GRCh38
rs528347691 Chr.12: 98502939-98502939 on Build GRCh38
rs1041149185 Chr.12: 98502836-98502836 on Build GRCh38
rs182470683 Chr.12: 98502697-98502697 on Build GRCh38
rs577834296 Chr.12: 98502801-98502801 on Build GRCh38
rs368372902 Chr.12: 98502945-98502945 on Build GRCh38
rs1042754640 Chr.12: 98502766-98502766 on Build GRCh38
rs1008246049 Chr.12: 98502592-98502592 on Build GRCh38
rs892528980 Chr.12: 98502629-98502629 on Build GRCh38
rs985503559 Chr.12: 98502726-98502726 on Build GRCh38
rs143072699 Chr.12: 98502837-98502838 on Build GRCh38
rs761144789 Chr.12: 98502579-98502579 on Build GRCh38
rs1024740392 Chr.12: 98502891-98502891 on Build GRCh38
rs1052927416 Chr.12: 98502784-98502784 on Build GRCh38
rs779290582 Chr.12: 98502620-98502620 on Build GRCh38
rs1042585847 Chr.12: 98502794-98502794 on Build GRCh38
rs960525172 Chr.12: 98502935-98502935 on Build GRCh38
rs537076758 Chr.12: 98502709-98502709 on Build GRCh38
rs574536954 Chr.12: 98503031-98503032 on Build GRCh38
rs1000548397 Chr.12: 98502668-98502668 on Build GRCh38

View Full Product Details