Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120491260-120491769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLL1
Entrez Gene ID
8655
Gene Name
dynein light chain LC8-type 1
Gene Aliases
DLC1, DLC8, DNCL1, DNCLC1, LC8, LC8a, PIN, hdlc1
UniGene
Hs.5120
Gene Chromosome Location
Chr.12: 120469857-120498495 on Build GRCh38
Transcripts
NM_001037494.1 NR_038854.1

Gene Symbol
NRAV
Entrez Gene ID
100506668
Gene Name
negative regulator of antiviral response (non-protein coding)
Gene Aliases
DYNLL1-AS1, DYNLL1AS1
UniGene
Hs.660080
Gene Chromosome Location
Chr.12: 120490338-120495946 on Build GRCh38
Transcripts
NM_001037494.1 NR_038854.1

SNP Detail

SNP ID Chromosome Location
rs562229337 Chr.12: 120491688-120491688 on Build GRCh38
rs768070727 Chr.12: 120491424-120491424 on Build GRCh38
rs71886391 Chr.12: 120491340-120491356 on Build GRCh38
rs978290286 Chr.12: 120491552-120491552 on Build GRCh38
rs140316317 Chr.12: 120491701-120491701 on Build GRCh38
rs962505048 Chr.12: 120491411-120491411 on Build GRCh38
rs1002773082 Chr.12: 120491739-120491739 on Build GRCh38
rs776422628 Chr.12: 120491644-120491644 on Build GRCh38
rs1048033849 Chr.12: 120491662-120491662 on Build GRCh38
rs1003949845 Chr.12: 120491356-120491356 on Build GRCh38
rs996882315 Chr.12: 120491444-120491444 on Build GRCh38
rs10789 Chr.12: 120491339-120491339 on Build GRCh38
rs182393260 Chr.12: 120491591-120491591 on Build GRCh38
rs117807066 Chr.12: 120491660-120491660 on Build GRCh38
rs955290919 Chr.12: 120491482-120491482 on Build GRCh38
rs1027991833 Chr.12: 120491479-120491479 on Build GRCh38
rs886637563 Chr.12: 120491351-120491351 on Build GRCh38
rs925328286 Chr.12: 120491620-120491620 on Build GRCh38
rs1003495005 Chr.12: 120491445-120491445 on Build GRCh38
rs921629653 Chr.12: 120491655-120491655 on Build GRCh38
rs577658493 Chr.12: 120491294-120491294 on Build GRCh38
rs990931912 Chr.12: 120491520-120491520 on Build GRCh38
rs544243578 Chr.12: 120491732-120491732 on Build GRCh38
rs968248083 Chr.12: 120491551-120491551 on Build GRCh38
rs1014804701 Chr.12: 120491402-120491402 on Build GRCh38
rs540030397 Chr.12: 120491305-120491305 on Build GRCh38
rs557325982 Chr.12: 120491597-120491597 on Build GRCh38
rs887637764 Chr.12: 120491376-120491376 on Build GRCh38
rs746767187 Chr.12: 120491607-120491607 on Build GRCh38
rs1013847099 Chr.12: 120491328-120491328 on Build GRCh38
rs1004719921 Chr.12: 120491389-120491389 on Build GRCh38
rs11065140 Chr.12: 120491738-120491738 on Build GRCh38
rs11612721 Chr.12: 120491639-120491639 on Build GRCh38
rs913723388 Chr.12: 120491590-120491590 on Build GRCh38
rs1035006271 Chr.12: 120491468-120491468 on Build GRCh38
rs757522161 Chr.12: 120491339-120491340 on Build GRCh38
rs944964609 Chr.12: 120491640-120491640 on Build GRCh38
rs940518052 Chr.12: 120491708-120491708 on Build GRCh38
rs1006772551 Chr.12: 120491712-120491712 on Build GRCh38
rs886711378 Chr.12: 120491698-120491698 on Build GRCh38
rs374222538 Chr.12: 120491349-120491365 on Build GRCh38
rs917961305 Chr.12: 120491656-120491656 on Build GRCh38
rs773870997 Chr.12: 120491580-120491580 on Build GRCh38
rs1040102769 Chr.12: 120491323-120491323 on Build GRCh38
rs975733838 Chr.12: 120491647-120491647 on Build GRCh38
rs1048808231 Chr.12: 120491674-120491674 on Build GRCh38
rs758640945 Chr.12: 120491534-120491534 on Build GRCh38
rs887575218 Chr.12: 120491687-120491687 on Build GRCh38
rs938114866 Chr.12: 120491637-120491637 on Build GRCh38
rs768623377 Chr.12: 120491619-120491619 on Build GRCh38
rs1017073861 Chr.12: 120491398-120491398 on Build GRCh38

View Full Product Details