Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123083725-123084226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100507091
Entrez Gene ID
100507091
Gene Name
uncharacterized LOC100507091
Gene Aliases
-
UniGene
Hs.683912
Gene Chromosome Location
Chr.12: 123081384-123084744 on Build GRCh38
Transcripts
NR_038290.1 NM_001300801.1 NM_020845.2

Gene Symbol
PITPNM2
Entrez Gene ID
57605
Gene Name
phosphatidylinositol transfer protein membrane associated 2
Gene Aliases
NIR-3, NIR3, RDGB2, RDGBA2
UniGene
Hs.272759
Gene Chromosome Location
Chr.12: 122983480-123110489 on Build GRCh38
Transcripts
NR_038290.1 NM_001300801.1 NM_020845.2

SNP Detail

SNP ID Chromosome Location
rs929509196 Chr.12: 123084175-123084175 on Build GRCh38
rs372933217 Chr.12: 123084102-123084102 on Build GRCh38
rs543637004 Chr.12: 123083883-123083883 on Build GRCh38
rs938329384 Chr.12: 123084111-123084111 on Build GRCh38
rs918954719 Chr.12: 123084095-123084095 on Build GRCh38
rs760914036 Chr.12: 123083940-123083940 on Build GRCh38
rs1043309866 Chr.12: 123083859-123083859 on Build GRCh38
rs562346006 Chr.12: 123083884-123083884 on Build GRCh38
rs529537153 Chr.12: 123083886-123083886 on Build GRCh38
rs140962311 Chr.12: 123083978-123083978 on Build GRCh38
rs1028372936 Chr.12: 123084045-123084045 on Build GRCh38
rs901171648 Chr.12: 123084011-123084011 on Build GRCh38
rs1043234824 Chr.12: 123084191-123084191 on Build GRCh38
rs868388555 Chr.12: 123083779-123083779 on Build GRCh38
rs4148862 Chr.12: 123084072-123084072 on Build GRCh38
rs948903153 Chr.12: 123084161-123084161 on Build GRCh38
rs565693570 Chr.12: 123084077-123084077 on Build GRCh38
rs992991681 Chr.12: 123084145-123084145 on Build GRCh38
rs867295258 Chr.12: 123084036-123084036 on Build GRCh38
rs1056877304 Chr.12: 123083756-123083756 on Build GRCh38
rs966047070 Chr.12: 123083916-123083916 on Build GRCh38
rs189059746 Chr.12: 123083802-123083802 on Build GRCh38
rs976417687 Chr.12: 123083931-123083931 on Build GRCh38
rs928305927 Chr.12: 123084100-123084100 on Build GRCh38
rs776706030 Chr.12: 123084153-123084153 on Build GRCh38
rs777736002 Chr.12: 123083888-123083888 on Build GRCh38
rs999051168 Chr.12: 123083914-123083914 on Build GRCh38
rs1057118253 Chr.12: 123084141-123084141 on Build GRCh38
rs979826943 Chr.12: 123084086-123084086 on Build GRCh38
rs563997147 Chr.12: 123083784-123083784 on Build GRCh38
rs1007374791 Chr.12: 123084069-123084069 on Build GRCh38
rs904756189 Chr.12: 123083903-123083903 on Build GRCh38
rs192161344 Chr.12: 123083901-123083901 on Build GRCh38
rs948852128 Chr.12: 123083851-123083851 on Build GRCh38
rs953335007 Chr.12: 123084056-123084056 on Build GRCh38
rs779519164 Chr.12: 123083841-123083841 on Build GRCh38
rs989975426 Chr.12: 123084043-123084043 on Build GRCh38
rs920819471 Chr.12: 123084076-123084076 on Build GRCh38
rs1031553467 Chr.12: 123083954-123083954 on Build GRCh38

View Full Product Details