Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36365351-36365856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPG20
Entrez Gene ID
23111
Gene Name
spastic paraplegia 20 (Troyer syndrome)
Gene Aliases
SPARTIN, TAHCCP1
UniGene
Hs.440414
Gene Chromosome Location
Chr.13: 36301638-36370180 on Build GRCh38
Transcripts
NM_001142294.1 NR_045180.1 NR_045181.1

Gene Symbol
SPG20-AS1
Entrez Gene ID
100507135
Gene Name
SPG20 antisense RNA 1
Gene Aliases
C13orf43, SPG20OS
UniGene
Hs.631781
Gene Chromosome Location
Chr.13: 36346431-36369735 on Build GRCh38
Transcripts
NM_001142294.1 NR_045180.1 NR_045181.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6358766 c.78C>A p.A26A Chr.13: 36365514-36365514 on Build GRCh38
7240912 c.115C>T p.P39S Chr.13: 36365551-36365551 on Build GRCh38
5495905 c.153G>A p.M51I Chr.13: 36365589-36365589 on Build GRCh38
5650467 c.260C>G p.S87C Chr.13: 36365821-36365821 on Build GRCh38
4914869 c.223G>C p.G75R Chr.13: 36365784-36365784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902576136 Chr.13: 36365748-36365748 on Build GRCh38
rs138518561 Chr.13: 36365690-36365690 on Build GRCh38
rs929729403 Chr.13: 36365534-36365534 on Build GRCh38
rs747371517 Chr.13: 36365766-36365766 on Build GRCh38
rs144909537 Chr.13: 36365395-36365395 on Build GRCh38
rs550807884 Chr.13: 36365678-36365678 on Build GRCh38
rs553095493 Chr.13: 36365712-36365713 on Build GRCh38
rs762625229 Chr.13: 36365541-36365541 on Build GRCh38
rs778899756 Chr.13: 36365645-36365645 on Build GRCh38
rs918316881 Chr.13: 36365501-36365501 on Build GRCh38
rs1000022833 Chr.13: 36365755-36365755 on Build GRCh38
rs199938032 Chr.13: 36365592-36365592 on Build GRCh38
rs540408813 Chr.13: 36365555-36365555 on Build GRCh38
rs367781902 Chr.13: 36365759-36365759 on Build GRCh38
rs557936993 Chr.13: 36365549-36365549 on Build GRCh38
rs991186183 Chr.13: 36365415-36365415 on Build GRCh38
rs924837893 Chr.13: 36365647-36365647 on Build GRCh38
rs963113137 Chr.13: 36365556-36365556 on Build GRCh38
rs1023582897 Chr.13: 36365832-36365832 on Build GRCh38
rs183590044 Chr.13: 36365745-36365745 on Build GRCh38
rs748260482 Chr.13: 36365683-36365683 on Build GRCh38
rs966496912 Chr.13: 36365383-36365383 on Build GRCh38
rs755958098 Chr.13: 36365811-36365811 on Build GRCh38
rs561264724 Chr.13: 36365550-36365550 on Build GRCh38
rs1051731657 Chr.13: 36365732-36365732 on Build GRCh38
rs916244122 Chr.13: 36365421-36365421 on Build GRCh38
rs3736920 Chr.13: 36365621-36365621 on Build GRCh38
rs3736919 Chr.13: 36365551-36365551 on Build GRCh38
rs957944655 Chr.13: 36365401-36365401 on Build GRCh38
rs533847438 Chr.13: 36365576-36365576 on Build GRCh38
rs573706721 Chr.13: 36365767-36365767 on Build GRCh38
rs997399162 Chr.13: 36365469-36365469 on Build GRCh38
rs192275381 Chr.13: 36365465-36365465 on Build GRCh38
rs887093651 Chr.13: 36365540-36365540 on Build GRCh38
rs893666934 Chr.13: 36365798-36365798 on Build GRCh38
rs981840815 Chr.13: 36365448-36365448 on Build GRCh38
rs767313770 Chr.13: 36365605-36365605 on Build GRCh38
rs999340949 Chr.13: 36365389-36365389 on Build GRCh38
rs1054234560 Chr.13: 36365413-36365415 on Build GRCh38
rs971630512 Chr.13: 36365599-36365599 on Build GRCh38
rs778019158 Chr.13: 36365697-36365697 on Build GRCh38
rs188888044 Chr.13: 36365424-36365424 on Build GRCh38

View Full Product Details