Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 100847158-100847687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEG3
Entrez Gene ID
55384
Gene Name
maternally expressed 3 (non-protein coding)
Gene Aliases
FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
UniGene
Hs.654863
Gene Chromosome Location
Chr.14: 100826108-100861026 on Build GRCh38
Transcripts
NR_002766.2 NR_003530.2 NR_003531.3 NR_033358.1 NR_033359.1 NR_033360.1 NR_046464.1 NR_046465.2 NR_046466.1 NR_046467.1 NR_046469.1 NR_046470.2 NR_046471.1 NR_046472.1 NR_046473.1

SNP Detail

SNP ID Chromosome Location
rs548458889 Chr.14: 100847583-100847583 on Build GRCh38
rs886898709 Chr.14: 100847316-100847316 on Build GRCh38
rs966439640 Chr.14: 100847290-100847290 on Build GRCh38
rs564370773 Chr.14: 100847408-100847408 on Build GRCh38
rs1029267926 Chr.14: 100847353-100847353 on Build GRCh38
rs555912240 Chr.14: 100847184-100847184 on Build GRCh38
rs879725051 Chr.14: 100847639-100847639 on Build GRCh38
rs898388949 Chr.14: 100847291-100847291 on Build GRCh38
rs939397565 Chr.14: 100847664-100847664 on Build GRCh38
rs578032215 Chr.14: 100847492-100847492 on Build GRCh38
rs555641514 Chr.14: 100847205-100847205 on Build GRCh38
rs979629078 Chr.14: 100847443-100847443 on Build GRCh38
rs1055273824 Chr.14: 100847204-100847204 on Build GRCh38
rs767106549 Chr.14: 100847354-100847354 on Build GRCh38
rs1019705378 Chr.14: 100847318-100847318 on Build GRCh38
rs1045460694 Chr.14: 100847595-100847595 on Build GRCh38
rs1046873147 Chr.14: 100847309-100847309 on Build GRCh38
rs963717848 Chr.14: 100847336-100847336 on Build GRCh38
rs3742394 Chr.14: 100847539-100847539 on Build GRCh38
rs755018187 Chr.14: 100847324-100847324 on Build GRCh38
rs1022962057 Chr.14: 100847275-100847275 on Build GRCh38
rs1044967627 Chr.14: 100847288-100847288 on Build GRCh38
rs534641991 Chr.14: 100847290-100847290 on Build GRCh38
rs543586671 Chr.14: 100847521-100847521 on Build GRCh38
rs907766458 Chr.14: 100847535-100847535 on Build GRCh38
rs996849383 Chr.14: 100847337-100847337 on Build GRCh38
rs45439095 Chr.14: 100847305-100847305 on Build GRCh38
rs3742395 Chr.14: 100847649-100847649 on Build GRCh38
rs956785714 Chr.14: 100847525-100847525 on Build GRCh38
rs562669625 Chr.14: 100847532-100847532 on Build GRCh38
rs150902250 Chr.14: 100847317-100847317 on Build GRCh38
rs972681611 Chr.14: 100847548-100847548 on Build GRCh38
rs370065924 Chr.14: 100847435-100847435 on Build GRCh38
rs766311710 Chr.14: 100847268-100847268 on Build GRCh38
rs572747322 Chr.14: 100847446-100847446 on Build GRCh38

View Full Product Details