Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 100982814-100983320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNHG24
Entrez Gene ID
101929369
Gene Name
small nucleolar RNA host gene 24
Gene Aliases
-
UniGene
Hs.118101
Gene Chromosome Location
Chr.14: 100967652-100986650 on Build GRCh38
Transcripts
NR_110178.1 NR_003215.1

Gene Symbol
SNORD114-22
Entrez Gene ID
767600
Gene Name
small nucleolar RNA, C/D box 114-22
Gene Aliases
14q(II-22)
UniGene
Gene Chromosome Location
Chr.14: 100982926-100982996 on Build GRCh38
Transcripts
NR_110178.1 NR_003215.1

SNP Detail

SNP ID Chromosome Location
rs751739805 Chr.14: 100982992-100982992 on Build GRCh38
rs762991706 Chr.14: 100982986-100982986 on Build GRCh38
rs905633801 Chr.14: 100983291-100983291 on Build GRCh38
rs1031406100 Chr.14: 100982906-100982906 on Build GRCh38
rs756637061 Chr.14: 100982998-100982998 on Build GRCh38
rs557622559 Chr.14: 100983253-100983253 on Build GRCh38
rs752726409 Chr.14: 100982931-100982931 on Build GRCh38
rs376016873 Chr.14: 100983028-100983028 on Build GRCh38
rs764269913 Chr.14: 100982988-100982988 on Build GRCh38
rs759031754 Chr.14: 100983074-100983074 on Build GRCh38
rs751355158 Chr.14: 100982860-100982860 on Build GRCh38
rs980373258 Chr.14: 100983144-100983144 on Build GRCh38
rs34924174 Chr.14: 100983051-100983051 on Build GRCh38
rs936941464 Chr.14: 100982905-100982905 on Build GRCh38
rs199765872 Chr.14: 100982950-100982950 on Build GRCh38
rs773156518 Chr.14: 100982965-100982965 on Build GRCh38
rs761718233 Chr.14: 100982901-100982901 on Build GRCh38
rs765304553 Chr.14: 100982928-100982928 on Build GRCh38
rs933544126 Chr.14: 100983192-100983192 on Build GRCh38
rs914117176 Chr.14: 100982924-100982924 on Build GRCh38
rs750754778 Chr.14: 100982996-100982996 on Build GRCh38
rs955792087 Chr.14: 100982929-100982929 on Build GRCh38
rs778027397 Chr.14: 100982935-100982935 on Build GRCh38
rs748805567 Chr.14: 100983007-100983007 on Build GRCh38
rs573651617 Chr.14: 100982916-100982916 on Build GRCh38
rs768245585 Chr.14: 100983020-100983020 on Build GRCh38
rs536315811 Chr.14: 100983119-100983119 on Build GRCh38
rs781475903 Chr.14: 100982995-100982995 on Build GRCh38
rs117820428 Chr.14: 100982944-100982944 on Build GRCh38
rs959967685 Chr.14: 100983159-100983159 on Build GRCh38
rs747747983 Chr.14: 100983024-100983024 on Build GRCh38
rs890786802 Chr.14: 100982870-100982870 on Build GRCh38
rs766995703 Chr.14: 100983177-100983177 on Build GRCh38
rs181982977 Chr.14: 100983022-100983022 on Build GRCh38
rs774190849 Chr.14: 100982953-100982953 on Build GRCh38
rs988274868 Chr.14: 100983187-100983187 on Build GRCh38
rs530794658 Chr.14: 100983027-100983027 on Build GRCh38
rs891516859 Chr.14: 100983066-100983066 on Build GRCh38
rs748822857 Chr.14: 100983015-100983015 on Build GRCh38
rs758643404 Chr.14: 100982932-100982932 on Build GRCh38
rs983390104 Chr.14: 100983172-100983172 on Build GRCh38
rs546700994 Chr.14: 100983142-100983142 on Build GRCh38
rs770879880 Chr.14: 100983044-100983044 on Build GRCh38
rs567577794 Chr.14: 100983080-100983080 on Build GRCh38
rs898878443 Chr.14: 100983254-100983254 on Build GRCh38
rs774225712 Chr.14: 100982892-100982892 on Build GRCh38
rs1035554886 Chr.14: 100983150-100983150 on Build GRCh38
rs201132403 Chr.14: 100982943-100982943 on Build GRCh38
rs1025379078 Chr.14: 100983048-100983048 on Build GRCh38
rs1007873258 Chr.14: 100983031-100983031 on Build GRCh38
rs780479967 Chr.14: 100983000-100983000 on Build GRCh38
rs914170158 Chr.14: 100983189-100983189 on Build GRCh38
rs980845446 Chr.14: 100983069-100983069 on Build GRCh38
rs370928850 Chr.14: 100982994-100982994 on Build GRCh38
rs374584252 Chr.14: 100982936-100982936 on Build GRCh38
rs980041667 Chr.14: 100982945-100982945 on Build GRCh38
rs1011692416 Chr.14: 100983262-100983262 on Build GRCh38
rs781567663 Chr.14: 100982940-100982940 on Build GRCh38
rs528699800 Chr.14: 100982960-100982960 on Build GRCh38
rs181290142 Chr.14: 100982942-100982942 on Build GRCh38
rs969530290 Chr.14: 100983058-100983058 on Build GRCh38
rs773912254 Chr.14: 100983029-100983029 on Build GRCh38
rs565491859 Chr.14: 100983173-100983173 on Build GRCh38
rs760596800 Chr.14: 100982968-100982968 on Build GRCh38
rs398044040 Chr.14: 100983056-100983056 on Build GRCh38
rs929157452 Chr.14: 100983257-100983257 on Build GRCh38
rs138232754 Chr.14: 100983209-100983209 on Build GRCh38
rs185933136 Chr.14: 100982951-100982951 on Build GRCh38
rs368491650 Chr.14: 100983200-100983200 on Build GRCh38
rs752810623 Chr.14: 100982975-100982975 on Build GRCh38
rs891464795 Chr.14: 100983258-100983258 on Build GRCh38
rs910194527 Chr.14: 100983227-100983227 on Build GRCh38
rs1047533661 Chr.14: 100983264-100983264 on Build GRCh38
rs767517677 Chr.14: 100982914-100982914 on Build GRCh38
rs760283689 Chr.14: 100983228-100983228 on Build GRCh38
rs1011327548 Chr.14: 100983130-100983130 on Build GRCh38
rs547843136 Chr.14: 100983107-100983107 on Build GRCh38
rs932149275 Chr.14: 100983217-100983217 on Build GRCh38
rs376512105 Chr.14: 100982962-100982962 on Build GRCh38
rs190698525 Chr.14: 100983012-100983012 on Build GRCh38
rs369650892 Chr.14: 100983284-100983284 on Build GRCh38
rs766294874 Chr.14: 100982974-100982974 on Build GRCh38
rs770457811 Chr.14: 100983038-100983039 on Build GRCh38

View Full Product Details