Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 56886068-56886565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC145783
Entrez Gene ID
145783
Gene Name
uncharacterized LOC145783
Gene Aliases
-
UniGene
Hs.620525
Gene Chromosome Location
Chr.15: 56886170-56918499 on Build GRCh38
Transcripts
NR_015419.2

SNP Detail

SNP ID Chromosome Location
rs761418299 Chr.15: 56886435-56886435 on Build GRCh38
rs945605021 Chr.15: 56886300-56886300 on Build GRCh38
rs200701628 Chr.15: 56886327-56886331 on Build GRCh38
rs1057280863 Chr.15: 56886334-56886334 on Build GRCh38
rs763868054 Chr.15: 56886336-56886336 on Build GRCh38
rs536032501 Chr.15: 56886240-56886240 on Build GRCh38
rs796504564 Chr.15: 56886302-56886302 on Build GRCh38
rs965072171 Chr.15: 56886269-56886269 on Build GRCh38
rs1026126553 Chr.15: 56886194-56886194 on Build GRCh38
rs928100999 Chr.15: 56886326-56886326 on Build GRCh38
rs954777353 Chr.15: 56886204-56886204 on Build GRCh38
rs993342439 Chr.15: 56886187-56886187 on Build GRCh38
rs12439868 Chr.15: 56886217-56886217 on Build GRCh38
rs978248482 Chr.15: 56886311-56886311 on Build GRCh38
rs578098647 Chr.15: 56886509-56886509 on Build GRCh38
rs751043986 Chr.15: 56886415-56886415 on Build GRCh38
rs1012874876 Chr.15: 56886155-56886155 on Build GRCh38
rs759585593 Chr.15: 56886271-56886271 on Build GRCh38
rs36036066 Chr.15: 56886286-56886287 on Build GRCh38
rs769444424 Chr.15: 56886282-56886282 on Build GRCh38
rs915466793 Chr.15: 56886297-56886297 on Build GRCh38
rs948375053 Chr.15: 56886373-56886373 on Build GRCh38
rs538344831 Chr.15: 56886459-56886459 on Build GRCh38
rs938410619 Chr.15: 56886123-56886123 on Build GRCh38
rs1017640478 Chr.15: 56886252-56886252 on Build GRCh38
rs371561007 Chr.15: 56886196-56886196 on Build GRCh38
rs964770664 Chr.15: 56886462-56886462 on Build GRCh38
rs1037340521 Chr.15: 56886374-56886374 on Build GRCh38
rs756697210 Chr.15: 56886494-56886494 on Build GRCh38
rs899025326 Chr.15: 56886182-56886182 on Build GRCh38
rs1047605275 Chr.15: 56886432-56886432 on Build GRCh38
rs987522199 Chr.15: 56886250-56886250 on Build GRCh38
rs746746158 Chr.15: 56886317-56886317 on Build GRCh38
rs1056807305 Chr.15: 56886129-56886129 on Build GRCh38
rs775402889 Chr.15: 56886301-56886301 on Build GRCh38
rs899097693 Chr.15: 56886404-56886404 on Build GRCh38
rs74018416 Chr.15: 56886472-56886472 on Build GRCh38
rs993373434 Chr.15: 56886426-56886426 on Build GRCh38
rs11855646 Chr.15: 56886330-56886330 on Build GRCh38
rs1041477753 Chr.15: 56886090-56886090 on Build GRCh38
rs201173102 Chr.15: 56886251-56886251 on Build GRCh38
rs1009023971 Chr.15: 56886439-56886439 on Build GRCh38
rs1016576188 Chr.15: 56886157-56886157 on Build GRCh38
rs902953922 Chr.15: 56886102-56886102 on Build GRCh38
rs894124904 Chr.15: 56886142-56886142 on Build GRCh38
rs915602245 Chr.15: 56886344-56886344 on Build GRCh38
rs569212103 Chr.15: 56886369-56886369 on Build GRCh38

View Full Product Details