Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 526743-527242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00235
Entrez Gene ID
64493
Gene Name
long intergenic non-protein coding RNA 235
Gene Aliases
C16orf10, LA16c-356B8.1, NCRNA00235
UniGene
Hs.721553
Gene Chromosome Location
Chr.16: 526847-527407 on Build GRCh38
Transcripts
NR_024121.1

SNP Detail

SNP ID Chromosome Location
rs3913741 Chr.16: 527068-527068 on Build GRCh38
rs964460743 Chr.16: 527023-527023 on Build GRCh38
rs910571776 Chr.16: 527022-527022 on Build GRCh38
rs1024911258 Chr.16: 526971-526971 on Build GRCh38
rs878939696 Chr.16: 527046-527046 on Build GRCh38
rs1040616185 Chr.16: 526799-526799 on Build GRCh38
rs964754245 Chr.16: 527040-527040 on Build GRCh38
rs927986021 Chr.16: 527094-527094 on Build GRCh38
rs943639853 Chr.16: 526793-526793 on Build GRCh38
rs923549363 Chr.16: 527063-527063 on Build GRCh38
rs760315793 Chr.16: 527029-527029 on Build GRCh38
rs1051282859 Chr.16: 527191-527191 on Build GRCh38
rs187710489 Chr.16: 526769-526769 on Build GRCh38
rs1008639918 Chr.16: 526986-526986 on Build GRCh38
rs562266004 Chr.16: 526983-526983 on Build GRCh38
rs529451747 Chr.16: 527071-527071 on Build GRCh38
rs1044306167 Chr.16: 527162-527162 on Build GRCh38
rs929537003 Chr.16: 527073-527073 on Build GRCh38
rs555386291 Chr.16: 526780-526780 on Build GRCh38
rs941789532 Chr.16: 527128-527128 on Build GRCh38
rs567409974 Chr.16: 526833-526833 on Build GRCh38
rs1011498028 Chr.16: 526924-526924 on Build GRCh38
rs537669237 Chr.16: 526849-526849 on Build GRCh38
rs138740746 Chr.16: 526944-526944 on Build GRCh38
rs902666482 Chr.16: 527163-527163 on Build GRCh38
rs920417258 Chr.16: 527078-527078 on Build GRCh38
rs770629779 Chr.16: 526821-526821 on Build GRCh38
rs753341870 Chr.16: 527181-527181 on Build GRCh38
rs895357818 Chr.16: 526881-526881 on Build GRCh38
rs772841852 Chr.16: 527008-527008 on Build GRCh38
rs563101045 Chr.16: 527127-527127 on Build GRCh38
rs544733090 Chr.16: 526938-526938 on Build GRCh38
rs987167856 Chr.16: 526772-526772 on Build GRCh38
rs1043869905 Chr.16: 526929-526929 on Build GRCh38
rs973658161 Chr.16: 527039-527039 on Build GRCh38
rs990800871 Chr.16: 527142-527142 on Build GRCh38
rs555805666 Chr.16: 526871-526871 on Build GRCh38
rs894799978 Chr.16: 526907-526907 on Build GRCh38
rs371928359 Chr.16: 527023-527026 on Build GRCh38
rs746092100 Chr.16: 527009-527009 on Build GRCh38
rs910548157 Chr.16: 526792-526792 on Build GRCh38
rs1046454493 Chr.16: 527218-527218 on Build GRCh38
rs577303812 Chr.16: 526875-526875 on Build GRCh38
rs902063972 Chr.16: 526806-526806 on Build GRCh38
rs1055209172 Chr.16: 526874-526874 on Build GRCh38
rs746861405 Chr.16: 526819-526819 on Build GRCh38
rs950636391 Chr.16: 527079-527079 on Build GRCh38
rs929477920 Chr.16: 526813-526813 on Build GRCh38
rs1013255915 Chr.16: 527182-527182 on Build GRCh38
rs766064194 Chr.16: 527126-527126 on Build GRCh38
rs939333118 Chr.16: 527139-527139 on Build GRCh38
rs990173437 Chr.16: 527010-527010 on Build GRCh38
rs571954576 Chr.16: 526978-526978 on Build GRCh38
rs902780184 Chr.16: 526932-526932 on Build GRCh38
rs544801992 Chr.16: 527081-527081 on Build GRCh38
rs887561524 Chr.16: 526836-526836 on Build GRCh38
rs190840197 Chr.16: 526979-526979 on Build GRCh38
rs939447670 Chr.16: 526861-526861 on Build GRCh38
rs891337625 Chr.16: 527206-527206 on Build GRCh38
rs909361743 Chr.16: 527118-527118 on Build GRCh38
rs964898743 Chr.16: 526791-526791 on Build GRCh38
rs770010881 Chr.16: 527147-527147 on Build GRCh38
rs894866367 Chr.16: 527174-527174 on Build GRCh38
rs566917953 Chr.16: 526764-526764 on Build GRCh38
rs916664185 Chr.16: 527145-527145 on Build GRCh38
rs1055222587 Chr.16: 526855-526855 on Build GRCh38

View Full Product Details