Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81667010-81667509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMIP
Entrez Gene ID
80790
Gene Name
c-Maf inducing protein
Gene Aliases
TCMIP
UniGene
Hs.594095
Gene Chromosome Location
Chr.16: 81445170-81711762 on Build GRCh38
Transcripts
NM_030629.2 NM_198390.2 XM_005256179.4 XM_005256181.2 XM_005256182.1 XM_011523352.1 XM_011523353.1 XM_017023733.1 NR_045112.1

Gene Symbol
LOC100129617
Entrez Gene ID
100129617
Gene Name
uncharacterized LOC100129617
Gene Aliases
-
UniGene
Hs.657151
Gene Chromosome Location
Chr.16: 81665354-81667276 on Build GRCh38
Transcripts
NM_030629.2 NM_198390.2 XM_005256179.4 XM_005256181.2 XM_005256182.1 XM_011523352.1 XM_011523353.1 XM_017023733.1 NR_045112.1

SNP Detail

SNP ID Chromosome Location
rs948010815 Chr.16: 81667363-81667363 on Build GRCh38
rs1040673771 Chr.16: 81667244-81667244 on Build GRCh38
rs144082144 Chr.16: 81667428-81667428 on Build GRCh38
rs915371669 Chr.16: 81667183-81667183 on Build GRCh38
rs966236649 Chr.16: 81667379-81667379 on Build GRCh38
rs534691022 Chr.16: 81667110-81667110 on Build GRCh38
rs920949594 Chr.16: 81667275-81667278 on Build GRCh38
rs112066886 Chr.16: 81667189-81667189 on Build GRCh38
rs137939191 Chr.16: 81667368-81667368 on Build GRCh38
rs974969918 Chr.16: 81667388-81667388 on Build GRCh38
rs1052713534 Chr.16: 81667390-81667390 on Build GRCh38
rs911757749 Chr.16: 81667254-81667254 on Build GRCh38
rs148815953 Chr.16: 81667120-81667120 on Build GRCh38
rs987191491 Chr.16: 81667209-81667209 on Build GRCh38
rs192036627 Chr.16: 81667471-81667471 on Build GRCh38
rs181125117 Chr.16: 81667119-81667119 on Build GRCh38
rs989583128 Chr.16: 81667179-81667179 on Build GRCh38
rs764846095 Chr.16: 81667352-81667352 on Build GRCh38
rs766311949 Chr.16: 81667316-81667316 on Build GRCh38
rs930507534 Chr.16: 81667331-81667331 on Build GRCh38
rs373716404 Chr.16: 81667378-81667378 on Build GRCh38
rs1022793598 Chr.16: 81667367-81667367 on Build GRCh38
rs897211609 Chr.16: 81667100-81667100 on Build GRCh38
rs978317231 Chr.16: 81667194-81667194 on Build GRCh38
rs1033709574 Chr.16: 81667155-81667155 on Build GRCh38
rs1046403346 Chr.16: 81667369-81667369 on Build GRCh38
rs532793497 Chr.16: 81667299-81667299 on Build GRCh38
rs1055054244 Chr.16: 81667450-81667450 on Build GRCh38
rs114456149 Chr.16: 81667303-81667303 on Build GRCh38
rs866493359 Chr.16: 81667128-81667128 on Build GRCh38
rs183860525 Chr.16: 81667139-81667139 on Build GRCh38
rs757242498 Chr.16: 81667181-81667185 on Build GRCh38
rs970710756 Chr.16: 81667129-81667129 on Build GRCh38
rs1022317876 Chr.16: 81667473-81667474 on Build GRCh38
rs115552139 Chr.16: 81667115-81667115 on Build GRCh38
rs1128658 Chr.16: 81667203-81667203 on Build GRCh38
rs996578862 Chr.16: 81667111-81667112 on Build GRCh38
rs893963666 Chr.16: 81667401-81667401 on Build GRCh38
rs374168999 Chr.16: 81667145-81667145 on Build GRCh38
rs182490967 Chr.16: 81667371-81667371 on Build GRCh38
rs895096629 Chr.16: 81667451-81667451 on Build GRCh38
rs944981319 Chr.16: 81667228-81667228 on Build GRCh38
rs1036891294 Chr.16: 81667302-81667302 on Build GRCh38
rs188182385 Chr.16: 81667325-81667325 on Build GRCh38
rs999801414 Chr.16: 81667372-81667372 on Build GRCh38
rs1048141727 Chr.16: 81667283-81667283 on Build GRCh38
rs959362711 Chr.16: 81667336-81667336 on Build GRCh38
rs932366628 Chr.16: 81667282-81667282 on Build GRCh38
rs989208802 Chr.16: 81667180-81667180 on Build GRCh38
rs187124759 Chr.16: 81667404-81667404 on Build GRCh38
rs957752276 Chr.16: 81667152-81667152 on Build GRCh38
rs1011112251 Chr.16: 81667409-81667409 on Build GRCh38
rs566836102 Chr.16: 81667214-81667214 on Build GRCh38
rs943692392 Chr.16: 81667054-81667054 on Build GRCh38
rs959415057 Chr.16: 81667164-81667164 on Build GRCh38
rs35168185 Chr.16: 81667470-81667471 on Build GRCh38
rs76453765 Chr.16: 81667127-81667127 on Build GRCh38
rs62043839 Chr.16: 81667419-81667419 on Build GRCh38
rs747380767 Chr.16: 81667439-81667439 on Build GRCh38
rs1048100403 Chr.16: 81667413-81667413 on Build GRCh38
rs1048456146 Chr.16: 81667334-81667334 on Build GRCh38
rs527593091 Chr.16: 81667435-81667435 on Build GRCh38
rs755746664 Chr.16: 81667470-81667470 on Build GRCh38
rs1037244348 Chr.16: 81667432-81667432 on Build GRCh38
rs370442613 Chr.16: 81667356-81667356 on Build GRCh38
rs147338352 Chr.16: 81667442-81667442 on Build GRCh38
rs543254946 Chr.16: 81667111-81667111 on Build GRCh38
rs916702583 Chr.16: 81667353-81667353 on Build GRCh38
rs1012237922 Chr.16: 81667118-81667118 on Build GRCh38

View Full Product Details