Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 86488096-86488605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FENDRR
Entrez Gene ID
400550
Gene Name
FOXF1 adjacent non-coding developmental regulatory RNA
Gene Aliases
FOXF1-AS1, TCONS_00024240, lincFOXF1, onco-lncRNA-21
UniGene
Hs.448825
Gene Chromosome Location
Chr.16: 86474525-86508860 on Build GRCh38
Transcripts
NR_033925.1 NR_036444.1

SNP Detail

SNP ID Chromosome Location
rs186296384 Chr.16: 86488477-86488477 on Build GRCh38
rs550492308 Chr.16: 86488184-86488184 on Build GRCh38
rs1001091190 Chr.16: 86488318-86488318 on Build GRCh38
rs973616188 Chr.16: 86488186-86488186 on Build GRCh38
rs371023761 Chr.16: 86488201-86488201 on Build GRCh38
rs938724752 Chr.16: 86488400-86488400 on Build GRCh38
rs951011715 Chr.16: 86488394-86488394 on Build GRCh38
rs553008517 Chr.16: 86488557-86488557 on Build GRCh38
rs908222457 Chr.16: 86488257-86488257 on Build GRCh38
rs115423814 Chr.16: 86488182-86488182 on Build GRCh38
rs9923388 Chr.16: 86488404-86488404 on Build GRCh38
rs758107929 Chr.16: 86488561-86488562 on Build GRCh38
rs929558443 Chr.16: 86488143-86488143 on Build GRCh38
rs1033756747 Chr.16: 86488347-86488347 on Build GRCh38
rs982827932 Chr.16: 86488247-86488247 on Build GRCh38
rs75201375 Chr.16: 86488367-86488367 on Build GRCh38
rs564355484 Chr.16: 86488178-86488185 on Build GRCh38
rs765719346 Chr.16: 86488346-86488346 on Build GRCh38
rs1009450693 Chr.16: 86488294-86488294 on Build GRCh38
rs778709360 Chr.16: 86488493-86488495 on Build GRCh38
rs758700116 Chr.16: 86488396-86488396 on Build GRCh38
rs1042349689 Chr.16: 86488416-86488416 on Build GRCh38
rs1027888013 Chr.16: 86488190-86488190 on Build GRCh38
rs781031688 Chr.16: 86488486-86488486 on Build GRCh38
rs546002255 Chr.16: 86488521-86488521 on Build GRCh38
rs116084373 Chr.16: 86488298-86488298 on Build GRCh38
rs535526633 Chr.16: 86488518-86488518 on Build GRCh38
rs903659556 Chr.16: 86488302-86488302 on Build GRCh38
rs976496142 Chr.16: 86488405-86488405 on Build GRCh38
rs1003647208 Chr.16: 86488150-86488150 on Build GRCh38
rs573005277 Chr.16: 86488577-86488577 on Build GRCh38
rs906517706 Chr.16: 86488537-86488537 on Build GRCh38
rs945325816 Chr.16: 86488338-86488338 on Build GRCh38
rs750904194 Chr.16: 86488315-86488315 on Build GRCh38
rs568647904 Chr.16: 86488403-86488403 on Build GRCh38
rs549889297 Chr.16: 86488157-86488157 on Build GRCh38
rs932511212 Chr.16: 86488324-86488324 on Build GRCh38
rs989282490 Chr.16: 86488484-86488484 on Build GRCh38
rs932460316 Chr.16: 86488475-86488475 on Build GRCh38
rs1000742408 Chr.16: 86488443-86488443 on Build GRCh38
rs994998399 Chr.16: 86488370-86488370 on Build GRCh38
rs766848129 Chr.16: 86488132-86488132 on Build GRCh38
rs1039279560 Chr.16: 86488453-86488453 on Build GRCh38
rs555447123 Chr.16: 86488377-86488377 on Build GRCh38
rs866075278 Chr.16: 86488344-86488344 on Build GRCh38
rs909404838 Chr.16: 86488356-86488356 on Build GRCh38
rs1015643103 Chr.16: 86488402-86488402 on Build GRCh38
rs945246370 Chr.16: 86488507-86488507 on Build GRCh38
rs1027793735 Chr.16: 86488382-86488382 on Build GRCh38
rs757196715 Chr.16: 86488442-86488442 on Build GRCh38
rs952402004 Chr.16: 86488196-86488196 on Build GRCh38
rs938730645 Chr.16: 86488230-86488230 on Build GRCh38
rs1053684642 Chr.16: 86488333-86488333 on Build GRCh38
rs997233057 Chr.16: 86488343-86488343 on Build GRCh38
rs1042755823 Chr.16: 86488342-86488342 on Build GRCh38
rs943747388 Chr.16: 86488263-86488263 on Build GRCh38
rs927439581 Chr.16: 86488389-86488389 on Build GRCh38
rs755052690 Chr.16: 86488565-86488565 on Build GRCh38
rs900252909 Chr.16: 86488470-86488470 on Build GRCh38
rs769525312 Chr.16: 86488523-86488523 on Build GRCh38
rs983640538 Chr.16: 86488355-86488355 on Build GRCh38
rs976507503 Chr.16: 86488274-86488274 on Build GRCh38
rs777903450 Chr.16: 86488399-86488399 on Build GRCh38
rs138218741 Chr.16: 86488334-86488334 on Build GRCh38
rs557251435 Chr.16: 86488419-86488419 on Build GRCh38
rs780230744 Chr.16: 86488487-86488487 on Build GRCh38
rs570663267 Chr.16: 86488185-86488185 on Build GRCh38

View Full Product Details