Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7017506-7017999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf49
Entrez Gene ID
124944
Gene Name
chromosome 17 open reading frame 49
Gene Aliases
BAP18, HEPIS
UniGene
Hs.511801
Gene Chromosome Location
Chr.17: 7014737-7017524 on Build GRCh38
Transcripts
NM_001142798.2 NM_001142799.2 NM_174893.3 NR_029712.1 NR_030178.1 NR_038310.1 NR_037717.1

Gene Symbol
MIR195
Entrez Gene ID
406971
Gene Name
microRNA 195
Gene Aliases
MIRN195, miRNA195, mir-195
UniGene
Gene Chromosome Location
Chr.17: 7017615-7017701 on Build GRCh38
Transcripts
NM_001142798.2 NM_001142799.2 NM_174893.3 NR_029712.1 NR_030178.1 NR_038310.1 NR_037717.1

Gene Symbol
MIR497
Entrez Gene ID
574456
Gene Name
microRNA 497
Gene Aliases
MIRN497, hsa-mir-497, mir-497
UniGene
Gene Chromosome Location
Chr.17: 7017911-7018022 on Build GRCh38
Transcripts
NM_001142798.2 NM_001142799.2 NM_174893.3 NR_029712.1 NR_030178.1 NR_038310.1 NR_037717.1

Gene Symbol
MIR497HG
Entrez Gene ID
100506755
Gene Name
mir-497-195 cluster host gene
Gene Aliases
MIR195HG
UniGene
Hs.710442
Gene Chromosome Location
Chr.17: 7015818-7019654 on Build GRCh38
Transcripts
NM_001142798.2 NM_001142799.2 NM_174893.3 NR_029712.1 NR_030178.1 NR_038310.1 NR_037717.1

Gene Symbol
RNASEK-C17orf49
Entrez Gene ID
100529209
Gene Name
RNASEK-C17orf49 readthrough
Gene Aliases
-
UniGene
Hs.511801
Gene Chromosome Location
Chr.17: 7012417-7017524 on Build GRCh38
Transcripts
NM_001142798.2 NM_001142799.2 NM_174893.3 NR_029712.1 NR_030178.1 NR_038310.1 NR_037717.1

SNP Detail

SNP ID Chromosome Location
rs748112857 Chr.17: 7017643-7017643 on Build GRCh38
rs759100327 Chr.17: 7017662-7017662 on Build GRCh38
rs918633347 Chr.17: 7017760-7017760 on Build GRCh38
rs975705809 Chr.17: 7017668-7017668 on Build GRCh38
rs1054818867 Chr.17: 7017910-7017910 on Build GRCh38
rs770696988 Chr.17: 7017912-7017912 on Build GRCh38
rs940662829 Chr.17: 7017917-7017917 on Build GRCh38
rs758977398 Chr.17: 7017577-7017577 on Build GRCh38
rs1055637087 Chr.17: 7017621-7017621 on Build GRCh38
rs1010038510 Chr.17: 7017914-7017914 on Build GRCh38
rs1045712676 Chr.17: 7017624-7017624 on Build GRCh38
rs371654899 Chr.17: 7017690-7017690 on Build GRCh38
rs370178733 Chr.17: 7017750-7017750 on Build GRCh38
rs905765974 Chr.17: 7017626-7017626 on Build GRCh38
rs901567473 Chr.17: 7017936-7017936 on Build GRCh38
rs376953960 Chr.17: 7017671-7017671 on Build GRCh38
rs561346783 Chr.17: 7017735-7017735 on Build GRCh38
rs751401247 Chr.17: 7017542-7017542 on Build GRCh38
rs752245895 Chr.17: 7017721-7017721 on Build GRCh38
rs766120856 Chr.17: 7017585-7017585 on Build GRCh38
rs755634302 Chr.17: 7017939-7017940 on Build GRCh38
rs755637140 Chr.17: 7017809-7017809 on Build GRCh38
rs781050107 Chr.17: 7017636-7017636 on Build GRCh38
rs986142931 Chr.17: 7017714-7017714 on Build GRCh38
rs1037716604 Chr.17: 7017923-7017923 on Build GRCh38
rs779240683 Chr.17: 7017605-7017605 on Build GRCh38
rs758618444 Chr.17: 7017716-7017716 on Build GRCh38
rs41283391 Chr.17: 7017569-7017569 on Build GRCh38
rs912928493 Chr.17: 7017533-7017533 on Build GRCh38
rs997278260 Chr.17: 7017949-7017949 on Build GRCh38
rs371979786 Chr.17: 7017657-7017657 on Build GRCh38
rs773079799 Chr.17: 7017651-7017651 on Build GRCh38
rs368631457 Chr.17: 7017596-7017596 on Build GRCh38
rs757629737 Chr.17: 7017693-7017693 on Build GRCh38
rs185833869 Chr.17: 7017583-7017583 on Build GRCh38
rs199582948 Chr.17: 7017603-7017603 on Build GRCh38
rs972884408 Chr.17: 7017733-7017733 on Build GRCh38
rs950156246 Chr.17: 7017764-7017764 on Build GRCh38
rs764461715 Chr.17: 7017688-7017688 on Build GRCh38
rs1045680288 Chr.17: 7017838-7017838 on Build GRCh38
rs773075275 Chr.17: 7017555-7017555 on Build GRCh38
rs532434307 Chr.17: 7017854-7017854 on Build GRCh38
rs756498778 Chr.17: 7017591-7017591 on Build GRCh38
rs764669899 Chr.17: 7017971-7017971 on Build GRCh38
rs547014253 Chr.17: 7017772-7017772 on Build GRCh38
rs545230167 Chr.17: 7017599-7017599 on Build GRCh38
rs1033301416 Chr.17: 7017803-7017803 on Build GRCh38
rs768022471 Chr.17: 7017579-7017579 on Build GRCh38
rs552422036 Chr.17: 7017855-7017855 on Build GRCh38
rs745486258 Chr.17: 7017918-7017918 on Build GRCh38
rs774053148 Chr.17: 7017915-7017915 on Build GRCh38
rs568912389 Chr.17: 7017970-7017970 on Build GRCh38
rs777503530 Chr.17: 7017742-7017742 on Build GRCh38
rs765964712 Chr.17: 7017574-7017574 on Build GRCh38
rs765571240 Chr.17: 7017700-7017700 on Build GRCh38
rs772223218 Chr.17: 7017620-7017620 on Build GRCh38
rs905888361 Chr.17: 7017848-7017848 on Build GRCh38
rs941562138 Chr.17: 7017732-7017732 on Build GRCh38
rs761226479 Chr.17: 7017674-7017674 on Build GRCh38
rs937255984 Chr.17: 7017861-7017861 on Build GRCh38
rs769526119 Chr.17: 7017646-7017646 on Build GRCh38
rs780159486 Chr.17: 7017720-7017720 on Build GRCh38
rs565304343 Chr.17: 7017608-7017608 on Build GRCh38
rs760066547 Chr.17: 7017962-7017962 on Build GRCh38
rs530157493 Chr.17: 7017744-7017744 on Build GRCh38
rs965793254 Chr.17: 7017665-7017665 on Build GRCh38
rs1028648384 Chr.17: 7017954-7017954 on Build GRCh38
rs560472854 Chr.17: 7017803-7017803 on Build GRCh38
rs767045828 Chr.17: 7017669-7017669 on Build GRCh38
rs369945617 Chr.17: 7017929-7017929 on Build GRCh38
rs757266327 Chr.17: 7017568-7017568 on Build GRCh38
rs745948637 Chr.17: 7017606-7017606 on Build GRCh38
rs575717543 Chr.17: 7017656-7017656 on Build GRCh38
rs376663065 Chr.17: 7017713-7017713 on Build GRCh38
rs749882696 Chr.17: 7017712-7017713 on Build GRCh38
rs574312111 Chr.17: 7017565-7017565 on Build GRCh38
rs754060403 Chr.17: 7017597-7017597 on Build GRCh38
rs1001872040 Chr.17: 7017629-7017629 on Build GRCh38

View Full Product Details