Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38852753-38853252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL23
Entrez Gene ID
9349
Gene Name
ribosomal protein L23
Gene Aliases
L23, rpL17
UniGene
Hs.406300
Gene Chromosome Location
Chr.17: 38850068-38853800 on Build GRCh38
Transcripts
NM_000978.3 NR_002576.1

Gene Symbol
SNORA21
Entrez Gene ID
619505
Gene Name
small nucleolar RNA, H/ACA box 21
Gene Aliases
ACA21
UniGene
Hs.738742
Gene Chromosome Location
Chr.17: 38852863-38852995 on Build GRCh38
Transcripts
NM_000978.3 NR_002576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1736218 c.83G>T p.C28F Chr.17: 38853036-38853036 on Build GRCh38
4065925 c.44G>A p.R15Q Chr.17: 38853075-38853075 on Build GRCh38
1224209 c.88G>A p.D30N Chr.17: 38853031-38853031 on Build GRCh38
3370694 c.45G>C p.R15R Chr.17: 38853074-38853074 on Build GRCh38
6220692 c.78C>T p.I26I Chr.17: 38853041-38853041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977734528 Chr.17: 38853154-38853154 on Build GRCh38
rs748407262 Chr.17: 38853176-38853177 on Build GRCh38
rs763366637 Chr.17: 38852845-38852848 on Build GRCh38
rs775995728 Chr.17: 38852873-38852873 on Build GRCh38
rs371656936 Chr.17: 38852991-38852991 on Build GRCh38
rs774851297 Chr.17: 38852993-38852993 on Build GRCh38
rs199890436 Chr.17: 38852941-38852941 on Build GRCh38
rs779749441 Chr.17: 38853149-38853149 on Build GRCh38
rs745351161 Chr.17: 38852953-38852953 on Build GRCh38
rs780146070 Chr.17: 38852780-38852780 on Build GRCh38
rs371838445 Chr.17: 38853012-38853012 on Build GRCh38
rs756964133 Chr.17: 38853139-38853139 on Build GRCh38
rs771731236 Chr.17: 38852832-38852832 on Build GRCh38
rs772367688 Chr.17: 38853110-38853110 on Build GRCh38
rs769979297 Chr.17: 38853181-38853181 on Build GRCh38
rs749869874 Chr.17: 38853069-38853069 on Build GRCh38
rs538144057 Chr.17: 38853009-38853009 on Build GRCh38
rs921576707 Chr.17: 38853077-38853077 on Build GRCh38
rs571796856 Chr.17: 38852793-38852793 on Build GRCh38
rs977808117 Chr.17: 38852843-38852843 on Build GRCh38
rs767702169 Chr.17: 38852906-38852906 on Build GRCh38
rs763362461 Chr.17: 38852896-38852896 on Build GRCh38
rs777412117 Chr.17: 38852944-38852944 on Build GRCh38
rs769254792 Chr.17: 38853075-38853075 on Build GRCh38
rs932148095 Chr.17: 38853202-38853202 on Build GRCh38
rs745417060 Chr.17: 38853064-38853064 on Build GRCh38
rs773238508 Chr.17: 38853113-38853113 on Build GRCh38
rs761023930 Chr.17: 38853221-38853221 on Build GRCh38
rs746725392 Chr.17: 38852947-38852947 on Build GRCh38
rs751328610 Chr.17: 38853039-38853039 on Build GRCh38
rs752517187 Chr.17: 38852933-38852933 on Build GRCh38
rs764206144 Chr.17: 38853222-38853222 on Build GRCh38
rs769121390 Chr.17: 38853068-38853068 on Build GRCh38
rs940977796 Chr.17: 38853215-38853216 on Build GRCh38
rs1021913440 Chr.17: 38852996-38852996 on Build GRCh38
rs991184556 Chr.17: 38852803-38852803 on Build GRCh38
rs557837476 Chr.17: 38852856-38852856 on Build GRCh38
rs940519225 Chr.17: 38853182-38853182 on Build GRCh38
rs755125164 Chr.17: 38852888-38852890 on Build GRCh38
rs760818652 Chr.17: 38853115-38853115 on Build GRCh38
rs368570213 Chr.17: 38853011-38853011 on Build GRCh38
rs757539507 Chr.17: 38852901-38852901 on Build GRCh38
rs746522864 Chr.17: 38852849-38852849 on Build GRCh38
rs965777034 Chr.17: 38853179-38853179 on Build GRCh38
rs977656573 Chr.17: 38853016-38853016 on Build GRCh38
rs549482642 Chr.17: 38852889-38852889 on Build GRCh38
rs764444281 Chr.17: 38852897-38852897 on Build GRCh38
rs907324778 Chr.17: 38852946-38852946 on Build GRCh38
rs770570176 Chr.17: 38852949-38852949 on Build GRCh38
rs763999661 Chr.17: 38853120-38853120 on Build GRCh38
rs34160491 Chr.17: 38853168-38853169 on Build GRCh38
rs922126687 Chr.17: 38852882-38852882 on Build GRCh38
rs531947694 Chr.17: 38853014-38853014 on Build GRCh38
rs921609918 Chr.17: 38852864-38852864 on Build GRCh38
rs754867417 Chr.17: 38852822-38852822 on Build GRCh38
rs780663707 Chr.17: 38853061-38853061 on Build GRCh38
rs767489718 Chr.17: 38852896-38852897 on Build GRCh38
rs187374217 Chr.17: 38853204-38853204 on Build GRCh38
rs1048987185 Chr.17: 38852899-38852899 on Build GRCh38
rs143537161 Chr.17: 38853057-38853057 on Build GRCh38
rs767012409 Chr.17: 38852870-38852870 on Build GRCh38
rs887595495 Chr.17: 38852918-38852918 on Build GRCh38
rs1239179 Chr.17: 38852819-38852819 on Build GRCh38
rs112538197 Chr.17: 38853020-38853020 on Build GRCh38
rs928466778 Chr.17: 38853207-38853207 on Build GRCh38
rs545426864 Chr.17: 38852779-38852779 on Build GRCh38
rs763414359 Chr.17: 38852976-38852976 on Build GRCh38
rs767633241 Chr.17: 38853198-38853198 on Build GRCh38
rs770356733 Chr.17: 38852961-38852961 on Build GRCh38
rs546393811 Chr.17: 38853140-38853140 on Build GRCh38
rs774870348 Chr.17: 38852978-38852978 on Build GRCh38
rs976275089 Chr.17: 38853188-38853188 on Build GRCh38
rs759987104 Chr.17: 38853117-38853117 on Build GRCh38
rs376234763 Chr.17: 38853119-38853119 on Build GRCh38
rs751381482 Chr.17: 38853123-38853123 on Build GRCh38
rs777615381 Chr.17: 38853029-38853029 on Build GRCh38
rs780797285 Chr.17: 38852952-38852952 on Build GRCh38
rs909167827 Chr.17: 38853148-38853148 on Build GRCh38
rs1034318357 Chr.17: 38852972-38852972 on Build GRCh38
rs747872009 Chr.17: 38852830-38852830 on Build GRCh38
rs112844799 Chr.17: 38852880-38852880 on Build GRCh38
rs752569951 Chr.17: 38853023-38853023 on Build GRCh38
rs755779594 Chr.17: 38853146-38853146 on Build GRCh38
rs750437159 Chr.17: 38852907-38852907 on Build GRCh38
rs115701245 Chr.17: 38853208-38853208 on Build GRCh38
rs755564906 Chr.17: 38853156-38853156 on Build GRCh38
rs778806893 Chr.17: 38852823-38852823 on Build GRCh38
rs751988514 Chr.17: 38852871-38852873 on Build GRCh38
rs753019421 Chr.17: 38853144-38853145 on Build GRCh38
rs377383657 Chr.17: 38852878-38852878 on Build GRCh38
rs879470503 Chr.17: 38853179-38853180 on Build GRCh38
rs201588765 Chr.17: 38852923-38852923 on Build GRCh38
rs748782781 Chr.17: 38853150-38853150 on Build GRCh38
rs985029054 Chr.17: 38853206-38853206 on Build GRCh38
rs772809261 Chr.17: 38852836-38852836 on Build GRCh38
rs367956826 Chr.17: 38853084-38853084 on Build GRCh38
rs182421097 Chr.17: 38852975-38852975 on Build GRCh38
rs767205327 Chr.17: 38853141-38853141 on Build GRCh38
rs923703688 Chr.17: 38852863-38852863 on Build GRCh38
rs770365884 Chr.17: 38852852-38852852 on Build GRCh38
rs767610446 Chr.17: 38853000-38853000 on Build GRCh38
rs754990364 Chr.17: 38852922-38852922 on Build GRCh38
rs374599622 Chr.17: 38853157-38853157 on Build GRCh38

View Full Product Details