Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58331110-58331604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR142
Entrez Gene ID
406934
Gene Name
microRNA 142
Gene Aliases
MIRN142, mir-142
UniGene
Gene Chromosome Location
Chr.17: 58331232-58331318 on Build GRCh38
Transcripts
NR_029683.1 NR_038410.1 NR_038411.1 NR_038412.1 NR_038413.1

Gene Symbol
TSPOAP1-AS1
Entrez Gene ID
100506779
Gene Name
TSPOAP1 antisense RNA 1
Gene Aliases
BZRAP1-AS1
UniGene
Hs.718477
Gene Chromosome Location
Chr.17: 58325450-58353727 on Build GRCh38
Transcripts
NR_029683.1 NR_038410.1 NR_038411.1 NR_038412.1 NR_038413.1

SNP Detail

SNP ID Chromosome Location
rs1012276158 Chr.17: 58331580-58331580 on Build GRCh38
rs565279408 Chr.17: 58331540-58331540 on Build GRCh38
rs941524335 Chr.17: 58331203-58331203 on Build GRCh38
rs936929683 Chr.17: 58331534-58331534 on Build GRCh38
rs533848920 Chr.17: 58331330-58331330 on Build GRCh38
rs956570669 Chr.17: 58331383-58331383 on Build GRCh38
rs764363741 Chr.17: 58331206-58331206 on Build GRCh38
rs912341854 Chr.17: 58331476-58331476 on Build GRCh38
rs578020025 Chr.17: 58331216-58331216 on Build GRCh38
rs1013266906 Chr.17: 58331393-58331393 on Build GRCh38
rs536795315 Chr.17: 58331444-58331444 on Build GRCh38
rs969577053 Chr.17: 58331177-58331177 on Build GRCh38
rs867010562 Chr.17: 58331300-58331300 on Build GRCh38
rs751449960 Chr.17: 58331336-58331336 on Build GRCh38
rs111619549 Chr.17: 58331546-58331546 on Build GRCh38
rs764879289 Chr.17: 58331217-58331217 on Build GRCh38
rs540592228 Chr.17: 58331178-58331178 on Build GRCh38
rs760054303 Chr.17: 58331179-58331179 on Build GRCh38
rs377637047 Chr.17: 58331248-58331248 on Build GRCh38
rs759796305 Chr.17: 58331205-58331205 on Build GRCh38
rs550842646 Chr.17: 58331238-58331238 on Build GRCh38
rs918844908 Chr.17: 58331554-58331554 on Build GRCh38
rs762351214 Chr.17: 58331333-58331333 on Build GRCh38
rs540599972 Chr.17: 58331537-58331537 on Build GRCh38
rs778163817 Chr.17: 58331276-58331276 on Build GRCh38
rs756004203 Chr.17: 58331364-58331364 on Build GRCh38
rs767576832 Chr.17: 58331355-58331355 on Build GRCh38
rs999457643 Chr.17: 58331417-58331417 on Build GRCh38
rs980957680 Chr.17: 58331462-58331462 on Build GRCh38
rs902799738 Chr.17: 58331176-58331176 on Build GRCh38
rs781201763 Chr.17: 58331255-58331255 on Build GRCh38
rs774962601 Chr.17: 58331204-58331204 on Build GRCh38
rs371054667 Chr.17: 58331326-58331326 on Build GRCh38
rs918813605 Chr.17: 58331214-58331214 on Build GRCh38
rs772570355 Chr.17: 58331308-58331308 on Build GRCh38
rs991801333 Chr.17: 58331158-58331158 on Build GRCh38
rs956373593 Chr.17: 58331156-58331156 on Build GRCh38
rs773361226 Chr.17: 58331190-58331190 on Build GRCh38
rs574594031 Chr.17: 58331435-58331435 on Build GRCh38
rs1015502031 Chr.17: 58331369-58331369 on Build GRCh38
rs963220101 Chr.17: 58331479-58331479 on Build GRCh38
rs1030910083 Chr.17: 58331140-58331140 on Build GRCh38
rs767926119 Chr.17: 58331207-58331207 on Build GRCh38
rs973670230 Chr.17: 58331380-58331380 on Build GRCh38
rs1025100029 Chr.17: 58331446-58331446 on Build GRCh38
rs763408747 Chr.17: 58331581-58331581 on Build GRCh38
rs749401775 Chr.17: 58331194-58331194 on Build GRCh38
rs537940062 Chr.17: 58331361-58331361 on Build GRCh38
rs750753199 Chr.17: 58331485-58331485 on Build GRCh38
rs761047970 Chr.17: 58331327-58331327 on Build GRCh38
rs747963068 Chr.17: 58331259-58331259 on Build GRCh38
rs908932757 Chr.17: 58331478-58331478 on Build GRCh38
rs570378710 Chr.17: 58331337-58331337 on Build GRCh38
rs1042990569 Chr.17: 58331508-58331508 on Build GRCh38
rs374051002 Chr.17: 58331189-58331189 on Build GRCh38
rs528796268 Chr.17: 58331172-58331172 on Build GRCh38
rs573562920 Chr.17: 58331530-58331530 on Build GRCh38
rs746452985 Chr.17: 58331288-58331288 on Build GRCh38
rs749883234 Chr.17: 58331219-58331219 on Build GRCh38
rs147467596 Chr.17: 58331167-58331167 on Build GRCh38
rs868344410 Chr.17: 58331154-58331154 on Build GRCh38
rs771294176 Chr.17: 58331200-58331200 on Build GRCh38
rs746909875 Chr.17: 58331257-58331265 on Build GRCh38
rs953773341 Chr.17: 58331451-58331451 on Build GRCh38
rs781307080 Chr.17: 58331313-58331314 on Build GRCh38
rs556025330 Chr.17: 58331419-58331419 on Build GRCh38
rs776662612 Chr.17: 58331433-58331433 on Build GRCh38
rs932405761 Chr.17: 58331141-58331141 on Build GRCh38
rs919607046 Chr.17: 58331411-58331411 on Build GRCh38
rs556317406 Chr.17: 58331163-58331163 on Build GRCh38
rs924179998 Chr.17: 58331512-58331512 on Build GRCh38
rs776259559 Chr.17: 58331210-58331210 on Build GRCh38
rs562696473 Chr.17: 58331264-58331264 on Build GRCh38
rs930260397 Chr.17: 58331558-58331558 on Build GRCh38
rs776347554 Chr.17: 58331222-58331222 on Build GRCh38
rs756348475 Chr.17: 58331260-58331260 on Build GRCh38
rs372865698 Chr.17: 58331328-58331328 on Build GRCh38
rs555222379 Chr.17: 58331461-58331461 on Build GRCh38
rs761546420 Chr.17: 58331331-58331331 on Build GRCh38
rs1042632999 Chr.17: 58331171-58331171 on Build GRCh38
rs577208311 Chr.17: 58331576-58331576 on Build GRCh38
rs547987105 Chr.17: 58331278-58331278 on Build GRCh38
rs766200458 Chr.17: 58331225-58331225 on Build GRCh38
rs529802001 Chr.17: 58331277-58331277 on Build GRCh38
rs372956615 Chr.17: 58331135-58331135 on Build GRCh38
rs201627422 Chr.17: 58331334-58331334 on Build GRCh38
rs961630457 Chr.17: 58331380-58331380 on Build GRCh38
rs944214131 Chr.17: 58331496-58331496 on Build GRCh38
rs755018180 Chr.17: 58331314-58331317 on Build GRCh38
rs891121951 Chr.17: 58331249-58331249 on Build GRCh38
rs751567736 Chr.17: 58331227-58331227 on Build GRCh38

View Full Product Details