Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 30298750-30299272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR302F
Entrez Gene ID
100302131
Gene Name
microRNA 302f
Gene Aliases
MIRN302F, hsa-mir-302f
UniGene
Gene Chromosome Location
Chr.18: 30298910-30298960 on Build GRCh38
Transcripts
NR_031684.1

SNP Detail

SNP ID Chromosome Location
rs779679276 Chr.18: 30298877-30298877 on Build GRCh38
rs1042685173 Chr.18: 30299201-30299201 on Build GRCh38
rs921128718 Chr.18: 30299144-30299144 on Build GRCh38
rs371050780 Chr.18: 30299061-30299061 on Build GRCh38
rs371547375 Chr.18: 30298898-30298898 on Build GRCh38
rs927374505 Chr.18: 30298837-30298837 on Build GRCh38
rs756335403 Chr.18: 30298900-30298900 on Build GRCh38
rs868685692 Chr.18: 30298932-30298932 on Build GRCh38
rs1001713638 Chr.18: 30298985-30298985 on Build GRCh38
rs753655781 Chr.18: 30299010-30299010 on Build GRCh38
rs929269686 Chr.18: 30298795-30298795 on Build GRCh38
rs778021887 Chr.18: 30298942-30298942 on Build GRCh38
rs1034477144 Chr.18: 30299084-30299084 on Build GRCh38
rs532807928 Chr.18: 30299091-30299091 on Build GRCh38
rs936513890 Chr.18: 30299159-30299159 on Build GRCh38
rs943486010 Chr.18: 30298843-30298843 on Build GRCh38
rs952615141 Chr.18: 30299050-30299050 on Build GRCh38
rs981593972 Chr.18: 30298836-30298836 on Build GRCh38
rs543657312 Chr.18: 30298946-30298946 on Build GRCh38
rs1011160309 Chr.18: 30298962-30298962 on Build GRCh38
rs947677852 Chr.18: 30299167-30299167 on Build GRCh38
rs1021277857 Chr.18: 30298964-30298964 on Build GRCh38
rs755997188 Chr.18: 30298874-30298874 on Build GRCh38
rs770158789 Chr.18: 30298958-30298958 on Build GRCh38
rs943904582 Chr.18: 30298948-30298948 on Build GRCh38
rs747724110 Chr.18: 30298896-30298896 on Build GRCh38
rs529495765 Chr.18: 30298998-30298998 on Build GRCh38
rs959018060 Chr.18: 30298864-30298864 on Build GRCh38
rs186915888 Chr.18: 30298986-30298986 on Build GRCh38
rs982735884 Chr.18: 30299127-30299127 on Build GRCh38
rs892163207 Chr.18: 30299031-30299031 on Build GRCh38
rs754640160 Chr.18: 30298886-30298886 on Build GRCh38
rs1049094570 Chr.18: 30298817-30298817 on Build GRCh38
rs368801782 Chr.18: 30298880-30298880 on Build GRCh38
rs779081218 Chr.18: 30299085-30299085 on Build GRCh38
rs1020543891 Chr.18: 30298825-30298825 on Build GRCh38
rs767346727 Chr.18: 30298971-30298971 on Build GRCh38
rs759770332 Chr.18: 30298955-30298955 on Build GRCh38
rs112507660 Chr.18: 30299015-30299015 on Build GRCh38
rs1039203025 Chr.18: 30298849-30298849 on Build GRCh38
rs971577779 Chr.18: 30298827-30298827 on Build GRCh38
rs749435098 Chr.18: 30298947-30298947 on Build GRCh38
rs78549380 Chr.18: 30299101-30299101 on Build GRCh38
rs370419387 Chr.18: 30299014-30299014 on Build GRCh38
rs775405313 Chr.18: 30298960-30298960 on Build GRCh38
rs1053530649 Chr.18: 30299021-30299021 on Build GRCh38
rs750165615 Chr.18: 30298862-30298862 on Build GRCh38
rs943935709 Chr.18: 30299142-30299142 on Build GRCh38
rs1039485660 Chr.18: 30298968-30298968 on Build GRCh38
rs1053222916 Chr.18: 30299146-30299149 on Build GRCh38
rs951484496 Chr.18: 30299111-30299111 on Build GRCh38
rs997767297 Chr.18: 30298869-30298869 on Build GRCh38
rs956277081 Chr.18: 30299129-30299129 on Build GRCh38
rs752520839 Chr.18: 30298811-30298811 on Build GRCh38
rs1010937951 Chr.18: 30298819-30298819 on Build GRCh38
rs564439110 Chr.18: 30298839-30298839 on Build GRCh38
rs767046330 Chr.18: 30299057-30299057 on Build GRCh38
rs777136689 Chr.18: 30298952-30298952 on Build GRCh38
rs1003060496 Chr.18: 30299070-30299070 on Build GRCh38
rs918469168 Chr.18: 30299027-30299027 on Build GRCh38
rs1009216438 Chr.18: 30299049-30299049 on Build GRCh38
rs912495661 Chr.18: 30299139-30299139 on Build GRCh38
rs576440207 Chr.18: 30298861-30298861 on Build GRCh38
rs569655340 Chr.18: 30299143-30299143 on Build GRCh38
rs750444762 Chr.18: 30298981-30298981 on Build GRCh38
rs778346102 Chr.18: 30298891-30298891 on Build GRCh38
rs760592676 Chr.18: 30298979-30299028 on Build GRCh38
rs763145470 Chr.18: 30298966-30298966 on Build GRCh38
rs905553604 Chr.18: 30299212-30299212 on Build GRCh38
rs747320577 Chr.18: 30298908-30298908 on Build GRCh38

View Full Product Details