Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 32937108-32937653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00486
Entrez Gene ID
285045
Gene Name
long intergenic non-protein coding RNA 486
Gene Aliases
-
UniGene
Hs.434660
Gene Chromosome Location
Chr.2: 32825443-32937203 on Build GRCh38
Transcripts
NR_027099.1 NR_027097.1

Gene Symbol
LOC100271832
Entrez Gene ID
100271832
Gene Name
uncharacterized LOC100271832
Gene Aliases
-
UniGene
Hs.743603
Gene Chromosome Location
Chr.2: 32927127-32946136 on Build GRCh38
Transcripts
NR_027099.1 NR_027097.1

SNP Detail

SNP ID Chromosome Location
rs940874295 Chr.2: 32937141-32937141 on Build GRCh38
rs903241045 Chr.2: 32937246-32937246 on Build GRCh38
rs992135786 Chr.2: 32937422-32937422 on Build GRCh38
rs928786654 Chr.2: 32937381-32937381 on Build GRCh38
rs926201040 Chr.2: 32937162-32937162 on Build GRCh38
rs756439281 Chr.2: 32937344-32937344 on Build GRCh38
rs146962182 Chr.2: 32937271-32937271 on Build GRCh38
rs1041718789 Chr.2: 32937451-32937451 on Build GRCh38
rs151202939 Chr.2: 32937330-32937330 on Build GRCh38
rs1040265730 Chr.2: 32937475-32937475 on Build GRCh38
rs868381662 Chr.2: 32937605-32937605 on Build GRCh38
rs956455379 Chr.2: 32937214-32937216 on Build GRCh38
rs140292984 Chr.2: 32937582-32937582 on Build GRCh38
rs1045852519 Chr.2: 32937176-32937176 on Build GRCh38
rs981151139 Chr.2: 32937625-32937625 on Build GRCh38
rs903272014 Chr.2: 32937469-32937469 on Build GRCh38
rs937554103 Chr.2: 32937179-32937179 on Build GRCh38
rs1005844604 Chr.2: 32937562-32937562 on Build GRCh38
rs965998263 Chr.2: 32937298-32937298 on Build GRCh38
rs750073862 Chr.2: 32937207-32937207 on Build GRCh38
rs928892060 Chr.2: 32937177-32937177 on Build GRCh38
rs187445331 Chr.2: 32937377-32937377 on Build GRCh38
rs536775197 Chr.2: 32937342-32937342 on Build GRCh38
rs748856792 Chr.2: 32937397-32937397 on Build GRCh38
rs35966458 Chr.2: 32937587-32937588 on Build GRCh38
rs1017977979 Chr.2: 32937578-32937578 on Build GRCh38
rs192737818 Chr.2: 32937544-32937544 on Build GRCh38
rs529718639 Chr.2: 32937606-32937606 on Build GRCh38
rs755805648 Chr.2: 32937258-32937258 on Build GRCh38
rs377153962 Chr.2: 32937345-32937345 on Build GRCh38
rs996489186 Chr.2: 32937314-32937314 on Build GRCh38
rs914610563 Chr.2: 32937423-32937423 on Build GRCh38
rs571505355 Chr.2: 32937184-32937184 on Build GRCh38
rs555884253 Chr.2: 32937515-32937515 on Build GRCh38
rs534221340 Chr.2: 32937190-32937190 on Build GRCh38
rs758795206 Chr.2: 32937414-32937414 on Build GRCh38
rs996137005 Chr.2: 32937530-32937530 on Build GRCh38
rs901671252 Chr.2: 32937501-32937501 on Build GRCh38
rs1029338513 Chr.2: 32937543-32937543 on Build GRCh38
rs961731090 Chr.2: 32937359-32937359 on Build GRCh38
rs947384245 Chr.2: 32937427-32937427 on Build GRCh38
rs1011688383 Chr.2: 32937222-32937222 on Build GRCh38
rs778283907 Chr.2: 32937571-32937571 on Build GRCh38
rs545334135 Chr.2: 32937483-32937483 on Build GRCh38
rs927400460 Chr.2: 32937357-32937357 on Build GRCh38
rs893122301 Chr.2: 32937194-32937194 on Build GRCh38
rs1041813754 Chr.2: 32937235-32937235 on Build GRCh38
rs183864348 Chr.2: 32937592-32937592 on Build GRCh38
rs553941769 Chr.2: 32937282-32937282 on Build GRCh38
rs117875913 Chr.2: 32937474-32937474 on Build GRCh38
rs560583432 Chr.2: 32937595-32937595 on Build GRCh38

View Full Product Details