Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86023128-86023535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC90784
Entrez Gene ID
90784
Gene Name
uncharacterized LOC90784
Gene Aliases
-
UniGene
Hs.594051
Gene Chromosome Location
Chr.2: 86020216-86023868 on Build GRCh38
Transcripts
NR_026984.1

SNP Detail

SNP ID Chromosome Location
rs996376878 Chr.2: 86023271-86023271 on Build GRCh38
rs769778175 Chr.2: 86023328-86023328 on Build GRCh38
rs1008841998 Chr.2: 86023465-86023465 on Build GRCh38
rs920311722 Chr.2: 86023304-86023304 on Build GRCh38
rs145921018 Chr.2: 86023236-86023236 on Build GRCh38
rs762559037 Chr.2: 86023329-86023329 on Build GRCh38
rs1028914386 Chr.2: 86023272-86023272 on Build GRCh38
rs1012581397 Chr.2: 86023405-86023405 on Build GRCh38
rs902509203 Chr.2: 86023222-86023222 on Build GRCh38
rs999636982 Chr.2: 86023223-86023223 on Build GRCh38
rs931450223 Chr.2: 86023171-86023171 on Build GRCh38
rs1028867312 Chr.2: 86023435-86023435 on Build GRCh38
rs565882961 Chr.2: 86023288-86023288 on Build GRCh38
rs766224681 Chr.2: 86023343-86023343 on Build GRCh38
rs144912102 Chr.2: 86023214-86023214 on Build GRCh38
rs931500443 Chr.2: 86023313-86023313 on Build GRCh38
rs961687868 Chr.2: 86023291-86023291 on Build GRCh38
rs1053882449 Chr.2: 86023359-86023359 on Build GRCh38
rs949666617 Chr.2: 86023333-86023333 on Build GRCh38
rs916739840 Chr.2: 86023332-86023332 on Build GRCh38
rs138406344 Chr.2: 86023443-86023443 on Build GRCh38
rs547748870 Chr.2: 86023250-86023250 on Build GRCh38
rs149001863 Chr.2: 86023233-86023233 on Build GRCh38
rs935298698 Chr.2: 86023358-86023358 on Build GRCh38
rs1055395329 Chr.2: 86023178-86023178 on Build GRCh38
rs772897410 Chr.2: 86023284-86023284 on Build GRCh38
rs994878208 Chr.2: 86023507-86023507 on Build GRCh38
rs142914316 Chr.2: 86023476-86023476 on Build GRCh38
rs893845463 Chr.2: 86023372-86023372 on Build GRCh38
rs745739024 Chr.2: 86023289-86023289 on Build GRCh38
rs902538815 Chr.2: 86023351-86023351 on Build GRCh38
rs894945618 Chr.2: 86023388-86023388 on Build GRCh38
rs973195983 Chr.2: 86023300-86023300 on Build GRCh38
rs895361895 Chr.2: 86023201-86023201 on Build GRCh38
rs776008103 Chr.2: 86023213-86023213 on Build GRCh38
rs943612158 Chr.2: 86023256-86023256 on Build GRCh38
rs76873798 Chr.2: 86023477-86023477 on Build GRCh38
rs188038319 Chr.2: 86023416-86023416 on Build GRCh38
rs920272447 Chr.2: 86023285-86023285 on Build GRCh38
rs996274748 Chr.2: 86023381-86023381 on Build GRCh38
rs968709337 Chr.2: 86023193-86023193 on Build GRCh38

View Full Product Details