Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112643987-112644493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ42351
Entrez Gene ID
400999
Gene Name
uncharacterized LOC400999
Gene Aliases
-
UniGene
Hs.585221
Gene Chromosome Location
Chr.2: 112641830-112644180 on Build GRCh38
Transcripts
NR_033871.1

SNP Detail

SNP ID Chromosome Location
rs985806754 Chr.2: 112644025-112644025 on Build GRCh38
rs544065149 Chr.2: 112644072-112644072 on Build GRCh38
rs748945167 Chr.2: 112644229-112644229 on Build GRCh38
rs189621350 Chr.2: 112644113-112644113 on Build GRCh38
rs1045683489 Chr.2: 112644023-112644023 on Build GRCh38
rs6737094 Chr.2: 112644140-112644140 on Build GRCh38
rs368141636 Chr.2: 112644323-112644323 on Build GRCh38
rs954227609 Chr.2: 112644238-112644238 on Build GRCh38
rs972579894 Chr.2: 112644327-112644327 on Build GRCh38
rs36035433 Chr.2: 112644167-112644168 on Build GRCh38
rs536783786 Chr.2: 112644411-112644411 on Build GRCh38
rs985388990 Chr.2: 112644248-112644248 on Build GRCh38
rs531993153 Chr.2: 112644363-112644363 on Build GRCh38
rs905555003 Chr.2: 112644106-112644106 on Build GRCh38
rs765611689 Chr.2: 112644344-112644344 on Build GRCh38
rs957178001 Chr.2: 112644179-112644179 on Build GRCh38
rs755679699 Chr.2: 112644252-112644268 on Build GRCh38
rs1045530214 Chr.2: 112644391-112644391 on Build GRCh38
rs1029601966 Chr.2: 112644233-112644233 on Build GRCh38
rs180869769 Chr.2: 112644442-112644442 on Build GRCh38
rs962537992 Chr.2: 112644291-112644291 on Build GRCh38
rs922497559 Chr.2: 112644022-112644022 on Build GRCh38
rs781035863 Chr.2: 112644104-112644104 on Build GRCh38
rs887823726 Chr.2: 112644292-112644292 on Build GRCh38
rs977109886 Chr.2: 112644170-112644170 on Build GRCh38
rs115093506 Chr.2: 112644196-112644196 on Build GRCh38
rs571479825 Chr.2: 112644387-112644387 on Build GRCh38
rs1036749261 Chr.2: 112644068-112644068 on Build GRCh38
rs928577591 Chr.2: 112644282-112644282 on Build GRCh38
rs929303304 Chr.2: 112644283-112644283 on Build GRCh38
rs941226953 Chr.2: 112644041-112644041 on Build GRCh38
rs773873689 Chr.2: 112644366-112644366 on Build GRCh38
rs140961271 Chr.2: 112644232-112644232 on Build GRCh38
rs35226648 Chr.2: 112644018-112644018 on Build GRCh38
rs767021388 Chr.2: 112644469-112644469 on Build GRCh38
rs866989629 Chr.2: 112644419-112644419 on Build GRCh38
rs769538262 Chr.2: 112644156-112644156 on Build GRCh38
rs894051032 Chr.2: 112644443-112644443 on Build GRCh38
rs114443043 Chr.2: 112644143-112644143 on Build GRCh38
rs377136597 Chr.2: 112644465-112644465 on Build GRCh38
rs757296292 Chr.2: 112644073-112644073 on Build GRCh38
rs768240924 Chr.2: 112644329-112644329 on Build GRCh38
rs567447504 Chr.2: 112644463-112644463 on Build GRCh38
rs150190638 Chr.2: 112644378-112644378 on Build GRCh38
rs1017777430 Chr.2: 112644137-112644137 on Build GRCh38
rs527894571 Chr.2: 112644434-112644434 on Build GRCh38
rs953977255 Chr.2: 112644024-112644024 on Build GRCh38
rs909798956 Chr.2: 112644278-112644278 on Build GRCh38
rs1031837725 Chr.2: 112644176-112644176 on Build GRCh38
rs796675732 Chr.2: 112644322-112644322 on Build GRCh38
rs12469675 Chr.2: 112644157-112644157 on Build GRCh38
rs909716064 Chr.2: 112644037-112644037 on Build GRCh38
rs151050382 Chr.2: 112644084-112644084 on Build GRCh38
rs758804261 Chr.2: 112644250-112644253 on Build GRCh38

View Full Product Details