Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165857386-165857898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100506124
Entrez Gene ID
100506124
Gene Name
uncharacterized LOC100506124
Gene Aliases
-
UniGene
Hs.709870
Gene Chromosome Location
Chr.2: 165857475-165871941 on Build GRCh38
Transcripts
NR_045375.2

SNP Detail

SNP ID Chromosome Location
rs752224342 Chr.2: 165857602-165857602 on Build GRCh38
rs901928464 Chr.2: 165857461-165857461 on Build GRCh38
rs910280632 Chr.2: 165857749-165857750 on Build GRCh38
rs528600882 Chr.2: 165857870-165857870 on Build GRCh38
rs934148808 Chr.2: 165857806-165857806 on Build GRCh38
rs540676334 Chr.2: 165857689-165857689 on Build GRCh38
rs1010565354 Chr.2: 165857821-165857821 on Build GRCh38
rs945210267 Chr.2: 165857447-165857447 on Build GRCh38
rs769919832 Chr.2: 165857547-165857547 on Build GRCh38
rs918954577 Chr.2: 165857556-165857556 on Build GRCh38
rs766211452 Chr.2: 165857543-165857543 on Build GRCh38
rs933373274 Chr.2: 165857464-165857464 on Build GRCh38
rs898012829 Chr.2: 165857537-165857537 on Build GRCh38
rs545008113 Chr.2: 165857607-165857607 on Build GRCh38
rs571815262 Chr.2: 165857648-165857648 on Build GRCh38
rs889094791 Chr.2: 165857497-165857497 on Build GRCh38
rs774676217 Chr.2: 165857678-165857678 on Build GRCh38
rs748210608 Chr.2: 165857421-165857421 on Build GRCh38
rs2304002 Chr.2: 165857585-165857585 on Build GRCh38
rs760579098 Chr.2: 165857526-165857526 on Build GRCh38
rs979293975 Chr.2: 165857433-165857433 on Build GRCh38
rs913753008 Chr.2: 165857442-165857442 on Build GRCh38
rs1046616464 Chr.2: 165857727-165857727 on Build GRCh38
rs958496062 Chr.2: 165857480-165857480 on Build GRCh38
rs906670535 Chr.2: 165857743-165857743 on Build GRCh38
rs943123310 Chr.2: 165857772-165857772 on Build GRCh38
rs1016504677 Chr.2: 165857530-165857530 on Build GRCh38
rs966655311 Chr.2: 165857873-165857873 on Build GRCh38
rs188364527 Chr.2: 165857714-165857714 on Build GRCh38
rs1050388890 Chr.2: 165857487-165857487 on Build GRCh38
rs1020281062 Chr.2: 165857655-165857655 on Build GRCh38
rs369930236 Chr.2: 165857459-165857459 on Build GRCh38
rs889146622 Chr.2: 165857694-165857694 on Build GRCh38
rs117614081 Chr.2: 165857601-165857601 on Build GRCh38
rs377546769 Chr.2: 165857812-165857812 on Build GRCh38
rs976621643 Chr.2: 165857672-165857672 on Build GRCh38
rs1039914673 Chr.2: 165857593-165857593 on Build GRCh38
rs1006899882 Chr.2: 165857520-165857520 on Build GRCh38
rs1021007074 Chr.2: 165857846-165857846 on Build GRCh38
rs1033410054 Chr.2: 165857799-165857799 on Build GRCh38
rs957981565 Chr.2: 165857803-165857803 on Build GRCh38
rs543642897 Chr.2: 165857713-165857713 on Build GRCh38
rs76865594 Chr.2: 165857720-165857720 on Build GRCh38
rs1024123864 Chr.2: 165857703-165857703 on Build GRCh38
rs991293061 Chr.2: 165857483-165857483 on Build GRCh38
rs2304003 Chr.2: 165857571-165857571 on Build GRCh38
rs183813363 Chr.2: 165857625-165857625 on Build GRCh38
rs1042326505 Chr.2: 165857837-165857837 on Build GRCh38
rs1025111301 Chr.2: 165857567-165857567 on Build GRCh38
rs922400935 Chr.2: 165857674-165857674 on Build GRCh38
rs542659495 Chr.2: 165857697-165857697 on Build GRCh38
rs532881517 Chr.2: 165857708-165857708 on Build GRCh38
rs1042234007 Chr.2: 165857448-165857448 on Build GRCh38
rs926060746 Chr.2: 165857455-165857455 on Build GRCh38
rs552557512 Chr.2: 165857740-165857740 on Build GRCh38
rs545586565 Chr.2: 165857564-165857564 on Build GRCh38
rs61742640 Chr.2: 165857606-165857606 on Build GRCh38
rs35295094 Chr.2: 165857436-165857437 on Build GRCh38
rs865817232 Chr.2: 165857831-165857831 on Build GRCh38
rs151215258 Chr.2: 165857773-165857773 on Build GRCh38

View Full Product Details