Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176173772-176174277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAGLR
Entrez Gene ID
401022
Gene Name
HOXD antisense growth-associated long non-coding RNA
Gene Aliases
HOXD-AS1, Mdgt
UniGene
Hs.98661
Gene Chromosome Location
Chr.2: 176173189-176188958 on Build GRCh38
Transcripts
NR_033979.2 NR_110458.1 NR_110459.1 NR_110460.1 NR_110461.1 NR_110462.1 NR_110463.1 NR_110464.1 NR_110465.1 NR_110466.1 NR_110467.1 NR_110468.1

SNP Detail

SNP ID Chromosome Location
rs570379353 Chr.2: 176174099-176174099 on Build GRCh38
rs956664895 Chr.2: 176173933-176173933 on Build GRCh38
rs527569320 Chr.2: 176173952-176173952 on Build GRCh38
rs1037760503 Chr.2: 176174248-176174248 on Build GRCh38
rs989222276 Chr.2: 176173936-176173936 on Build GRCh38
rs140118522 Chr.2: 176173802-176173802 on Build GRCh38
rs924585212 Chr.2: 176174217-176174217 on Build GRCh38
rs890828770 Chr.2: 176173889-176173889 on Build GRCh38
rs934589774 Chr.2: 176174243-176174243 on Build GRCh38
rs1003157512 Chr.2: 176173896-176173896 on Build GRCh38
rs753527178 Chr.2: 176173800-176173800 on Build GRCh38
rs149736491 Chr.2: 176173877-176173878 on Build GRCh38
rs1012129800 Chr.2: 176173819-176173819 on Build GRCh38
rs1007900004 Chr.2: 176173891-176173891 on Build GRCh38
rs993693179 Chr.2: 176173995-176173995 on Build GRCh38
rs537552887 Chr.2: 176174210-176174210 on Build GRCh38
rs981454692 Chr.2: 176174198-176174198 on Build GRCh38
rs1023491980 Chr.2: 176173860-176173860 on Build GRCh38
rs761027471 Chr.2: 176174098-176174098 on Build GRCh38
rs952237607 Chr.2: 176174044-176174044 on Build GRCh38
rs201841613 Chr.2: 176173889-176173889 on Build GRCh38
rs764747905 Chr.2: 176173971-176173971 on Build GRCh38
rs1030633193 Chr.2: 176173923-176173923 on Build GRCh38
rs1052205900 Chr.2: 176174176-176174176 on Build GRCh38
rs560472703 Chr.2: 176173893-176173893 on Build GRCh38
rs975016582 Chr.2: 176174069-176174069 on Build GRCh38
rs934535706 Chr.2: 176173804-176173804 on Build GRCh38
rs144198623 Chr.2: 176174034-176174037 on Build GRCh38
rs775842987 Chr.2: 176173906-176173906 on Build GRCh38
rs968860701 Chr.2: 176174065-176174065 on Build GRCh38
rs759885625 Chr.2: 176173806-176173806 on Build GRCh38
rs907829109 Chr.2: 176174193-176174193 on Build GRCh38
rs1054311169 Chr.2: 176173818-176173818 on Build GRCh38
rs552219580 Chr.2: 176174020-176174020 on Build GRCh38
rs549887811 Chr.2: 176174211-176174211 on Build GRCh38
rs962286482 Chr.2: 176173932-176173932 on Build GRCh38
rs1027806926 Chr.2: 176174026-176174026 on Build GRCh38
rs1475969 Chr.2: 176174122-176174122 on Build GRCh38

View Full Product Details