Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 177284442-177284946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100130691
Entrez Gene ID
100130691
Gene Name
uncharacterized LOC100130691
Gene Aliases
-
UniGene
Hs.707723
Gene Chromosome Location
Chr.2: 177283508-177392691 on Build GRCh38
Transcripts
NR_026966.1

SNP Detail

SNP ID Chromosome Location
rs186971560 Chr.2: 177284741-177284741 on Build GRCh38
rs1019876814 Chr.2: 177284492-177284492 on Build GRCh38
rs536657261 Chr.2: 177284631-177284631 on Build GRCh38
rs997045899 Chr.2: 177284824-177284824 on Build GRCh38
rs371459073 Chr.2: 177284645-177284645 on Build GRCh38
rs953162610 Chr.2: 177284834-177284834 on Build GRCh38
rs769842688 Chr.2: 177284819-177284819 on Build GRCh38
rs764424840 Chr.2: 177284806-177284806 on Build GRCh38
rs191741899 Chr.2: 177284544-177284544 on Build GRCh38
rs1004199662 Chr.2: 177284755-177284755 on Build GRCh38
rs6433659 Chr.2: 177284636-177284636 on Build GRCh38
rs750531671 Chr.2: 177284815-177284816 on Build GRCh38
rs117633229 Chr.2: 177284832-177284832 on Build GRCh38
rs925030090 Chr.2: 177284847-177284847 on Build GRCh38
rs979067457 Chr.2: 177284583-177284583 on Build GRCh38
rs979183567 Chr.2: 177284836-177284836 on Build GRCh38
rs545726197 Chr.2: 177284512-177284512 on Build GRCh38
rs1052340000 Chr.2: 177284610-177284610 on Build GRCh38
rs990700922 Chr.2: 177284788-177284788 on Build GRCh38
rs1053846925 Chr.2: 177284845-177284845 on Build GRCh38
rs570371253 Chr.2: 177284910-177284910 on Build GRCh38
rs762399603 Chr.2: 177284607-177284607 on Build GRCh38
rs917854563 Chr.2: 177284916-177284916 on Build GRCh38
rs533706967 Chr.2: 177284485-177284485 on Build GRCh38
rs995347382 Chr.2: 177284716-177284716 on Build GRCh38
rs931716816 Chr.2: 177284883-177284883 on Build GRCh38
rs562658785 Chr.2: 177284510-177284510 on Build GRCh38
rs1048501405 Chr.2: 177284727-177284727 on Build GRCh38
rs921718352 Chr.2: 177284537-177284537 on Build GRCh38
rs569099011 Chr.2: 177284793-177284793 on Build GRCh38
rs183000380 Chr.2: 177284695-177284695 on Build GRCh38
rs949293725 Chr.2: 177284529-177284529 on Build GRCh38
rs975976118 Chr.2: 177284502-177284502 on Build GRCh38
rs1046087726 Chr.2: 177284536-177284536 on Build GRCh38
rs959027562 Chr.2: 177284763-177284763 on Build GRCh38
rs962007525 Chr.2: 177284713-177284713 on Build GRCh38
rs148424696 Chr.2: 177284606-177284606 on Build GRCh38
rs953222314 Chr.2: 177284550-177284550 on Build GRCh38
rs956395871 Chr.2: 177284868-177284868 on Build GRCh38
rs1014937021 Chr.2: 177284709-177284709 on Build GRCh38
rs995963539 Chr.2: 177284686-177284686 on Build GRCh38
rs1019909294 Chr.2: 177284795-177284795 on Build GRCh38
rs949276283 Chr.2: 177284634-177284634 on Build GRCh38
rs867987286 Chr.2: 177284684-177284684 on Build GRCh38
rs1051020430 Chr.2: 177284677-177284678 on Build GRCh38
rs75153968 Chr.2: 177284560-177284560 on Build GRCh38
rs546637536 Chr.2: 177284689-177284689 on Build GRCh38
rs376695788 Chr.2: 177284817-177284817 on Build GRCh38
rs952551401 Chr.2: 177284503-177284503 on Build GRCh38
rs905279401 Chr.2: 177284662-177284662 on Build GRCh38
rs890747364 Chr.2: 177284678-177284678 on Build GRCh38
rs965599261 Chr.2: 177284497-177284497 on Build GRCh38
rs549523675 Chr.2: 177284630-177284630 on Build GRCh38
rs915025630 Chr.2: 177284882-177284882 on Build GRCh38
rs556858127 Chr.2: 177284765-177284765 on Build GRCh38
rs987752736 Chr.2: 177284912-177284912 on Build GRCh38

View Full Product Details