Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215615436-215615854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00607
Entrez Gene ID
646324
Gene Name
long intergenic non-protein coding RNA 607
Gene Aliases
-
UniGene
Hs.655713
Gene Chromosome Location
Chr.2: 215611563-215843536 on Build GRCh38
Transcripts
NR_037195.1

SNP Detail

SNP ID Chromosome Location
rs1014192888 Chr.2: 215615737-215615737 on Build GRCh38
rs142134755 Chr.2: 215615679-215615679 on Build GRCh38
rs1055601333 Chr.2: 215615672-215615672 on Build GRCh38
rs192649174 Chr.2: 215615527-215615527 on Build GRCh38
rs963886665 Chr.2: 215615503-215615503 on Build GRCh38
rs956437582 Chr.2: 215615535-215615535 on Build GRCh38
rs557074729 Chr.2: 215615634-215615634 on Build GRCh38
rs374157800 Chr.2: 215615820-215615820 on Build GRCh38
rs554166561 Chr.2: 215615738-215615738 on Build GRCh38
rs532788420 Chr.2: 215615531-215615531 on Build GRCh38
rs185591307 Chr.2: 215615592-215615592 on Build GRCh38
rs571740738 Chr.2: 215615680-215615680 on Build GRCh38
rs1028292110 Chr.2: 215615483-215615483 on Build GRCh38
rs946467803 Chr.2: 215615603-215615603 on Build GRCh38
rs995554445 Chr.2: 215615717-215615717 on Build GRCh38
rs886807738 Chr.2: 215615781-215615781 on Build GRCh38
rs987664485 Chr.2: 215615581-215615581 on Build GRCh38
rs867998431 Chr.2: 215615648-215615648 on Build GRCh38
rs954287329 Chr.2: 215615508-215615508 on Build GRCh38
rs550916811 Chr.2: 215615541-215615541 on Build GRCh38
rs778042143 Chr.2: 215615671-215615671 on Build GRCh38
rs537991289 Chr.2: 215615651-215615651 on Build GRCh38
rs141058768 Chr.2: 215615762-215615762 on Build GRCh38
rs889824339 Chr.2: 215615747-215615747 on Build GRCh38
rs1005187593 Chr.2: 215615787-215615787 on Build GRCh38
rs1028241409 Chr.2: 215615746-215615747 on Build GRCh38
rs151088693 Chr.2: 215615699-215615699 on Build GRCh38
rs946045007 Chr.2: 215615631-215615631 on Build GRCh38
rs190267359 Chr.2: 215615653-215615653 on Build GRCh38
rs912727622 Chr.2: 215615532-215615532 on Build GRCh38
rs55664438 Chr.2: 215615555-215615555 on Build GRCh38
rs549361650 Chr.2: 215615621-215615621 on Build GRCh38
rs898554955 Chr.2: 215615715-215615715 on Build GRCh38
rs995441889 Chr.2: 215615480-215615480 on Build GRCh38
rs1016694693 Chr.2: 215615801-215615801 on Build GRCh38
rs550627507 Chr.2: 215615475-215615475 on Build GRCh38

View Full Product Details