Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238226522-238227060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC151174
Entrez Gene ID
151174
Gene Name
uncharacterized LOC151174
Gene Aliases
-
UniGene
Hs.424165
Gene Chromosome Location
Chr.2: 238225113-238231677 on Build GRCh38
Transcripts
NR_026925.1 NR_026926.1

SNP Detail

SNP ID Chromosome Location
rs999171696 Chr.2: 238226933-238226933 on Build GRCh38
rs56193113 Chr.2: 238226775-238226775 on Build GRCh38
rs1007061846 Chr.2: 238226912-238226912 on Build GRCh38
rs1033520462 Chr.2: 238226791-238226791 on Build GRCh38
rs1046483458 Chr.2: 238226754-238226754 on Build GRCh38
rs909661426 Chr.2: 238226988-238226988 on Build GRCh38
rs371009906 Chr.2: 238227017-238227017 on Build GRCh38
rs879415064 Chr.2: 238226602-238226602 on Build GRCh38
rs529277057 Chr.2: 238226571-238226571 on Build GRCh38
rs971527147 Chr.2: 238226901-238226901 on Build GRCh38
rs1000252314 Chr.2: 238226640-238226640 on Build GRCh38
rs781278148 Chr.2: 238226980-238226980 on Build GRCh38
rs746005655 Chr.2: 238227002-238227002 on Build GRCh38
rs983872296 Chr.2: 238226687-238226687 on Build GRCh38
rs972326985 Chr.2: 238226923-238226923 on Build GRCh38
rs1002800384 Chr.2: 238226625-238226625 on Build GRCh38
rs951032619 Chr.2: 238226944-238226944 on Build GRCh38
rs1026526677 Chr.2: 238226934-238226934 on Build GRCh38
rs927962992 Chr.2: 238226774-238226774 on Build GRCh38
rs916742741 Chr.2: 238226957-238226957 on Build GRCh38
rs142910248 Chr.2: 238226591-238226591 on Build GRCh38
rs978325186 Chr.2: 238226683-238226683 on Build GRCh38
rs752198653 Chr.2: 238226868-238226868 on Build GRCh38
rs929697169 Chr.2: 238226559-238226559 on Build GRCh38
rs151097904 Chr.2: 238226730-238226730 on Build GRCh38
rs901111578 Chr.2: 238226729-238226729 on Build GRCh38
rs1039538469 Chr.2: 238226691-238226691 on Build GRCh38
rs534210778 Chr.2: 238226668-238226668 on Build GRCh38
rs752069102 Chr.2: 238226793-238226793 on Build GRCh38
rs944330565 Chr.2: 238226567-238226567 on Build GRCh38
rs903307225 Chr.2: 238226637-238226637 on Build GRCh38
rs1036395837 Chr.2: 238227029-238227029 on Build GRCh38
rs909598337 Chr.2: 238226688-238226688 on Build GRCh38
rs373447152 Chr.2: 238227034-238227034 on Build GRCh38
rs1046472139 Chr.2: 238226610-238226610 on Build GRCh38
rs1048746563 Chr.2: 238226898-238226898 on Build GRCh38
rs1013524325 Chr.2: 238226824-238226824 on Build GRCh38
rs1019294909 Chr.2: 238226914-238226914 on Build GRCh38
rs571180689 Chr.2: 238226577-238226577 on Build GRCh38
rs995426613 Chr.2: 238226860-238226860 on Build GRCh38
rs933804027 Chr.2: 238227032-238227032 on Build GRCh38
rs969554403 Chr.2: 238226997-238226997 on Build GRCh38
rs182352652 Chr.2: 238226690-238226690 on Build GRCh38
rs533512548 Chr.2: 238226670-238226670 on Build GRCh38
rs959131908 Chr.2: 238226655-238226655 on Build GRCh38
rs929604015 Chr.2: 238226673-238226673 on Build GRCh38
rs56151916 Chr.2: 238226736-238226736 on Build GRCh38
rs757712081 Chr.2: 238226947-238226947 on Build GRCh38
rs545910248 Chr.2: 238226737-238226737 on Build GRCh38
rs377742887 Chr.2: 238227001-238227001 on Build GRCh38
rs928056190 Chr.2: 238227013-238227013 on Build GRCh38
rs897010978 Chr.2: 238226645-238226645 on Build GRCh38
rs958567032 Chr.2: 238226939-238226939 on Build GRCh38
rs879447752 Chr.2: 238226634-238226634 on Build GRCh38
rs942507772 Chr.2: 238226589-238226589 on Build GRCh38
rs991638816 Chr.2: 238226656-238226656 on Build GRCh38
rs965034229 Chr.2: 238226928-238226928 on Build GRCh38
rs1026808527 Chr.2: 238226666-238226666 on Build GRCh38
rs377380829 Chr.2: 238226592-238226592 on Build GRCh38
rs189740688 Chr.2: 238226669-238226669 on Build GRCh38
rs566772328 Chr.2: 238226620-238226620 on Build GRCh38
rs963813314 Chr.2: 238226999-238226999 on Build GRCh38
rs1020226047 Chr.2: 238226626-238226626 on Build GRCh38
rs1033075834 Chr.2: 238226647-238226647 on Build GRCh38
rs187719383 Chr.2: 238226834-238226834 on Build GRCh38
rs551539099 Chr.2: 238226597-238226597 on Build GRCh38
rs551028079 Chr.2: 238226586-238226586 on Build GRCh38
rs1025011883 Chr.2: 238226667-238226667 on Build GRCh38

View Full Product Details