Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240959355-240959846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROCC2
Entrez Gene ID
728763
Gene Name
ciliary rootlet coiled-coil, rootletin family member 2
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.2: 240919048-240993311 on Build GRCh38
Transcripts
XM_017005450.1 NR_033841.1

Gene Symbol
LOC200772
Entrez Gene ID
200772
Gene Name
uncharacterized LOC200772
Gene Aliases
-
UniGene
Hs.647893
Gene Chromosome Location
Chr.2: 240954617-240967451 on Build GRCh38
Transcripts
XM_017005450.1 NR_033841.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7214118 c.2742G>A p.A914A Chr.2: 240959417-240959417 on Build GRCh38
6326336 c.2751G>A p.R917R Chr.2: 240959426-240959426 on Build GRCh38
4913130 c.2754C>T p.G918G Chr.2: 240959429-240959429 on Build GRCh38
5043809 c.2704G>C p.E902Q Chr.2: 240959379-240959379 on Build GRCh38
5518391 c.2755G>A p.D919N Chr.2: 240959430-240959430 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs10169006 Chr.2: 240959407-240959407 on Build GRCh38
rs529530316 Chr.2: 240959804-240959804 on Build GRCh38
rs10204458 Chr.2: 240959819-240959819 on Build GRCh38
rs776462029 Chr.2: 240959592-240959592 on Build GRCh38
rs1054852169 Chr.2: 240959429-240959429 on Build GRCh38
rs988413570 Chr.2: 240959403-240959403 on Build GRCh38
rs996231800 Chr.2: 240959621-240959621 on Build GRCh38
rs1004997986 Chr.2: 240959648-240959648 on Build GRCh38
rs533225855 Chr.2: 240959635-240959635 on Build GRCh38
rs181685204 Chr.2: 240959825-240959825 on Build GRCh38
rs1009210171 Chr.2: 240959632-240959632 on Build GRCh38
rs968253401 Chr.2: 240959649-240959649 on Build GRCh38
rs776772300 Chr.2: 240959438-240959438 on Build GRCh38
rs184411436 Chr.2: 240959425-240959425 on Build GRCh38
rs1033917712 Chr.2: 240959815-240959815 on Build GRCh38
rs12468630 Chr.2: 240959701-240959701 on Build GRCh38
rs908084083 Chr.2: 240959396-240959396 on Build GRCh38
rs772368079 Chr.2: 240959416-240959416 on Build GRCh38
rs955019660 Chr.2: 240959397-240959397 on Build GRCh38
rs978329105 Chr.2: 240959820-240959820 on Build GRCh38
rs890826207 Chr.2: 240959630-240959630 on Build GRCh38
rs10179839 Chr.2: 240959530-240959530 on Build GRCh38
rs959564411 Chr.2: 240959687-240959687 on Build GRCh38
rs952188462 Chr.2: 240959394-240959394 on Build GRCh38
rs1050846723 Chr.2: 240959763-240959763 on Build GRCh38
rs915097309 Chr.2: 240959459-240959459 on Build GRCh38
rs1008940373 Chr.2: 240959601-240959601 on Build GRCh38
rs755365298 Chr.2: 240959406-240959406 on Build GRCh38
rs749832738 Chr.2: 240959558-240959558 on Build GRCh38
rs116656887 Chr.2: 240959387-240959387 on Build GRCh38
rs1046979444 Chr.2: 240959527-240959527 on Build GRCh38
rs972037170 Chr.2: 240959673-240959673 on Build GRCh38
rs73006099 Chr.2: 240959609-240959609 on Build GRCh38
rs748250797 Chr.2: 240959414-240959414 on Build GRCh38
rs938061100 Chr.2: 240959449-240959449 on Build GRCh38
rs899655359 Chr.2: 240959579-240959579 on Build GRCh38
rs979632936 Chr.2: 240959668-240959668 on Build GRCh38
rs12468628 Chr.2: 240959694-240959694 on Build GRCh38
rs902422161 Chr.2: 240959581-240959581 on Build GRCh38
rs937730058 Chr.2: 240959426-240959426 on Build GRCh38
rs529009293 Chr.2: 240959604-240959604 on Build GRCh38
rs941307017 Chr.2: 240959731-240959731 on Build GRCh38
rs181892420 Chr.2: 240959624-240959624 on Build GRCh38
rs914961545 Chr.2: 240959809-240959809 on Build GRCh38
rs932486785 Chr.2: 240959760-240959760 on Build GRCh38
rs996326836 Chr.2: 240959386-240959386 on Build GRCh38
rs186252623 Chr.2: 240959637-240959637 on Build GRCh38
rs929295102 Chr.2: 240959524-240959524 on Build GRCh38
rs557476860 Chr.2: 240959498-240959498 on Build GRCh38
rs1001564627 Chr.2: 240959805-240959805 on Build GRCh38
rs1015214136 Chr.2: 240959654-240959654 on Build GRCh38
rs747131299 Chr.2: 240959417-240959417 on Build GRCh38
rs992675932 Chr.2: 240959689-240959689 on Build GRCh38
rs189103417 Chr.2: 240959738-240959738 on Build GRCh38
rs908474750 Chr.2: 240959721-240959721 on Build GRCh38
rs983668401 Chr.2: 240959395-240959395 on Build GRCh38
rs573603137 Chr.2: 240959699-240959699 on Build GRCh38
rs759358618 Chr.2: 240959491-240959491 on Build GRCh38
rs546714038 Chr.2: 240959733-240959733 on Build GRCh38
rs562559583 Chr.2: 240959628-240959628 on Build GRCh38
rs890858776 Chr.2: 240959774-240959774 on Build GRCh38
rs1034299156 Chr.2: 240959686-240959686 on Build GRCh38
rs1009241074 Chr.2: 240959787-240959787 on Build GRCh38
rs1020600904 Chr.2: 240959802-240959802 on Build GRCh38
rs941205898 Chr.2: 240959541-240959541 on Build GRCh38

View Full Product Details