Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 19263867-19264278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100130264
Entrez Gene ID
100130264
Gene Name
uncharacterized LOC100130264
Gene Aliases
-
UniGene
Hs.683814
Gene Chromosome Location
Chr.20: 19242302-19284596 on Build GRCh38
Transcripts
NR_024564.1 NM_020689.3

Gene Symbol
SLC24A3
Entrez Gene ID
57419
Gene Name
solute carrier family 24 member 3
Gene Aliases
NCKX3
UniGene
Hs.654790
Gene Chromosome Location
Chr.20: 19212646-19722897 on Build GRCh38
Transcripts
NR_024564.1 NM_020689.3

SNP Detail

SNP ID Chromosome Location
rs561917166 Chr.20: 19263960-19263960 on Build GRCh38
rs989437665 Chr.20: 19263907-19263907 on Build GRCh38
rs988287171 Chr.20: 19264066-19264066 on Build GRCh38
rs369171233 Chr.20: 19264069-19264070 on Build GRCh38
rs1030426521 Chr.20: 19264073-19264073 on Build GRCh38
rs1005678232 Chr.20: 19263945-19263945 on Build GRCh38
rs916476345 Chr.20: 19264172-19264172 on Build GRCh38
rs77217711 Chr.20: 19263993-19263993 on Build GRCh38
rs956493306 Chr.20: 19264020-19264020 on Build GRCh38
rs764083598 Chr.20: 19264057-19264057 on Build GRCh38
rs139502053 Chr.20: 19263965-19263965 on Build GRCh38
rs144159715 Chr.20: 19264019-19264019 on Build GRCh38
rs756992533 Chr.20: 19263926-19263926 on Build GRCh38
rs888618540 Chr.20: 19263942-19263942 on Build GRCh38
rs914958051 Chr.20: 19264067-19264067 on Build GRCh38
rs187405099 Chr.20: 19264102-19264102 on Build GRCh38
rs141013401 Chr.20: 19264189-19264189 on Build GRCh38
rs968304881 Chr.20: 19264089-19264089 on Build GRCh38
rs980602160 Chr.20: 19264090-19264090 on Build GRCh38
rs954474946 Chr.20: 19264074-19264074 on Build GRCh38
rs982121250 Chr.20: 19264201-19264201 on Build GRCh38
rs528068811 Chr.20: 19264132-19264132 on Build GRCh38
rs917248424 Chr.20: 19264233-19264233 on Build GRCh38
rs530546307 Chr.20: 19264091-19264091 on Build GRCh38
rs191626153 Chr.20: 19263909-19263909 on Build GRCh38
rs537584563 Chr.20: 19264220-19264220 on Build GRCh38
rs752566186 Chr.20: 19264137-19264137 on Build GRCh38
rs11477124 Chr.20: 19264159-19264159 on Build GRCh38
rs184621774 Chr.20: 19263994-19263994 on Build GRCh38
rs948038933 Chr.20: 19263914-19263914 on Build GRCh38
rs1008052274 Chr.20: 19264158-19264158 on Build GRCh38
rs546528767 Chr.20: 19264167-19264167 on Build GRCh38
rs568338119 Chr.20: 19264056-19264057 on Build GRCh38
rs571460387 Chr.20: 19264169-19264169 on Build GRCh38
rs869208854 Chr.20: 19264176-19264176 on Build GRCh38
rs1057196604 Chr.20: 19264182-19264182 on Build GRCh38
rs930080683 Chr.20: 19263960-19263960 on Build GRCh38
rs768575263 Chr.20: 19264122-19264122 on Build GRCh38
rs192101767 Chr.20: 19264199-19264199 on Build GRCh38
rs576484417 Chr.20: 19264121-19264121 on Build GRCh38
rs772835507 Chr.20: 19263940-19263940 on Build GRCh38
rs939234881 Chr.20: 19264176-19264176 on Build GRCh38
rs745486961 Chr.20: 19263973-19263973 on Build GRCh38
rs978268859 Chr.20: 19264097-19264097 on Build GRCh38
rs1030837728 Chr.20: 19264117-19264117 on Build GRCh38
rs6136668 Chr.20: 19264210-19264210 on Build GRCh38
rs765145489 Chr.20: 19264159-19264160 on Build GRCh38
rs34043614 Chr.20: 19264174-19264176 on Build GRCh38

View Full Product Details