Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 26083073-26083584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM182A
Entrez Gene ID
284800
Gene Name
family with sequence similarity 182 member A
Gene Aliases
C20orf91, bB329D4.1
UniGene
Hs.516956
Gene Chromosome Location
Chr.20: 26054614-26086917 on Build GRCh38
Transcripts
NR_026713.1

SNP Detail

SNP ID Chromosome Location
rs796149361 Chr.20: 26083283-26083283 on Build GRCh38
rs550468479 Chr.20: 26083558-26083558 on Build GRCh38
rs917075348 Chr.20: 26083345-26083345 on Build GRCh38
rs563946671 Chr.20: 26083208-26083208 on Build GRCh38
rs200244609 Chr.20: 26083348-26083348 on Build GRCh38
rs1025649694 Chr.20: 26083211-26083211 on Build GRCh38
rs149468670 Chr.20: 26083440-26083440 on Build GRCh38
rs199612347 Chr.20: 26083417-26083417 on Build GRCh38
rs758287521 Chr.20: 26083223-26083240 on Build GRCh38
rs73349814 Chr.20: 26083217-26083217 on Build GRCh38
rs372635886 Chr.20: 26083552-26083552 on Build GRCh38
rs529171234 Chr.20: 26083249-26083249 on Build GRCh38
rs912890263 Chr.20: 26083542-26083542 on Build GRCh38
rs78544920 Chr.20: 26083116-26083116 on Build GRCh38
rs779568819 Chr.20: 26083503-26083503 on Build GRCh38
rs201789604 Chr.20: 26083347-26083347 on Build GRCh38
rs200150456 Chr.20: 26083284-26083284 on Build GRCh38
rs772589721 Chr.20: 26083512-26083512 on Build GRCh38
rs571060940 Chr.20: 26083465-26083465 on Build GRCh38
rs375151925 Chr.20: 26083400-26083400 on Build GRCh38
rs551457171 Chr.20: 26083456-26083456 on Build GRCh38
rs45552838 Chr.20: 26083236-26083236 on Build GRCh38
rs141921294 Chr.20: 26083444-26083444 on Build GRCh38
rs868291927 Chr.20: 26083445-26083445 on Build GRCh38
rs570290294 Chr.20: 26083560-26083560 on Build GRCh38
rs376920563 Chr.20: 26083312-26083312 on Build GRCh38
rs45443092 Chr.20: 26083198-26083198 on Build GRCh38
rs201336962 Chr.20: 26083351-26083351 on Build GRCh38
rs78113344 Chr.20: 26083169-26083169 on Build GRCh38
rs960817532 Chr.20: 26083319-26083319 on Build GRCh38
rs969891837 Chr.20: 26083407-26083407 on Build GRCh38
rs200746510 Chr.20: 26083224-26083224 on Build GRCh38
rs57915816 Chr.20: 26083402-26083402 on Build GRCh38
rs879180712 Chr.20: 26083434-26083434 on Build GRCh38
rs45488097 Chr.20: 26083415-26083415 on Build GRCh38
rs45510692 Chr.20: 26083296-26083296 on Build GRCh38
rs369916281 Chr.20: 26083396-26083396 on Build GRCh38
rs1937056 Chr.20: 26083381-26083381 on Build GRCh38
rs202066058 Chr.20: 26083357-26083357 on Build GRCh38
rs370286572 Chr.20: 26083508-26083508 on Build GRCh38
rs200172661 Chr.20: 26083297-26083297 on Build GRCh38
rs376640594 Chr.20: 26083394-26083394 on Build GRCh38
rs112731559 Chr.20: 26083470-26083470 on Build GRCh38
rs935901510 Chr.20: 26083491-26083491 on Build GRCh38
rs962942887 Chr.20: 26083142-26083142 on Build GRCh38
rs79413385 Chr.20: 26083425-26083425 on Build GRCh38
rs113713481 Chr.20: 26083412-26083412 on Build GRCh38
rs925919966 Chr.20: 26083457-26083457 on Build GRCh38
rs759038916 Chr.20: 26083256-26083256 on Build GRCh38
rs878939661 Chr.20: 26083246-26083246 on Build GRCh38
rs548730345 Chr.20: 26083319-26083319 on Build GRCh38
rs190285444 Chr.20: 26083209-26083209 on Build GRCh38
rs75807365 Chr.20: 26083310-26083310 on Build GRCh38
rs543719705 Chr.20: 26083226-26083226 on Build GRCh38
rs77390076 Chr.20: 26083423-26083423 on Build GRCh38
rs111728116 Chr.20: 26083397-26083397 on Build GRCh38
rs76817215 Chr.20: 26083385-26083385 on Build GRCh38
rs994195500 Chr.20: 26083177-26083177 on Build GRCh38
rs113631568 Chr.20: 26083277-26083277 on Build GRCh38
rs559328099 Chr.20: 26083507-26083507 on Build GRCh38
rs868045735 Chr.20: 26083399-26083399 on Build GRCh38
rs748792468 Chr.20: 26083250-26083250 on Build GRCh38

View Full Product Details