Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 48367966-48368468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00494
Entrez Gene ID
284749
Gene Name
long intergenic non-protein coding RNA 494
Gene Aliases
-
UniGene
Hs.299080
Gene Chromosome Location
Chr.20: 48359911-48370638 on Build GRCh38
Transcripts
NR_026958.1

SNP Detail

SNP ID Chromosome Location
rs564273572 Chr.20: 48367999-48367999 on Build GRCh38
rs35454782 Chr.20: 48368388-48368388 on Build GRCh38
rs529020458 Chr.20: 48368134-48368134 on Build GRCh38
rs906462985 Chr.20: 48368138-48368138 on Build GRCh38
rs954164890 Chr.20: 48368319-48368319 on Build GRCh38
rs531588869 Chr.20: 48368054-48368054 on Build GRCh38
rs1003679224 Chr.20: 48368436-48368436 on Build GRCh38
rs1023323242 Chr.20: 48368084-48368084 on Build GRCh38
rs894781267 Chr.20: 48368159-48368159 on Build GRCh38
rs976972969 Chr.20: 48368306-48368306 on Build GRCh38
rs868083178 Chr.20: 48368019-48368019 on Build GRCh38
rs975532384 Chr.20: 48368189-48368189 on Build GRCh38
rs921344170 Chr.20: 48368228-48368228 on Build GRCh38
rs939238501 Chr.20: 48368146-48368146 on Build GRCh38
rs865909048 Chr.20: 48368085-48368085 on Build GRCh38
rs962682811 Chr.20: 48368149-48368149 on Build GRCh38
rs149238163 Chr.20: 48367994-48367994 on Build GRCh38
rs965625536 Chr.20: 48368305-48368305 on Build GRCh38
rs976749738 Chr.20: 48368052-48368052 on Build GRCh38
rs146043675 Chr.20: 48368395-48368395 on Build GRCh38
rs1034871245 Chr.20: 48368442-48368442 on Build GRCh38
rs1010641304 Chr.20: 48368065-48368065 on Build GRCh38
rs370927479 Chr.20: 48368435-48368435 on Build GRCh38
rs1051277548 Chr.20: 48368235-48368235 on Build GRCh38
rs6095089 Chr.20: 48368056-48368056 on Build GRCh38
rs148412729 Chr.20: 48368102-48368102 on Build GRCh38
rs551348918 Chr.20: 48368307-48368307 on Build GRCh38
rs1015844821 Chr.20: 48368022-48368022 on Build GRCh38
rs1010520479 Chr.20: 48368408-48368408 on Build GRCh38
rs781068355 Chr.20: 48368266-48368272 on Build GRCh38
rs999472879 Chr.20: 48368049-48368049 on Build GRCh38
rs909911827 Chr.20: 48368147-48368147 on Build GRCh38
rs529227864 Chr.20: 48368234-48368234 on Build GRCh38
rs144139092 Chr.20: 48368273-48368273 on Build GRCh38
rs965407963 Chr.20: 48368024-48368024 on Build GRCh38
rs989608255 Chr.20: 48368332-48368332 on Build GRCh38
rs951347169 Chr.20: 48368129-48368129 on Build GRCh38
rs80173527 Chr.20: 48368434-48368434 on Build GRCh38
rs1027806421 Chr.20: 48368225-48368225 on Build GRCh38
rs547543296 Chr.20: 48368179-48368179 on Build GRCh38
rs1015730170 Chr.20: 48368260-48368260 on Build GRCh38
rs2260121 Chr.20: 48368444-48368444 on Build GRCh38
rs1035225423 Chr.20: 48368115-48368115 on Build GRCh38
rs983058902 Chr.20: 48368002-48368002 on Build GRCh38

View Full Product Details