Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 48368641-48369148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00494
Entrez Gene ID
284749
Gene Name
long intergenic non-protein coding RNA 494
Gene Aliases
-
UniGene
Hs.299080
Gene Chromosome Location
Chr.20: 48359911-48370638 on Build GRCh38
Transcripts
NR_026958.1

SNP Detail

SNP ID Chromosome Location
rs1022271832 Chr.20: 48368903-48368903 on Build GRCh38
rs889938540 Chr.20: 48369023-48369023 on Build GRCh38
rs11905003 Chr.20: 48368904-48368904 on Build GRCh38
rs534935915 Chr.20: 48368925-48368925 on Build GRCh38
rs939010371 Chr.20: 48368858-48368858 on Build GRCh38
rs34852081 Chr.20: 48368832-48368832 on Build GRCh38
rs923018297 Chr.20: 48369005-48369005 on Build GRCh38
rs368207773 Chr.20: 48368763-48368763 on Build GRCh38
rs148936178 Chr.20: 48368905-48368905 on Build GRCh38
rs533017071 Chr.20: 48368819-48368819 on Build GRCh38
rs374082589 Chr.20: 48368759-48368759 on Build GRCh38
rs978125889 Chr.20: 48368682-48368682 on Build GRCh38
rs1044727850 Chr.20: 48368698-48368698 on Build GRCh38
rs866815153 Chr.20: 48369100-48369100 on Build GRCh38
rs750972432 Chr.20: 48369019-48369019 on Build GRCh38
rs557100252 Chr.20: 48368940-48368940 on Build GRCh38
rs191675722 Chr.20: 48368968-48368968 on Build GRCh38
rs1032170784 Chr.20: 48368816-48368816 on Build GRCh38
rs758828982 Chr.20: 48369052-48369052 on Build GRCh38
rs939142798 Chr.20: 48368709-48368709 on Build GRCh38
rs1038114399 Chr.20: 48369095-48369095 on Build GRCh38
rs926313742 Chr.20: 48368857-48368857 on Build GRCh38
rs764707009 Chr.20: 48368990-48368990 on Build GRCh38
rs187516365 Chr.20: 48368730-48368730 on Build GRCh38
rs576275271 Chr.20: 48368712-48368712 on Build GRCh38
rs1016745423 Chr.20: 48368745-48368745 on Build GRCh38
rs757762949 Chr.20: 48369032-48369032 on Build GRCh38
rs766825174 Chr.20: 48369076-48369076 on Build GRCh38
rs907562794 Chr.20: 48368969-48368969 on Build GRCh38
rs35367410 Chr.20: 48368722-48368722 on Build GRCh38
rs1011327782 Chr.20: 48368665-48368665 on Build GRCh38
rs775133089 Chr.20: 48368750-48368750 on Build GRCh38
rs567851414 Chr.20: 48368922-48368922 on Build GRCh38
rs932470342 Chr.20: 48369119-48369119 on Build GRCh38
rs990279884 Chr.20: 48368823-48368823 on Build GRCh38
rs1050361299 Chr.20: 48369014-48369014 on Build GRCh38
rs954111226 Chr.20: 48368663-48368663 on Build GRCh38
rs986928711 Chr.20: 48368814-48368814 on Build GRCh38
rs34772061 Chr.20: 48368659-48368659 on Build GRCh38
rs996795795 Chr.20: 48368908-48368908 on Build GRCh38
rs943915553 Chr.20: 48368660-48368660 on Build GRCh38
rs970180710 Chr.20: 48368970-48368970 on Build GRCh38
rs928871656 Chr.20: 48368998-48368998 on Build GRCh38
rs975863160 Chr.20: 48368999-48368999 on Build GRCh38
rs538714868 Chr.20: 48368938-48368938 on Build GRCh38
rs1043944800 Chr.20: 48369070-48369070 on Build GRCh38
rs939749534 Chr.20: 48368900-48368900 on Build GRCh38
rs559493452 Chr.20: 48368784-48368784 on Build GRCh38
rs6095090 Chr.20: 48368674-48368674 on Build GRCh38
rs952066715 Chr.20: 48368966-48368966 on Build GRCh38
rs886868729 Chr.20: 48368899-48368899 on Build GRCh38
rs898419794 Chr.20: 48368746-48368746 on Build GRCh38
rs760457829 Chr.20: 48368755-48368755 on Build GRCh38
rs555675083 Chr.20: 48369013-48369013 on Build GRCh38
rs990163849 Chr.20: 48368963-48368963 on Build GRCh38
rs535349719 Chr.20: 48368836-48368836 on Build GRCh38
rs868412322 Chr.20: 48368928-48368928 on Build GRCh38
rs546745639 Chr.20: 48368930-48368930 on Build GRCh38
rs11906911 Chr.20: 48368704-48368704 on Build GRCh38
rs11904974 Chr.20: 48368937-48368937 on Build GRCh38
rs956115469 Chr.20: 48368817-48368817 on Build GRCh38
rs6019092 Chr.20: 48368954-48368954 on Build GRCh38
rs1054754214 Chr.20: 48368672-48368672 on Build GRCh38
rs1056248615 Chr.20: 48368866-48368866 on Build GRCh38
rs992240034 Chr.20: 48369012-48369012 on Build GRCh38
rs948933884 Chr.20: 48368676-48368676 on Build GRCh38
rs886982999 Chr.20: 48368729-48368729 on Build GRCh38
rs541218836 Chr.20: 48368768-48368768 on Build GRCh38
rs938910311 Chr.20: 48369006-48369006 on Build GRCh38
rs1011129605 Chr.20: 48368888-48368889 on Build GRCh38
rs397834607 Chr.20: 48368846-48368846 on Build GRCh38

View Full Product Details