Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44933468-44933965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01547
Entrez Gene ID
84536
Gene Name
long intergenic non-protein coding RNA 1547
Gene Aliases
C21orf67, C21orf69, PRED54
UniGene
Hs.534504
Gene Chromosome Location
Chr.21: 44933284-44939913 on Build GRCh38
Transcripts
NR_027128.1

SNP Detail

SNP ID Chromosome Location
rs916364404 Chr.21: 44933718-44933718 on Build GRCh38
rs989721328 Chr.21: 44933876-44933876 on Build GRCh38
rs1021533448 Chr.21: 44933593-44933593 on Build GRCh38
rs73376586 Chr.21: 44933914-44933914 on Build GRCh38
rs984366756 Chr.21: 44933725-44933725 on Build GRCh38
rs891502056 Chr.21: 44933698-44933698 on Build GRCh38
rs9984679 Chr.21: 44933758-44933758 on Build GRCh38
rs9983985 Chr.21: 44933723-44933723 on Build GRCh38
rs367981595 Chr.21: 44933899-44933899 on Build GRCh38
rs972873325 Chr.21: 44933646-44933646 on Build GRCh38
rs1024019380 Chr.21: 44933710-44933710 on Build GRCh38
rs562515556 Chr.21: 44933898-44933898 on Build GRCh38
rs377425380 Chr.21: 44933548-44933548 on Build GRCh38
rs375444305 Chr.21: 44933855-44933855 on Build GRCh38
rs371438266 Chr.21: 44933826-44933826 on Build GRCh38
rs935898007 Chr.21: 44933842-44933842 on Build GRCh38
rs958272599 Chr.21: 44933860-44933860 on Build GRCh38
rs922424147 Chr.21: 44933653-44933653 on Build GRCh38
rs901385232 Chr.21: 44933830-44933830 on Build GRCh38
rs766325456 Chr.21: 44933868-44933868 on Build GRCh38
rs1032699491 Chr.21: 44933512-44933512 on Build GRCh38
rs982691000 Chr.21: 44933596-44933596 on Build GRCh38
rs371173682 Chr.21: 44933659-44933659 on Build GRCh38
rs995501707 Chr.21: 44933638-44933638 on Build GRCh38
rs372247796 Chr.21: 44933853-44933853 on Build GRCh38
rs141331122 Chr.21: 44933495-44933495 on Build GRCh38
rs370649049 Chr.21: 44933721-44933721 on Build GRCh38
rs185167346 Chr.21: 44933621-44933621 on Build GRCh38
rs560825101 Chr.21: 44933643-44933643 on Build GRCh38
rs7282201 Chr.21: 44933763-44933763 on Build GRCh38
rs182301348 Chr.21: 44933532-44933532 on Build GRCh38
rs79917387 Chr.21: 44933675-44933675 on Build GRCh38
rs536890344 Chr.21: 44933608-44933608 on Build GRCh38
rs906927114 Chr.21: 44933906-44933906 on Build GRCh38
rs7278544 Chr.21: 44933722-44933722 on Build GRCh38
rs368315685 Chr.21: 44933839-44933839 on Build GRCh38
rs1036317358 Chr.21: 44933755-44933755 on Build GRCh38
rs376272638 Chr.21: 44933696-44933696 on Build GRCh38
rs370340960 Chr.21: 44933752-44933752 on Build GRCh38
rs922676169 Chr.21: 44933811-44933811 on Build GRCh38
rs1007387477 Chr.21: 44933820-44933820 on Build GRCh38
rs62214491 Chr.21: 44933682-44933682 on Build GRCh38
rs747956275 Chr.21: 44933644-44933644 on Build GRCh38
rs1016880416 Chr.21: 44933741-44933741 on Build GRCh38
rs749251709 Chr.21: 44933717-44933717 on Build GRCh38
rs569559001 Chr.21: 44933549-44933549 on Build GRCh38
rs139251218 Chr.21: 44933762-44933762 on Build GRCh38
rs576853446 Chr.21: 44933683-44933683 on Build GRCh38
rs537832285 Chr.21: 44933709-44933709 on Build GRCh38
rs984062588 Chr.21: 44933639-44933639 on Build GRCh38
rs780862722 Chr.21: 44933594-44933594 on Build GRCh38
rs1004822792 Chr.21: 44933577-44933577 on Build GRCh38
rs955663303 Chr.21: 44933818-44933818 on Build GRCh38
rs80246834 Chr.21: 44933828-44933828 on Build GRCh38
rs1050041092 Chr.21: 44933668-44933668 on Build GRCh38
rs1010020694 Chr.21: 44933558-44933558 on Build GRCh38

View Full Product Details