Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26657327-26657828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIAT
Entrez Gene ID
440823
Gene Name
myocardial infarction associated transcript (non-protein coding)
Gene Aliases
C22orf35, GOMAFU, LINC00066, NCRNA00066, RNCR2, lncRNA-MIAT
UniGene
Hs.517502
Gene Chromosome Location
Chr.22: 26657482-26676478 on Build GRCh38
Transcripts
NR_003491.3 NR_033319.2 NR_033320.2 NR_033321.2

SNP Detail

SNP ID Chromosome Location
rs959523529 Chr.22: 26657551-26657551 on Build GRCh38
rs1015681342 Chr.22: 26657506-26657506 on Build GRCh38
rs544965424 Chr.22: 26657591-26657591 on Build GRCh38
rs988480408 Chr.22: 26657421-26657421 on Build GRCh38
rs940423341 Chr.22: 26657672-26657673 on Build GRCh38
rs1034881500 Chr.22: 26657538-26657538 on Build GRCh38
rs868071319 Chr.22: 26657356-26657356 on Build GRCh38
rs1050169304 Chr.22: 26657463-26657463 on Build GRCh38
rs976858484 Chr.22: 26657739-26657739 on Build GRCh38
rs914325085 Chr.22: 26657427-26657427 on Build GRCh38
rs1044991899 Chr.22: 26657736-26657736 on Build GRCh38
rs974799426 Chr.22: 26657443-26657443 on Build GRCh38
rs8142890 Chr.22: 26657414-26657414 on Build GRCh38
rs923725795 Chr.22: 26657365-26657365 on Build GRCh38
rs922790031 Chr.22: 26657756-26657756 on Build GRCh38
rs977875768 Chr.22: 26657363-26657363 on Build GRCh38
rs901418150 Chr.22: 26657452-26657452 on Build GRCh38
rs1020452063 Chr.22: 26657515-26657515 on Build GRCh38
rs932929626 Chr.22: 26657457-26657457 on Build GRCh38
rs1051523888 Chr.22: 26657420-26657420 on Build GRCh38
rs564045456 Chr.22: 26657605-26657605 on Build GRCh38
rs772993467 Chr.22: 26657553-26657553 on Build GRCh38
rs562619035 Chr.22: 26657476-26657476 on Build GRCh38
rs183564910 Chr.22: 26657804-26657804 on Build GRCh38
rs1035479500 Chr.22: 26657408-26657408 on Build GRCh38
rs1003360061 Chr.22: 26657512-26657512 on Build GRCh38
rs528425106 Chr.22: 26657644-26657644 on Build GRCh38
rs979005721 Chr.22: 26657522-26657522 on Build GRCh38
rs1036128747 Chr.22: 26657432-26657432 on Build GRCh38
rs944487229 Chr.22: 26657611-26657611 on Build GRCh38
rs940539268 Chr.22: 26657428-26657428 on Build GRCh38
rs1036184587 Chr.22: 26657673-26657673 on Build GRCh38
rs934825475 Chr.22: 26657438-26657438 on Build GRCh38
rs555729462 Chr.22: 26657371-26657371 on Build GRCh38
rs865894659 Chr.22: 26657470-26657470 on Build GRCh38
rs1001903288 Chr.22: 26657501-26657501 on Build GRCh38
rs546866826 Chr.22: 26657735-26657735 on Build GRCh38
rs968992258 Chr.22: 26657520-26657520 on Build GRCh38
rs921605794 Chr.22: 26657380-26657380 on Build GRCh38
rs994702439 Chr.22: 26657491-26657491 on Build GRCh38
rs561770699 Chr.22: 26657772-26657772 on Build GRCh38
rs1048910426 Chr.22: 26657496-26657496 on Build GRCh38
rs574167485 Chr.22: 26657398-26657398 on Build GRCh38
rs371656892 Chr.22: 26657587-26657587 on Build GRCh38
rs947042132 Chr.22: 26657436-26657436 on Build GRCh38
rs946418953 Chr.22: 26657354-26657354 on Build GRCh38
rs905862603 Chr.22: 26657499-26657499 on Build GRCh38
rs535658240 Chr.22: 26657425-26657425 on Build GRCh38
rs1022344440 Chr.22: 26657588-26657588 on Build GRCh38
rs930603680 Chr.22: 26657703-26657703 on Build GRCh38
rs1006017513 Chr.22: 26657502-26657502 on Build GRCh38
rs937371994 Chr.22: 26657770-26657770 on Build GRCh38
rs923671833 Chr.22: 26657609-26657609 on Build GRCh38
rs920423261 Chr.22: 26657685-26657685 on Build GRCh38
rs147062430 Chr.22: 26657510-26657510 on Build GRCh38

View Full Product Details