Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 27921848-27922348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC28-AS1
Entrez Gene ID
284900
Gene Name
TTC28 antisense RNA 1
Gene Aliases
TTC28-AS, TTC28AS, dJ353E16.2
UniGene
Hs.602319
Gene Chromosome Location
Chr.22: 27919376-28002679 on Build GRCh38
Transcripts
NR_026962.1 NR_026963.1

SNP Detail

SNP ID Chromosome Location
rs879797842 Chr.22: 27922242-27922243 on Build GRCh38
rs769744520 Chr.22: 27922186-27922186 on Build GRCh38
rs746625583 Chr.22: 27921956-27921956 on Build GRCh38
rs1016373583 Chr.22: 27921948-27921948 on Build GRCh38
rs1049215252 Chr.22: 27922107-27922107 on Build GRCh38
rs900344156 Chr.22: 27922181-27922181 on Build GRCh38
rs5752658 Chr.22: 27922044-27922044 on Build GRCh38
rs182437585 Chr.22: 27921899-27921899 on Build GRCh38
rs911893548 Chr.22: 27922245-27922245 on Build GRCh38
rs369735433 Chr.22: 27921897-27921897 on Build GRCh38
rs1031240085 Chr.22: 27922323-27922323 on Build GRCh38
rs550085818 Chr.22: 27922084-27922084 on Build GRCh38
rs576165449 Chr.22: 27921875-27921875 on Build GRCh38
rs906171728 Chr.22: 27922154-27922154 on Build GRCh38
rs945436170 Chr.22: 27921947-27921947 on Build GRCh38
rs1003610284 Chr.22: 27922163-27922163 on Build GRCh38
rs150337342 Chr.22: 27922164-27922164 on Build GRCh38
rs888229141 Chr.22: 27922108-27922108 on Build GRCh38
rs1023772648 Chr.22: 27922267-27922267 on Build GRCh38
rs1012750221 Chr.22: 27922259-27922259 on Build GRCh38
rs566104070 Chr.22: 27922286-27922286 on Build GRCh38
rs978176459 Chr.22: 27922227-27922227 on Build GRCh38
rs947267971 Chr.22: 27921874-27921874 on Build GRCh38
rs997435415 Chr.22: 27921894-27921894 on Build GRCh38
rs922246221 Chr.22: 27922305-27922305 on Build GRCh38
rs886452609 Chr.22: 27921935-27921935 on Build GRCh38
rs900455562 Chr.22: 27921888-27921888 on Build GRCh38
rs541326806 Chr.22: 27921923-27921923 on Build GRCh38
rs1039257952 Chr.22: 27922045-27922045 on Build GRCh38
rs915369008 Chr.22: 27921939-27921939 on Build GRCh38
rs191118442 Chr.22: 27921959-27921959 on Build GRCh38
rs761886310 Chr.22: 27922264-27922264 on Build GRCh38
rs10483140 Chr.22: 27922204-27922204 on Build GRCh38
rs540261740 Chr.22: 27922272-27922272 on Build GRCh38
rs1024306600 Chr.22: 27922067-27922067 on Build GRCh38
rs1015695527 Chr.22: 27922134-27922134 on Build GRCh38
rs959273289 Chr.22: 27922166-27922166 on Build GRCh38
rs1003729599 Chr.22: 27922296-27922296 on Build GRCh38
rs989347560 Chr.22: 27922175-27922175 on Build GRCh38
rs975128609 Chr.22: 27922302-27922302 on Build GRCh38
rs934348724 Chr.22: 27922117-27922117 on Build GRCh38
rs970885783 Chr.22: 27922290-27922290 on Build GRCh38
rs186993325 Chr.22: 27921927-27921927 on Build GRCh38
rs542722129 Chr.22: 27922018-27922018 on Build GRCh38
rs1022104513 Chr.22: 27922203-27922203 on Build GRCh38
rs1030303289 Chr.22: 27921918-27921918 on Build GRCh38
rs900737477 Chr.22: 27922081-27922081 on Build GRCh38
rs970916848 Chr.22: 27922080-27922080 on Build GRCh38
rs1016754051 Chr.22: 27922232-27922232 on Build GRCh38
rs569909343 Chr.22: 27922183-27922183 on Build GRCh38
rs1003881739 Chr.22: 27922131-27922131 on Build GRCh38
rs978095672 Chr.22: 27921954-27921954 on Build GRCh38

View Full Product Details