Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186491994-186492496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC253573
Entrez Gene ID
253573
Gene Name
uncharacterized LOC253573
Gene Aliases
-
UniGene
Hs.97386
Gene Chromosome Location
Chr.3: 186454981-186493661 on Build GRCh38
Transcripts
NR_033844.1

SNP Detail

SNP ID Chromosome Location
rs989109731 Chr.3: 186492280-186492280 on Build GRCh38
rs1005587917 Chr.3: 186492081-186492081 on Build GRCh38
rs964137169 Chr.3: 186492364-186492364 on Build GRCh38
rs1039743987 Chr.3: 186492416-186492416 on Build GRCh38
rs928222148 Chr.3: 186492305-186492305 on Build GRCh38
rs115684382 Chr.3: 186492324-186492324 on Build GRCh38
rs969682456 Chr.3: 186492292-186492292 on Build GRCh38
rs957751620 Chr.3: 186492273-186492273 on Build GRCh38
rs890370895 Chr.3: 186492442-186492445 on Build GRCh38
rs922865869 Chr.3: 186492073-186492073 on Build GRCh38
rs1047616086 Chr.3: 186492075-186492075 on Build GRCh38
rs9870051 Chr.3: 186492059-186492059 on Build GRCh38
rs769315016 Chr.3: 186492377-186492377 on Build GRCh38
rs530428877 Chr.3: 186492281-186492281 on Build GRCh38
rs542677943 Chr.3: 186492097-186492097 on Build GRCh38
rs930191538 Chr.3: 186492083-186492083 on Build GRCh38
rs930263609 Chr.3: 186492386-186492386 on Build GRCh38
rs558790874 Chr.3: 186492049-186492049 on Build GRCh38
rs1037836406 Chr.3: 186492103-186492103 on Build GRCh38
rs9870751 Chr.3: 186492464-186492464 on Build GRCh38
rs968688892 Chr.3: 186492319-186492319 on Build GRCh38
rs941385091 Chr.3: 186492367-186492367 on Build GRCh38
rs574422841 Chr.3: 186492118-186492118 on Build GRCh38
rs954731083 Chr.3: 186492225-186492225 on Build GRCh38
rs748446196 Chr.3: 186492093-186492093 on Build GRCh38
rs772440089 Chr.3: 186492145-186492145 on Build GRCh38
rs1035504771 Chr.3: 186492243-186492243 on Build GRCh38
rs1052139092 Chr.3: 186492441-186492441 on Build GRCh38
rs933993329 Chr.3: 186492438-186492438 on Build GRCh38
rs774910246 Chr.3: 186492435-186492435 on Build GRCh38
rs74845605 Chr.3: 186492076-186492077 on Build GRCh38
rs77954587 Chr.3: 186492313-186492313 on Build GRCh38
rs909974576 Chr.3: 186492326-186492326 on Build GRCh38
rs207464158 Chr.3: 186492235-186492235 on Build GRCh38
rs560972956 Chr.3: 186492468-186492468 on Build GRCh38
rs532713554 Chr.3: 186492414-186492414 on Build GRCh38
rs916839778 Chr.3: 186492058-186492058 on Build GRCh38
rs888527519 Chr.3: 186492078-186492078 on Build GRCh38
rs977330014 Chr.3: 186492376-186492376 on Build GRCh38
rs779431588 Chr.3: 186492067-186492067 on Build GRCh38
rs905722729 Chr.3: 186492180-186492180 on Build GRCh38
rs776334895 Chr.3: 186492216-186492216 on Build GRCh38
rs374483545 Chr.3: 186492442-186492442 on Build GRCh38
rs545134999 Chr.3: 186492371-186492371 on Build GRCh38
rs1030742956 Chr.3: 186492267-186492267 on Build GRCh38
rs1010122194 Chr.3: 186492277-186492277 on Build GRCh38
rs930149748 Chr.3: 186492075-186492078 on Build GRCh38
rs982803137 Chr.3: 186492299-186492299 on Build GRCh38
rs1005963710 Chr.3: 186492101-186492101 on Build GRCh38
rs899801749 Chr.3: 186492425-186492425 on Build GRCh38
rs1041190371 Chr.3: 186492024-186492024 on Build GRCh38
rs911309393 Chr.3: 186492345-186492345 on Build GRCh38
rs540889039 Chr.3: 186492086-186492086 on Build GRCh38
rs1001785343 Chr.3: 186492229-186492229 on Build GRCh38
rs541414422 Chr.3: 186492141-186492141 on Build GRCh38
rs996471299 Chr.3: 186492194-186492194 on Build GRCh38
rs66971127 Chr.3: 186492067-186492067 on Build GRCh38
rs562988684 Chr.3: 186492268-186492268 on Build GRCh38

View Full Product Details