Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1195989-1196498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100130872
Entrez Gene ID
100130872
Gene Name
uncharacterized LOC100130872
Gene Aliases
-
UniGene
Hs.302963
Gene Chromosome Location
Chr.4: 1195783-1208962 on Build GRCh38
Transcripts
NR_024569.1 NM_001199021.1

Gene Symbol
SPON2
Entrez Gene ID
10417
Gene Name
spondin 2
Gene Aliases
DIL-1, DIL1, M-SPONDIN, MINDIN
UniGene
Hs.302963
Gene Chromosome Location
Chr.4: 1166932-1208962 on Build GRCh38
Transcripts
NR_024569.1 NM_001199021.1

SNP Detail

SNP ID Chromosome Location
rs568993431 Chr.4: 1196338-1196338 on Build GRCh38
rs947293482 Chr.4: 1196149-1196149 on Build GRCh38
rs1047686436 Chr.4: 1196333-1196333 on Build GRCh38
rs578010580 Chr.4: 1196442-1196442 on Build GRCh38
rs539806047 Chr.4: 1196161-1196161 on Build GRCh38
rs933299525 Chr.4: 1196183-1196183 on Build GRCh38
rs199518978 Chr.4: 1196175-1196175 on Build GRCh38
rs773561663 Chr.4: 1196433-1196433 on Build GRCh38
rs937784798 Chr.4: 1196282-1196282 on Build GRCh38
rs912999531 Chr.4: 1196352-1196352 on Build GRCh38
rs561181866 Chr.4: 1196415-1196415 on Build GRCh38
rs765447184 Chr.4: 1196407-1196407 on Build GRCh38
rs549556585 Chr.4: 1196400-1196400 on Build GRCh38
rs954716864 Chr.4: 1196475-1196475 on Build GRCh38
rs1006140855 Chr.4: 1196396-1196396 on Build GRCh38
rs996682491 Chr.4: 1196426-1196426 on Build GRCh38
rs1013524483 Chr.4: 1196141-1196141 on Build GRCh38
rs546986948 Chr.4: 1196462-1196462 on Build GRCh38
rs896160821 Chr.4: 1196307-1196307 on Build GRCh38
rs554367493 Chr.4: 1196106-1196106 on Build GRCh38
rs886635853 Chr.4: 1196201-1196202 on Build GRCh38
rs1026214840 Chr.4: 1196142-1196142 on Build GRCh38
rs903894283 Chr.4: 1196071-1196071 on Build GRCh38
rs1001601993 Chr.4: 1196262-1196262 on Build GRCh38
rs1013221103 Chr.4: 1196327-1196327 on Build GRCh38
rs984590211 Chr.4: 1196154-1196154 on Build GRCh38
rs190884773 Chr.4: 1196448-1196448 on Build GRCh38
rs998057737 Chr.4: 1196090-1196090 on Build GRCh38
rs978569109 Chr.4: 1196377-1196377 on Build GRCh38
rs577889855 Chr.4: 1196180-1196180 on Build GRCh38
rs575496493 Chr.4: 1196053-1196053 on Build GRCh38
rs911105002 Chr.4: 1196383-1196383 on Build GRCh38
rs997647738 Chr.4: 1196196-1196196 on Build GRCh38
rs1018083723 Chr.4: 1196086-1196086 on Build GRCh38
rs550731261 Chr.4: 1196200-1196200 on Build GRCh38
rs947629929 Chr.4: 1196229-1196229 on Build GRCh38
rs747408674 Chr.4: 1196244-1196244 on Build GRCh38
rs891707143 Chr.4: 1196015-1196015 on Build GRCh38
rs545279539 Chr.4: 1196205-1196205 on Build GRCh38
rs984890402 Chr.4: 1196032-1196032 on Build GRCh38
rs943392469 Chr.4: 1196055-1196055 on Build GRCh38
rs896142487 Chr.4: 1196132-1196132 on Build GRCh38
rs1030376100 Chr.4: 1196447-1196447 on Build GRCh38
rs528598018 Chr.4: 1196452-1196452 on Build GRCh38
rs369656979 Chr.4: 1196277-1196277 on Build GRCh38
rs1033785384 Chr.4: 1196131-1196131 on Build GRCh38
rs1054856548 Chr.4: 1196293-1196293 on Build GRCh38
rs969683727 Chr.4: 1196108-1196108 on Build GRCh38
rs1053271441 Chr.4: 1196075-1196075 on Build GRCh38
rs950534958 Chr.4: 1196150-1196150 on Build GRCh38
rs975183625 Chr.4: 1196068-1196068 on Build GRCh38
rs769237437 Chr.4: 1196279-1196279 on Build GRCh38
rs911158709 Chr.4: 1196157-1196157 on Build GRCh38
rs185927091 Chr.4: 1196031-1196031 on Build GRCh38
rs1039044450 Chr.4: 1196162-1196162 on Build GRCh38
rs73067802 Chr.4: 1196228-1196228 on Build GRCh38
rs923295334 Chr.4: 1196069-1196069 on Build GRCh38
rs1001956337 Chr.4: 1196109-1196109 on Build GRCh38
rs947517742 Chr.4: 1196100-1196100 on Build GRCh38
rs1026515637 Chr.4: 1196099-1196099 on Build GRCh38
rs988607423 Chr.4: 1196334-1196334 on Build GRCh38
rs1035402015 Chr.4: 1196269-1196269 on Build GRCh38
rs906011388 Chr.4: 1196271-1196271 on Build GRCh38
rs760797886 Chr.4: 1196199-1196199 on Build GRCh38
rs970299510 Chr.4: 1196253-1196253 on Build GRCh38
rs1006406089 Chr.4: 1196204-1196204 on Build GRCh38
rs928333786 Chr.4: 1196115-1196115 on Build GRCh38
rs536489884 Chr.4: 1196089-1196089 on Build GRCh38
rs913091413 Chr.4: 1196147-1196147 on Build GRCh38
rs942665033 Chr.4: 1196387-1196387 on Build GRCh38
rs933603133 Chr.4: 1196074-1196074 on Build GRCh38
rs73793109 Chr.4: 1196357-1196357 on Build GRCh38
rs369416244 Chr.4: 1196156-1196156 on Build GRCh38
rs1043655372 Chr.4: 1196231-1196231 on Build GRCh38
rs1043462449 Chr.4: 1196101-1196101 on Build GRCh38

View Full Product Details