Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 119460371-119460895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC645513
Entrez Gene ID
645513
Gene Name
septin 7 pseudogene
Gene Aliases
-
UniGene
Hs.677546
Gene Chromosome Location
Chr.4: 119454783-119499592 on Build GRCh38
Transcripts
NR_037630.1

SNP Detail

SNP ID Chromosome Location
rs977731962 Chr.4: 119460760-119460760 on Build GRCh38
rs9994810 Chr.4: 119460435-119460435 on Build GRCh38
rs868534161 Chr.4: 119460624-119460624 on Build GRCh38
rs182285994 Chr.4: 119460414-119460414 on Build GRCh38
rs911069309 Chr.4: 119460825-119460825 on Build GRCh38
rs191841349 Chr.4: 119460727-119460727 on Build GRCh38
rs10020027 Chr.4: 119460724-119460724 on Build GRCh38
rs929950836 Chr.4: 119460659-119460659 on Build GRCh38
rs1011825522 Chr.4: 119460717-119460717 on Build GRCh38
rs1001599624 Chr.4: 119460703-119460703 on Build GRCh38
rs752355107 Chr.4: 119460407-119460410 on Build GRCh38
rs547215393 Chr.4: 119460614-119460614 on Build GRCh38
rs1045726605 Chr.4: 119460662-119460662 on Build GRCh38
rs190240887 Chr.4: 119460745-119460745 on Build GRCh38
rs955312034 Chr.4: 119460793-119460793 on Build GRCh38
rs933717883 Chr.4: 119460545-119460545 on Build GRCh38
rs540984616 Chr.4: 119460708-119460708 on Build GRCh38
rs530233958 Chr.4: 119460695-119460695 on Build GRCh38
rs9994730 Chr.4: 119460409-119460409 on Build GRCh38
rs941201046 Chr.4: 119460828-119460828 on Build GRCh38
rs527274199 Chr.4: 119460539-119460539 on Build GRCh38
rs7356290 Chr.4: 119460872-119460872 on Build GRCh38
rs985714914 Chr.4: 119460573-119460573 on Build GRCh38
rs373578542 Chr.4: 119460754-119460754 on Build GRCh38
rs941303602 Chr.4: 119460643-119460643 on Build GRCh38
rs777282084 Chr.4: 119460770-119460772 on Build GRCh38
rs184575506 Chr.4: 119460738-119460738 on Build GRCh38
rs7356491 Chr.4: 119460819-119460819 on Build GRCh38
rs866790644 Chr.4: 119460466-119460466 on Build GRCh38
rs911028953 Chr.4: 119460605-119460605 on Build GRCh38
rs192840506 Chr.4: 119460770-119460770 on Build GRCh38
rs977845135 Chr.4: 119460459-119460459 on Build GRCh38
rs201951101 Chr.4: 119460852-119460852 on Build GRCh38
rs922364949 Chr.4: 119460479-119460479 on Build GRCh38
rs897197159 Chr.4: 119460655-119460655 on Build GRCh38
rs6848530 Chr.4: 119460653-119460653 on Build GRCh38
rs1034961553 Chr.4: 119460705-119460705 on Build GRCh38
rs186636519 Chr.4: 119460484-119460484 on Build GRCh38
rs761862452 Chr.4: 119460666-119460666 on Build GRCh38

View Full Product Details