Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 173541631-173542147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAND2-AS1
Entrez Gene ID
79804
Gene Name
HAND2 antisense RNA 1 (head to head)
Gene Aliases
DEIN, NBLA00301
UniGene
Hs.61435
Gene Chromosome Location
Chr.4: 173528600-173591255 on Build GRCh38
Transcripts
NR_003679.2 NR_136192.1 NR_136193.1 NR_136194.1 NR_136195.1 NR_136196.1 NR_136197.1 NR_136198.1 NR_136199.1 NR_136200.1 NR_136201.1

SNP Detail

SNP ID Chromosome Location
rs965467974 Chr.4: 173541790-173541790 on Build GRCh38
rs576251569 Chr.4: 173541700-173541700 on Build GRCh38
rs796857004 Chr.4: 173542033-173542034 on Build GRCh38
rs1007046772 Chr.4: 173541788-173541788 on Build GRCh38
rs902128158 Chr.4: 173541710-173541710 on Build GRCh38
rs1050117097 Chr.4: 173542039-173542039 on Build GRCh38
rs992645754 Chr.4: 173541930-173541930 on Build GRCh38
rs1059045 Chr.4: 173541824-173541824 on Build GRCh38
rs762251782 Chr.4: 173541800-173541800 on Build GRCh38
rs942964259 Chr.4: 173542070-173542070 on Build GRCh38
rs369928714 Chr.4: 173541958-173541958 on Build GRCh38
rs1041379057 Chr.4: 173541667-173541667 on Build GRCh38
rs561639428 Chr.4: 173541876-173541876 on Build GRCh38
rs890072772 Chr.4: 173541761-173541761 on Build GRCh38
rs181454072 Chr.4: 173541662-173541662 on Build GRCh38
rs541333683 Chr.4: 173541859-173541859 on Build GRCh38
rs55727418 Chr.4: 173541842-173541842 on Build GRCh38
rs886661845 Chr.4: 173542062-173542062 on Build GRCh38
rs1001082926 Chr.4: 173541731-173541731 on Build GRCh38
rs1025682248 Chr.4: 173542107-173542107 on Build GRCh38
rs888555999 Chr.4: 173542111-173542111 on Build GRCh38
rs1016419794 Chr.4: 173542110-173542110 on Build GRCh38
rs925725358 Chr.4: 173541979-173541979 on Build GRCh38
rs549502045 Chr.4: 173542036-173542036 on Build GRCh38
rs1017529693 Chr.4: 173541679-173541679 on Build GRCh38
rs1026517386 Chr.4: 173541749-173541749 on Build GRCh38
rs372196771 Chr.4: 173542061-173542061 on Build GRCh38
rs776925359 Chr.4: 173541738-173541738 on Build GRCh38
rs111786259 Chr.4: 173541789-173541789 on Build GRCh38
rs935649991 Chr.4: 173541996-173541996 on Build GRCh38
rs904191825 Chr.4: 173541701-173541701 on Build GRCh38
rs1041179268 Chr.4: 173542079-173542079 on Build GRCh38
rs994332193 Chr.4: 173542094-173542094 on Build GRCh38
rs917086677 Chr.4: 173541937-173541937 on Build GRCh38
rs544979255 Chr.4: 173541820-173541820 on Build GRCh38
rs185794008 Chr.4: 173541756-173541756 on Build GRCh38
rs943432955 Chr.4: 173541971-173541971 on Build GRCh38
rs949976015 Chr.4: 173541906-173541906 on Build GRCh38
rs773890436 Chr.4: 173541838-173541838 on Build GRCh38
rs981755148 Chr.4: 173541923-173541923 on Build GRCh38
rs766950272 Chr.4: 173541949-173541949 on Build GRCh38
rs901347384 Chr.4: 173542084-173542084 on Build GRCh38
rs945797629 Chr.4: 173541663-173541663 on Build GRCh38
rs950947114 Chr.4: 173541751-173541751 on Build GRCh38
rs550010846 Chr.4: 173542041-173542041 on Build GRCh38
rs540513889 Chr.4: 173541863-173541863 on Build GRCh38
rs945774832 Chr.4: 173541942-173541942 on Build GRCh38
rs11132988 Chr.4: 173541924-173541924 on Build GRCh38
rs901843529 Chr.4: 173542016-173542016 on Build GRCh38
rs984127877 Chr.4: 173541849-173541849 on Build GRCh38

View Full Product Details