Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183494857-183495364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC389247
Entrez Gene ID
389247
Gene Name
uncharacterized LOC389247
Gene Aliases
-
UniGene
Hs.385787
Gene Chromosome Location
Chr.4: 183494737-183504515 on Build GRCh38
Transcripts
NR_033995.2

SNP Detail

SNP ID Chromosome Location
rs995335701 Chr.4: 183495277-183495277 on Build GRCh38
rs535804247 Chr.4: 183494951-183494951 on Build GRCh38
rs1005082314 Chr.4: 183495320-183495320 on Build GRCh38
rs578188874 Chr.4: 183495152-183495152 on Build GRCh38
rs999905564 Chr.4: 183495037-183495037 on Build GRCh38
rs745850214 Chr.4: 183495082-183495082 on Build GRCh38
rs148319518 Chr.4: 183495177-183495177 on Build GRCh38
rs945659446 Chr.4: 183495241-183495241 on Build GRCh38
rs1019052331 Chr.4: 183495321-183495321 on Build GRCh38
rs986220304 Chr.4: 183495182-183495182 on Build GRCh38
rs962298597 Chr.4: 183495136-183495136 on Build GRCh38
rs528753073 Chr.4: 183494983-183494983 on Build GRCh38
rs946504419 Chr.4: 183495029-183495029 on Build GRCh38
rs192684969 Chr.4: 183495185-183495185 on Build GRCh38
rs775619544 Chr.4: 183495294-183495294 on Build GRCh38
rs1043906335 Chr.4: 183495000-183495000 on Build GRCh38
rs73870572 Chr.4: 183495063-183495063 on Build GRCh38
rs974404512 Chr.4: 183494984-183494984 on Build GRCh38
rs976520905 Chr.4: 183495210-183495210 on Build GRCh38
rs756735580 Chr.4: 183494976-183494976 on Build GRCh38
rs541183092 Chr.4: 183494994-183494994 on Build GRCh38
rs73870573 Chr.4: 183495154-183495154 on Build GRCh38
rs532953564 Chr.4: 183495295-183495295 on Build GRCh38
rs143275499 Chr.4: 183495157-183495157 on Build GRCh38
rs749573731 Chr.4: 183495181-183495181 on Build GRCh38
rs755575828 Chr.4: 183495040-183495040 on Build GRCh38
rs771028551 Chr.4: 183495264-183495264 on Build GRCh38
rs753487444 Chr.4: 183494926-183494926 on Build GRCh38
rs866997855 Chr.4: 183494894-183494894 on Build GRCh38
rs186462399 Chr.4: 183495078-183495078 on Build GRCh38
rs986210248 Chr.4: 183494883-183494883 on Build GRCh38
rs373037665 Chr.4: 183495156-183495156 on Build GRCh38
rs756431832 Chr.4: 183495143-183495143 on Build GRCh38
rs558266158 Chr.4: 183495033-183495033 on Build GRCh38
rs950875632 Chr.4: 183494901-183494902 on Build GRCh38
rs984128563 Chr.4: 183494931-183494931 on Build GRCh38
rs544074998 Chr.4: 183495066-183495066 on Build GRCh38
rs1014312449 Chr.4: 183495094-183495094 on Build GRCh38
rs62358464 Chr.4: 183494993-183494993 on Build GRCh38
rs1023955499 Chr.4: 183495114-183495114 on Build GRCh38
rs913897661 Chr.4: 183495222-183495222 on Build GRCh38
rs748524232 Chr.4: 183495137-183495137 on Build GRCh38
rs184666366 Chr.4: 183495186-183495186 on Build GRCh38
rs1055485922 Chr.4: 183495330-183495330 on Build GRCh38
rs79572731 Chr.4: 183495323-183495323 on Build GRCh38
rs570239570 Chr.4: 183495273-183495273 on Build GRCh38
rs1012150249 Chr.4: 183495160-183495161 on Build GRCh38
rs966137555 Chr.4: 183495209-183495209 on Build GRCh38

View Full Product Details