Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 6827755-6828261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR4278
Entrez Gene ID
100422999
Gene Name
microRNA 4278
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.5: 6827853-6827921 on Build GRCh38
Transcripts
NR_036242.1

SNP Detail

SNP ID Chromosome Location
rs566963241 Chr.5: 6827943-6827943 on Build GRCh38
rs759881719 Chr.5: 6827922-6827922 on Build GRCh38
rs535530982 Chr.5: 6827956-6827956 on Build GRCh38
rs57914403 Chr.5: 6827850-6827850 on Build GRCh38
rs770377233 Chr.5: 6827838-6827838 on Build GRCh38
rs551279521 Chr.5: 6827910-6827910 on Build GRCh38
rs1008767831 Chr.5: 6828217-6828217 on Build GRCh38
rs763591108 Chr.5: 6827965-6827965 on Build GRCh38
rs955460468 Chr.5: 6827913-6827913 on Build GRCh38
rs556819159 Chr.5: 6828073-6828073 on Build GRCh38
rs1001225683 Chr.5: 6827858-6827858 on Build GRCh38
rs1030177385 Chr.5: 6828201-6828201 on Build GRCh38
rs1051816044 Chr.5: 6828120-6828120 on Build GRCh38
rs961702262 Chr.5: 6828230-6828230 on Build GRCh38
rs77813241 Chr.5: 6827833-6827833 on Build GRCh38
rs546816547 Chr.5: 6827942-6827942 on Build GRCh38
rs148744018 Chr.5: 6827935-6827935 on Build GRCh38
rs958900727 Chr.5: 6827959-6827959 on Build GRCh38
rs62351789 Chr.5: 6828169-6828169 on Build GRCh38
rs916944994 Chr.5: 6827888-6827888 on Build GRCh38
rs764997852 Chr.5: 6827861-6827861 on Build GRCh38
rs1038527624 Chr.5: 6827815-6827815 on Build GRCh38
rs923826292 Chr.5: 6828071-6828071 on Build GRCh38
rs955433055 Chr.5: 6828203-6828203 on Build GRCh38
rs1036305842 Chr.5: 6828185-6828185 on Build GRCh38
rs147461035 Chr.5: 6828220-6828220 on Build GRCh38
rs543721539 Chr.5: 6828136-6828137 on Build GRCh38
rs548774996 Chr.5: 6827824-6827824 on Build GRCh38
rs937742496 Chr.5: 6827832-6827832 on Build GRCh38
rs144590083 Chr.5: 6828050-6828050 on Build GRCh38
rs989421268 Chr.5: 6828091-6828091 on Build GRCh38
rs1052321291 Chr.5: 6827875-6827875 on Build GRCh38
rs62351788 Chr.5: 6827966-6827966 on Build GRCh38
rs902219077 Chr.5: 6828074-6828074 on Build GRCh38
rs370508169 Chr.5: 6828141-6828142 on Build GRCh38
rs1042452604 Chr.5: 6827846-6827846 on Build GRCh38
rs766738335 Chr.5: 6827883-6827883 on Build GRCh38
rs559047338 Chr.5: 6828173-6828173 on Build GRCh38
rs750201452 Chr.5: 6827880-6827880 on Build GRCh38
rs921760224 Chr.5: 6828102-6828104 on Build GRCh38
rs10039082 Chr.5: 6828237-6828237 on Build GRCh38
rs191393395 Chr.5: 6827919-6827919 on Build GRCh38
rs374082445 Chr.5: 6827805-6827805 on Build GRCh38
rs275440 Chr.5: 6828116-6828116 on Build GRCh38
rs908570471 Chr.5: 6827936-6827936 on Build GRCh38
rs949147964 Chr.5: 6828055-6828055 on Build GRCh38
rs553931684 Chr.5: 6827837-6827837 on Build GRCh38
rs763340974 Chr.5: 6827881-6827881 on Build GRCh38
rs778862056 Chr.5: 6828164-6828164 on Build GRCh38
rs1054142611 Chr.5: 6828159-6828159 on Build GRCh38
rs961647651 Chr.5: 6827941-6827941 on Build GRCh38
rs762193678 Chr.5: 6827819-6827819 on Build GRCh38
rs908754173 Chr.5: 6827801-6827801 on Build GRCh38

View Full Product Details