Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 9712203-9712735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC285692
Entrez Gene ID
285692
Gene Name
uncharacterized LOC285692
Gene Aliases
-
UniGene
Hs.651487
Gene Chromosome Location
Chr.5: 9641315-9903824 on Build GRCh38
Transcripts
NR_027112.2

SNP Detail

SNP ID Chromosome Location
rs757551851 Chr.5: 9712692-9712692 on Build GRCh38
rs973641405 Chr.5: 9712562-9712562 on Build GRCh38
rs1032767519 Chr.5: 9712474-9712474 on Build GRCh38
rs916358803 Chr.5: 9712632-9712632 on Build GRCh38
rs893802268 Chr.5: 9712489-9712489 on Build GRCh38
rs557997075 Chr.5: 9712651-9712651 on Build GRCh38
rs778543458 Chr.5: 9712233-9712233 on Build GRCh38
rs949853161 Chr.5: 9712286-9712286 on Build GRCh38
rs531686374 Chr.5: 9712304-9712304 on Build GRCh38
rs550457679 Chr.5: 9712654-9712654 on Build GRCh38
rs542421099 Chr.5: 9712279-9712279 on Build GRCh38
rs868243177 Chr.5: 9712371-9712371 on Build GRCh38
rs987636017 Chr.5: 9712515-9712515 on Build GRCh38
rs924570559 Chr.5: 9712355-9712355 on Build GRCh38
rs142080053 Chr.5: 9712413-9712413 on Build GRCh38
rs549351590 Chr.5: 9712622-9712622 on Build GRCh38
rs999443890 Chr.5: 9712437-9712437 on Build GRCh38
rs549362572 Chr.5: 9712305-9712305 on Build GRCh38
rs894995017 Chr.5: 9712327-9712327 on Build GRCh38
rs971531647 Chr.5: 9712664-9712664 on Build GRCh38
rs1046245730 Chr.5: 9712405-9712405 on Build GRCh38
rs935968016 Chr.5: 9712708-9712708 on Build GRCh38
rs551681211 Chr.5: 9712346-9712346 on Build GRCh38
rs1050451358 Chr.5: 9712506-9712506 on Build GRCh38
rs1027364655 Chr.5: 9712627-9712627 on Build GRCh38
rs564906025 Chr.5: 9712412-9712412 on Build GRCh38
rs991267690 Chr.5: 9712648-9712648 on Build GRCh38
rs977284783 Chr.5: 9712689-9712689 on Build GRCh38
rs567201027 Chr.5: 9712551-9712551 on Build GRCh38
rs1029061156 Chr.5: 9712495-9712495 on Build GRCh38
rs867791885 Chr.5: 9712298-9712298 on Build GRCh38
rs754314339 Chr.5: 9712493-9712493 on Build GRCh38
rs561354970 Chr.5: 9712362-9712362 on Build GRCh38
rs1009080586 Chr.5: 9712591-9712591 on Build GRCh38
rs1015481028 Chr.5: 9712592-9712592 on Build GRCh38
rs991236458 Chr.5: 9712226-9712226 on Build GRCh38
rs949113561 Chr.5: 9712636-9712636 on Build GRCh38
rs1013455767 Chr.5: 9712378-9712378 on Build GRCh38
rs1053800364 Chr.5: 9712431-9712431 on Build GRCh38
rs375876315 Chr.5: 9712644-9712644 on Build GRCh38
rs538173595 Chr.5: 9712649-9712649 on Build GRCh38
rs867378812 Chr.5: 9712621-9712621 on Build GRCh38
rs919568266 Chr.5: 9712246-9712246 on Build GRCh38
rs138657353 Chr.5: 9712288-9712288 on Build GRCh38
rs890651700 Chr.5: 9712528-9712528 on Build GRCh38
rs146331403 Chr.5: 9712554-9712554 on Build GRCh38
rs547489279 Chr.5: 9712548-9712548 on Build GRCh38
rs917113222 Chr.5: 9712235-9712235 on Build GRCh38

View Full Product Details