Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 51373551-51374086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC642366
Entrez Gene ID
642366
Gene Name
uncharacterized LOC642366
Gene Aliases
-
UniGene
Hs.544139
Gene Chromosome Location
Chr.5: 51372736-51383332 on Build GRCh38
Transcripts
NR_046243.1

SNP Detail

SNP ID Chromosome Location
rs1010357822 Chr.5: 51374054-51374054 on Build GRCh38
rs992914672 Chr.5: 51373888-51373888 on Build GRCh38
rs988714446 Chr.5: 51373584-51373584 on Build GRCh38
rs150507042 Chr.5: 51373636-51373636 on Build GRCh38
rs1050035306 Chr.5: 51373989-51373989 on Build GRCh38
rs147406754 Chr.5: 51373868-51373868 on Build GRCh38
rs920137142 Chr.5: 51373687-51373687 on Build GRCh38
rs911201601 Chr.5: 51373994-51373994 on Build GRCh38
rs569525251 Chr.5: 51373969-51373969 on Build GRCh38
rs779780475 Chr.5: 51374024-51374024 on Build GRCh38
rs77709439 Chr.5: 51374050-51374050 on Build GRCh38
rs183686289 Chr.5: 51374053-51374053 on Build GRCh38
rs1041164483 Chr.5: 51373765-51373766 on Build GRCh38
rs972840369 Chr.5: 51373931-51373931 on Build GRCh38
rs758809126 Chr.5: 51373667-51373667 on Build GRCh38
rs778080701 Chr.5: 51373723-51373723 on Build GRCh38
rs80351662 Chr.5: 51373610-51373610 on Build GRCh38
rs373022574 Chr.5: 51373748-51373748 on Build GRCh38
rs907605204 Chr.5: 51373904-51373904 on Build GRCh38
rs968642748 Chr.5: 51373604-51373604 on Build GRCh38
rs944024438 Chr.5: 51373998-51373998 on Build GRCh38
rs1037178286 Chr.5: 51373673-51373673 on Build GRCh38
rs527367355 Chr.5: 51373714-51373714 on Build GRCh38
rs889835504 Chr.5: 51373747-51373747 on Build GRCh38
rs780830962 Chr.5: 51373843-51373843 on Build GRCh38
rs780819097 Chr.5: 51373807-51373807 on Build GRCh38
rs73095531 Chr.5: 51373877-51373877 on Build GRCh38
rs533903260 Chr.5: 51373599-51373599 on Build GRCh38
rs920129082 Chr.5: 51373966-51373966 on Build GRCh38
rs748980233 Chr.5: 51374043-51374043 on Build GRCh38
rs931574915 Chr.5: 51373688-51373688 on Build GRCh38
rs71618116 Chr.5: 51373648-51373648 on Build GRCh38
rs769293284 Chr.5: 51373923-51373923 on Build GRCh38
rs561656464 Chr.5: 51373672-51373672 on Build GRCh38
rs114857048 Chr.5: 51373600-51373600 on Build GRCh38
rs774198417 Chr.5: 51374056-51374056 on Build GRCh38
rs757901366 Chr.5: 51373808-51373808 on Build GRCh38
rs1050185975 Chr.5: 51373721-51373721 on Build GRCh38
rs968614921 Chr.5: 51373821-51373821 on Build GRCh38
rs142842904 Chr.5: 51373754-51373754 on Build GRCh38
rs959762595 Chr.5: 51373887-51373887 on Build GRCh38
rs757282576 Chr.5: 51373654-51373654 on Build GRCh38
rs747517661 Chr.5: 51373731-51373731 on Build GRCh38
rs552554090 Chr.5: 51373878-51373878 on Build GRCh38
rs548163214 Chr.5: 51373665-51373665 on Build GRCh38

View Full Product Details