Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132331992-132332488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC553103
Entrez Gene ID
553103
Gene Name
uncharacterized LOC553103
Gene Aliases
-
UniGene
Hs.570908
Gene Chromosome Location
Chr.5: 132311276-132369916 on Build GRCh38
Transcripts
NR_110997.1 NM_003059.2 XM_006714675.3 XM_011543589.2 XM_017009776.1

Gene Symbol
SLC22A4
Entrez Gene ID
6583
Gene Name
solute carrier family 22 member 4
Gene Aliases
OCTN1
UniGene
Hs.310591
Gene Chromosome Location
Chr.5: 132294384-132344206 on Build GRCh38
Transcripts
NR_110997.1 NM_003059.2 XM_006714675.3 XM_011543589.2 XM_017009776.1

SNP Detail

SNP ID Chromosome Location
rs1035566071 Chr.5: 132332400-132332400 on Build GRCh38
rs990904404 Chr.5: 132332213-132332213 on Build GRCh38
rs538665401 Chr.5: 132332445-132332445 on Build GRCh38
rs951325339 Chr.5: 132332056-132332056 on Build GRCh38
rs983203667 Chr.5: 132332088-132332088 on Build GRCh38
rs748110879 Chr.5: 132332422-132332422 on Build GRCh38
rs536604984 Chr.5: 132332197-132332197 on Build GRCh38
rs1047320969 Chr.5: 132332379-132332380 on Build GRCh38
rs964237999 Chr.5: 132332039-132332039 on Build GRCh38
rs371788718 Chr.5: 132332224-132332224 on Build GRCh38
rs1018844798 Chr.5: 132332018-132332018 on Build GRCh38
rs558129341 Chr.5: 132332111-132332111 on Build GRCh38
rs752806056 Chr.5: 132332372-132332372 on Build GRCh38
rs572347397 Chr.5: 132332413-132332413 on Build GRCh38
rs183794268 Chr.5: 132332139-132332139 on Build GRCh38
rs73265586 Chr.5: 132332465-132332465 on Build GRCh38
rs777959719 Chr.5: 132332419-132332419 on Build GRCh38
rs145884243 Chr.5: 132332373-132332373 on Build GRCh38
rs1051265447 Chr.5: 132332313-132332314 on Build GRCh38
rs898235127 Chr.5: 132332351-132332351 on Build GRCh38
rs189565374 Chr.5: 132332318-132332318 on Build GRCh38
rs999175659 Chr.5: 132332238-132332238 on Build GRCh38
rs1039488296 Chr.5: 132332329-132332329 on Build GRCh38
rs924245672 Chr.5: 132332291-132332291 on Build GRCh38
rs541323371 Chr.5: 132332428-132332428 on Build GRCh38
rs964165799 Chr.5: 132332297-132332297 on Build GRCh38
rs946945598 Chr.5: 132332243-132332243 on Build GRCh38
rs1008887397 Chr.5: 132332263-132332263 on Build GRCh38
rs960672034 Chr.5: 132332203-132332203 on Build GRCh38
rs149819316 Chr.5: 132332272-132332272 on Build GRCh38
rs372239198 Chr.5: 132332424-132332424 on Build GRCh38
rs939249110 Chr.5: 132332116-132332116 on Build GRCh38
rs886912031 Chr.5: 132332383-132332384 on Build GRCh38
rs149520979 Chr.5: 132332379-132332379 on Build GRCh38
rs556934466 Chr.5: 132332221-132332221 on Build GRCh38
rs947929265 Chr.5: 132332150-132332150 on Build GRCh38
rs1056415831 Chr.5: 132332133-132332133 on Build GRCh38
rs903621050 Chr.5: 132332192-132332192 on Build GRCh38
rs756375138 Chr.5: 132332107-132332107 on Build GRCh38
rs272885 Chr.5: 132332043-132332043 on Build GRCh38
rs890825051 Chr.5: 132332256-132332256 on Build GRCh38
rs1002808859 Chr.5: 132332389-132332389 on Build GRCh38
rs576608043 Chr.5: 132332052-132332052 on Build GRCh38
rs564398232 Chr.5: 132332451-132332451 on Build GRCh38
rs1043419035 Chr.5: 132332161-132332161 on Build GRCh38
rs879579219 Chr.5: 132332313-132332313 on Build GRCh38
rs780385255 Chr.5: 132332241-132332241 on Build GRCh38
rs753709542 Chr.5: 132332317-132332317 on Build GRCh38
rs974151138 Chr.5: 132332040-132332040 on Build GRCh38
rs979514842 Chr.5: 132332429-132332429 on Build GRCh38

View Full Product Details