Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 6346049-6346571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LY86-AS1
Entrez Gene ID
285780
Gene Name
LY86 antisense RNA 1
Gene Aliases
LY86-AS, LY86AS
UniGene
Hs.655737
Gene Chromosome Location
Chr.6: 6346465-6622826 on Build GRCh38
Transcripts
NR_026970.1

SNP Detail

SNP ID Chromosome Location
rs746232823 Chr.6: 6346546-6346546 on Build GRCh38
rs796818050 Chr.6: 6346099-6346099 on Build GRCh38
rs1013795267 Chr.6: 6346362-6346362 on Build GRCh38
rs998830444 Chr.6: 6346184-6346184 on Build GRCh38
rs936531433 Chr.6: 6346222-6346222 on Build GRCh38
rs750251286 Chr.6: 6346327-6346327 on Build GRCh38
rs1048176580 Chr.6: 6346307-6346307 on Build GRCh38
rs566142792 Chr.6: 6346454-6346454 on Build GRCh38
rs867076561 Chr.6: 6346111-6346111 on Build GRCh38
rs550378036 Chr.6: 6346359-6346359 on Build GRCh38
rs764362982 Chr.6: 6346201-6346201 on Build GRCh38
rs943895538 Chr.6: 6346451-6346451 on Build GRCh38
rs780932348 Chr.6: 6346105-6346117 on Build GRCh38
rs182916493 Chr.6: 6346485-6346485 on Build GRCh38
rs565122349 Chr.6: 6346405-6346405 on Build GRCh38
rs908233507 Chr.6: 6346382-6346382 on Build GRCh38
rs866396706 Chr.6: 6346109-6346109 on Build GRCh38
rs925102784 Chr.6: 6346213-6346213 on Build GRCh38
rs1032454540 Chr.6: 6346410-6346410 on Build GRCh38
rs567579039 Chr.6: 6346527-6346527 on Build GRCh38
rs561590849 Chr.6: 6346259-6346259 on Build GRCh38
rs867659618 Chr.6: 6346093-6346093 on Build GRCh38
rs969449560 Chr.6: 6346404-6346404 on Build GRCh38
rs899498589 Chr.6: 6346304-6346304 on Build GRCh38
rs913850542 Chr.6: 6346253-6346253 on Build GRCh38
rs549029305 Chr.6: 6346493-6346493 on Build GRCh38
rs1024349418 Chr.6: 6346106-6346121 on Build GRCh38
rs564198451 Chr.6: 6346438-6346438 on Build GRCh38
rs9392768 Chr.6: 6346105-6346105 on Build GRCh38
rs1040707684 Chr.6: 6346290-6346290 on Build GRCh38
rs1048599695 Chr.6: 6346524-6346524 on Build GRCh38
rs6901377 Chr.6: 6346529-6346529 on Build GRCh38
rs932357562 Chr.6: 6346305-6346305 on Build GRCh38
rs373821304 Chr.6: 6346425-6346425 on Build GRCh38
rs980888221 Chr.6: 6346197-6346197 on Build GRCh38
rs12192554 Chr.6: 6346171-6346171 on Build GRCh38
rs761254162 Chr.6: 6346093-6346093 on Build GRCh38
rs976701867 Chr.6: 6346456-6346456 on Build GRCh38
rs988040577 Chr.6: 6346225-6346225 on Build GRCh38
rs371722485 Chr.6: 6346496-6346496 on Build GRCh38
rs1015115688 Chr.6: 6346346-6346346 on Build GRCh38
rs1031604723 Chr.6: 6346279-6346279 on Build GRCh38
rs969839021 Chr.6: 6346193-6346193 on Build GRCh38
rs943947818 Chr.6: 6346260-6346260 on Build GRCh38
rs1006367871 Chr.6: 6346333-6346333 on Build GRCh38
rs191123921 Chr.6: 6346430-6346430 on Build GRCh38
rs1025362579 Chr.6: 6346385-6346385 on Build GRCh38
rs957586565 Chr.6: 6346297-6346297 on Build GRCh38
rs541780502 Chr.6: 6346105-6346116 on Build GRCh38
rs1024994420 Chr.6: 6346183-6346183 on Build GRCh38
rs531829911 Chr.6: 6346334-6346334 on Build GRCh38
rs9504769 Chr.6: 6346087-6346087 on Build GRCh38
rs964916498 Chr.6: 6346332-6346332 on Build GRCh38
rs796533262 Chr.6: 6346088-6346093 on Build GRCh38
rs9504768 Chr.6: 6346086-6346086 on Build GRCh38
rs920918809 Chr.6: 6346470-6346470 on Build GRCh38
rs754565447 Chr.6: 6346540-6346540 on Build GRCh38

View Full Product Details