Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31462918-31463403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCP5
Entrez Gene ID
10866
Gene Name
HLA complex P5 (non-protein coding)
Gene Aliases
6S2650E, D6S2650E, P5-1
UniGene
Hs.691948
Gene Chromosome Location
Chr.6: 31463180-31465809 on Build GRCh38
Transcripts
NR_040662.1

SNP Detail

SNP ID Chromosome Location
rs572140230 Chr.6: 31463074-31463074 on Build GRCh38
rs928396346 Chr.6: 31463127-31463127 on Build GRCh38
rs955270022 Chr.6: 31462971-31462971 on Build GRCh38
rs45533135 Chr.6: 31463174-31463174 on Build GRCh38
rs1020160144 Chr.6: 31462950-31462950 on Build GRCh38
rs751101195 Chr.6: 31463004-31463005 on Build GRCh38
rs75425619 Chr.6: 31463035-31463035 on Build GRCh38
rs999506753 Chr.6: 31463195-31463195 on Build GRCh38
rs528002799 Chr.6: 31463224-31463224 on Build GRCh38
rs572791517 Chr.6: 31463014-31463014 on Build GRCh38
rs1018888741 Chr.6: 31463211-31463211 on Build GRCh38
rs892962270 Chr.6: 31463166-31463166 on Build GRCh38
rs999475672 Chr.6: 31462970-31462970 on Build GRCh38
rs373729297 Chr.6: 31463254-31463254 on Build GRCh38
rs185441393 Chr.6: 31462982-31462982 on Build GRCh38
rs973973611 Chr.6: 31463047-31463047 on Build GRCh38
rs762335695 Chr.6: 31463083-31463083 on Build GRCh38
rs945374017 Chr.6: 31463150-31463150 on Build GRCh38
rs545617611 Chr.6: 31463144-31463144 on Build GRCh38
rs544395859 Chr.6: 31463072-31463072 on Build GRCh38
rs578132870 Chr.6: 31463114-31463114 on Build GRCh38
rs573951487 Chr.6: 31463038-31463038 on Build GRCh38
rs1043000110 Chr.6: 31463155-31463155 on Build GRCh38
rs565814705 Chr.6: 31463358-31463358 on Build GRCh38
rs568929585 Chr.6: 31463275-31463275 on Build GRCh38
rs552547672 Chr.6: 31463113-31463113 on Build GRCh38
rs975041576 Chr.6: 31462960-31462960 on Build GRCh38
rs752456536 Chr.6: 31463355-31463355 on Build GRCh38
rs937085305 Chr.6: 31463140-31463140 on Build GRCh38
rs3099840 Chr.6: 31462944-31462944 on Build GRCh38
rs3132090 Chr.6: 31462975-31462975 on Build GRCh38
rs937846054 Chr.6: 31463349-31463349 on Build GRCh38
rs902530449 Chr.6: 31462962-31462962 on Build GRCh38
rs549668248 Chr.6: 31463229-31463229 on Build GRCh38
rs767301403 Chr.6: 31463343-31463343 on Build GRCh38
rs769468959 Chr.6: 31463172-31463172 on Build GRCh38
rs528862871 Chr.6: 31463253-31463253 on Build GRCh38
rs77803816 Chr.6: 31463286-31463286 on Build GRCh38
rs1055524705 Chr.6: 31463143-31463143 on Build GRCh38
rs6905676 Chr.6: 31463231-31463231 on Build GRCh38
rs189844075 Chr.6: 31463307-31463307 on Build GRCh38
rs903106060 Chr.6: 31463190-31463190 on Build GRCh38
rs974302581 Chr.6: 31463320-31463320 on Build GRCh38
rs1029040492 Chr.6: 31463247-31463247 on Build GRCh38
rs904106482 Chr.6: 31463158-31463158 on Build GRCh38
rs560829113 Chr.6: 31463167-31463167 on Build GRCh38
rs1001437142 Chr.6: 31463162-31463162 on Build GRCh38
rs941153342 Chr.6: 31463029-31463029 on Build GRCh38
rs781452345 Chr.6: 31463021-31463021 on Build GRCh38
rs2518030 Chr.6: 31463305-31463305 on Build GRCh38
rs6928071 Chr.6: 31463112-31463112 on Build GRCh38
rs762488441 Chr.6: 31463198-31463198 on Build GRCh38
rs896465444 Chr.6: 31463116-31463116 on Build GRCh38

View Full Product Details