Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31835104-31835535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf48
Entrez Gene ID
50854
Gene Name
chromosome 6 open reading frame 48
Gene Aliases
D6S57, G8
UniGene
Hs.640836
Gene Chromosome Location
Chr.6: 31834915-31839766 on Build GRCh38
Transcripts
NM_001040437.2 NM_001040438.2 NM_001287482.1 NM_001287483.1 NM_001287484.1 NM_001287485.1 NM_001287486.1 NM_001287487.1 NM_001287488.1 NR_002745.1

Gene Symbol
SNORD48
Entrez Gene ID
26801
Gene Name
small nucleolar RNA, C/D box 48
Gene Aliases
RNU48, U48
UniGene
Gene Chromosome Location
Chr.6: 31835263-31835326 on Build GRCh38
Transcripts
NM_001040437.2 NM_001040438.2 NM_001287482.1 NM_001287483.1 NM_001287484.1 NM_001287485.1 NM_001287486.1 NM_001287487.1 NM_001287488.1 NR_002745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792565 c.199G>A p.G67S Chr.6: 31835148-31835148 on Build GRCh38
164506 c.212C>T p.S71L Chr.6: 31835161-31835161 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs12206600 Chr.6: 31835486-31835486 on Build GRCh38
rs774850186 Chr.6: 31835364-31835364 on Build GRCh38
rs571492964 Chr.6: 31835270-31835270 on Build GRCh38
rs555120372 Chr.6: 31835383-31835383 on Build GRCh38
rs17201241 Chr.6: 31835297-31835297 on Build GRCh38
rs774666898 Chr.6: 31835427-31835427 on Build GRCh38
rs993097316 Chr.6: 31835373-31835373 on Build GRCh38
rs972497685 Chr.6: 31835158-31835158 on Build GRCh38
rs745475110 Chr.6: 31835405-31835405 on Build GRCh38
rs559553811 Chr.6: 31835423-31835423 on Build GRCh38
rs1007662283 Chr.6: 31835142-31835142 on Build GRCh38
rs758774103 Chr.6: 31835152-31835152 on Build GRCh38
rs577254705 Chr.6: 31835334-31835334 on Build GRCh38
rs144137493 Chr.6: 31835433-31835433 on Build GRCh38
rs147324511 Chr.6: 31835512-31835512 on Build GRCh38
rs541203586 Chr.6: 31835411-31835411 on Build GRCh38
rs770466619 Chr.6: 31835247-31835247 on Build GRCh38
rs17201248 Chr.6: 31835353-31835353 on Build GRCh38
rs777671614 Chr.6: 31835327-31835327 on Build GRCh38
rs759109723 Chr.6: 31835252-31835252 on Build GRCh38
rs765871203 Chr.6: 31835134-31835134 on Build GRCh38
rs577513583 Chr.6: 31835402-31835402 on Build GRCh38
rs752588668 Chr.6: 31835315-31835315 on Build GRCh38
rs751605959 Chr.6: 31835287-31835291 on Build GRCh38
rs769360916 Chr.6: 31835349-31835349 on Build GRCh38
rs185899920 Chr.6: 31835391-31835391 on Build GRCh38
rs933630716 Chr.6: 31835484-31835484 on Build GRCh38
rs548554139 Chr.6: 31835163-31835163 on Build GRCh38
rs796125280 Chr.6: 31835288-31835288 on Build GRCh38
rs751384803 Chr.6: 31835456-31835456 on Build GRCh38
rs764106272 Chr.6: 31835446-31835446 on Build GRCh38
rs763298451 Chr.6: 31835286-31835286 on Build GRCh38
rs762071179 Chr.6: 31835365-31835365 on Build GRCh38
rs940399898 Chr.6: 31835143-31835143 on Build GRCh38
rs937102933 Chr.6: 31835392-31835392 on Build GRCh38
rs780421821 Chr.6: 31835384-31835384 on Build GRCh38
rs576337048 Chr.6: 31835386-31835386 on Build GRCh38
rs777629896 Chr.6: 31835208-31835208 on Build GRCh38
rs113957351 Chr.6: 31835410-31835410 on Build GRCh38
rs916082442 Chr.6: 31835222-31835222 on Build GRCh38
rs751851149 Chr.6: 31835295-31835295 on Build GRCh38
rs776834227 Chr.6: 31835442-31835442 on Build GRCh38
rs750578446 Chr.6: 31835370-31835370 on Build GRCh38
rs746750160 Chr.6: 31835329-31835329 on Build GRCh38
rs114916638 Chr.6: 31835369-31835369 on Build GRCh38
rs1043218940 Chr.6: 31835262-31835262 on Build GRCh38
rs17201234 Chr.6: 31835180-31835180 on Build GRCh38
rs767403097 Chr.6: 31835255-31835258 on Build GRCh38
rs776321751 Chr.6: 31835248-31835248 on Build GRCh38
rs767725675 Chr.6: 31835311-31835311 on Build GRCh38
rs112576236 Chr.6: 31835409-31835409 on Build GRCh38
rs974545662 Chr.6: 31835298-31835298 on Build GRCh38
rs748554135 Chr.6: 31835429-31835429 on Build GRCh38
rs111432088 Chr.6: 31835161-31835161 on Build GRCh38
rs760776561 Chr.6: 31835375-31835375 on Build GRCh38
rs777299852 Chr.6: 31835136-31835136 on Build GRCh38
rs776080060 Chr.6: 31835275-31835275 on Build GRCh38
rs896040222 Chr.6: 31835205-31835205 on Build GRCh38
rs769016983 Chr.6: 31835358-31835358 on Build GRCh38
rs751768732 Chr.6: 31835153-31835153 on Build GRCh38
rs767905525 Chr.6: 31835305-31835307 on Build GRCh38
rs746622647 Chr.6: 31835238-31835238 on Build GRCh38
rs759520914 Chr.6: 31835443-31835443 on Build GRCh38
rs757147375 Chr.6: 31835463-31835463 on Build GRCh38
rs751424945 Chr.6: 31835397-31835397 on Build GRCh38
rs775804492 Chr.6: 31835285-31835287 on Build GRCh38
rs773442620 Chr.6: 31835431-31835431 on Build GRCh38
rs1023192469 Chr.6: 31835377-31835377 on Build GRCh38
rs760170087 Chr.6: 31835139-31835139 on Build GRCh38
rs948840432 Chr.6: 31835234-31835234 on Build GRCh38
rs748743527 Chr.6: 31835422-31835422 on Build GRCh38
rs1016748288 Chr.6: 31835347-31835347 on Build GRCh38
rs745920645 Chr.6: 31835171-31835171 on Build GRCh38
rs750972490 Chr.6: 31835322-31835322 on Build GRCh38
rs780994311 Chr.6: 31835403-31835403 on Build GRCh38
rs764473328 Chr.6: 31835288-31835288 on Build GRCh38
rs900813248 Chr.6: 31835299-31835299 on Build GRCh38
rs760933745 Chr.6: 31835432-31835432 on Build GRCh38
rs780974325 Chr.6: 31835339-31835339 on Build GRCh38
rs530252931 Chr.6: 31835466-31835466 on Build GRCh38
rs771836221 Chr.6: 31835185-31835185 on Build GRCh38
rs765804875 Chr.6: 31835145-31835145 on Build GRCh38
rs149369565 Chr.6: 31835132-31835132 on Build GRCh38
rs758149156 Chr.6: 31835319-31835319 on Build GRCh38
rs753313365 Chr.6: 31835390-31835391 on Build GRCh38
rs753175799 Chr.6: 31835150-31835150 on Build GRCh38
rs778810049 Chr.6: 31835173-31835173 on Build GRCh38
rs543459963 Chr.6: 31835390-31835390 on Build GRCh38
rs988538911 Chr.6: 31835438-31835438 on Build GRCh38
rs757417356 Chr.6: 31835291-31835297 on Build GRCh38
rs536712377 Chr.6: 31835382-31835382 on Build GRCh38
rs1014872602 Chr.6: 31835426-31835426 on Build GRCh38
rs998060807 Chr.6: 31835305-31835305 on Build GRCh38
rs747933428 Chr.6: 31835128-31835128 on Build GRCh38
rs745381122 Chr.6: 31835345-31835345 on Build GRCh38
rs922291210 Chr.6: 31835462-31835462 on Build GRCh38
rs993588932 Chr.6: 31835357-31835357 on Build GRCh38
rs1000185134 Chr.6: 31835467-31835467 on Build GRCh38
rs756452232 Chr.6: 31835338-31835338 on Build GRCh38
rs550009182 Chr.6: 31835253-31835253 on Build GRCh38
rs1049609535 Chr.6: 31835326-31835326 on Build GRCh38
rs756131149 Chr.6: 31835314-31835314 on Build GRCh38
rs760105482 Chr.6: 31835133-31835133 on Build GRCh38
rs369060498 Chr.6: 31835328-31835328 on Build GRCh38
rs34103657 Chr.6: 31835287-31835287 on Build GRCh38
rs571105497 Chr.6: 31835416-31835416 on Build GRCh38
rs771680819 Chr.6: 31835130-31835130 on Build GRCh38
rs567227239 Chr.6: 31835172-31835172 on Build GRCh38

View Full Product Details